Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 138191384-138191889 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC23
Entrez Gene ID
8697
Gene Name
cell division cycle 23
Gene Aliases
ANAPC8, APC8, CUT23
UniGene
Hs.73625
Gene Chromosome Location
Chr.5: 138187648-138213343 on Build GRCh38
Transcripts
NM_004661.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
236073 c.1384G>A p.V462M Chr.5: 138191514-138191514 on Build GRCh38
3728882 c.1363-6_1363-5insT p.? Chr.5: 138191540-138191541 on Build GRCh38
5356315 c.1363-6delT p.? Chr.5: 138191541-138191541 on Build GRCh38
1247746 c.1340_1341insA p.N447fs*32 Chr.5: 138191883-138191884 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs758282291 Chr.5: 138191570-138191570 on Build GRCh38
rs763956817 Chr.5: 138191576-138191576 on Build GRCh38
rs890559077 Chr.5: 138191440-138191440 on Build GRCh38
rs1051033662 Chr.5: 138191609-138191609 on Build GRCh38
rs746752090 Chr.5: 138191433-138191433 on Build GRCh38
rs745476402 Chr.5: 138191832-138191832 on Build GRCh38
rs959804412 Chr.5: 138191739-138191739 on Build GRCh38
rs561418205 Chr.5: 138191640-138191640 on Build GRCh38
rs905542569 Chr.5: 138191770-138191770 on Build GRCh38
rs776979044 Chr.5: 138191528-138191528 on Build GRCh38
rs541685032 Chr.5: 138191506-138191506 on Build GRCh38
rs777568233 Chr.5: 138191424-138191424 on Build GRCh38
rs371790062 Chr.5: 138191820-138191820 on Build GRCh38
rs921884164 Chr.5: 138191806-138191806 on Build GRCh38
rs761053901 Chr.5: 138191514-138191514 on Build GRCh38
rs780875301 Chr.5: 138191818-138191818 on Build GRCh38
rs769162155 Chr.5: 138191461-138191461 on Build GRCh38
rs995261114 Chr.5: 138191720-138191720 on Build GRCh38
rs751426683 Chr.5: 138191577-138191577 on Build GRCh38
rs576500803 Chr.5: 138191699-138191699 on Build GRCh38
rs543833153 Chr.5: 138191763-138191763 on Build GRCh38
rs752774053 Chr.5: 138191554-138191554 on Build GRCh38
rs969928871 Chr.5: 138191704-138191704 on Build GRCh38
rs765266755 Chr.5: 138191543-138191543 on Build GRCh38
rs947074647 Chr.5: 138191767-138191767 on Build GRCh38
rs1023894961 Chr.5: 138191686-138191686 on Build GRCh38
rs1035418233 Chr.5: 138191719-138191719 on Build GRCh38
rs768900826 Chr.5: 138191706-138191706 on Build GRCh38
rs779553359 Chr.5: 138191487-138191487 on Build GRCh38
rs748629037 Chr.5: 138191476-138191476 on Build GRCh38
rs773724901 Chr.5: 138191510-138191510 on Build GRCh38
rs756911699 Chr.5: 138191437-138191437 on Build GRCh38
rs774960468 Chr.5: 138191465-138191465 on Build GRCh38
rs370700895 Chr.5: 138191448-138191448 on Build GRCh38
rs367727615 Chr.5: 138191450-138191450 on Build GRCh38
rs371113139 Chr.5: 138191765-138191765 on Build GRCh38
rs1014907856 Chr.5: 138191565-138191565 on Build GRCh38
rs765489921 Chr.5: 138191791-138191791 on Build GRCh38
rs1004823002 Chr.5: 138191508-138191508 on Build GRCh38
rs766772517 Chr.5: 138191515-138191515 on Build GRCh38
rs931842152 Chr.5: 138191407-138191407 on Build GRCh38
rs1051769651 Chr.5: 138191423-138191423 on Build GRCh38
rs759704014 Chr.5: 138191530-138191530 on Build GRCh38
rs757036115 Chr.5: 138191580-138191580 on Build GRCh38
rs113013172 Chr.5: 138191748-138191748 on Build GRCh38
rs755749198 Chr.5: 138191851-138191851 on Build GRCh38
rs17228526 Chr.5: 138191408-138191408 on Build GRCh38
rs982292575 Chr.5: 138191715-138191715 on Build GRCh38
rs979052034 Chr.5: 138191776-138191776 on Build GRCh38
rs899408737 Chr.5: 138191596-138191596 on Build GRCh38

View Full Product Details