Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 138337430-138337926 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC25C
Entrez Gene ID
995
Gene Name
cell division cycle 25C
Gene Aliases
CDC25, PPP1R60
UniGene
Hs.656
Gene Chromosome Location
Chr.5: 138285265-138338355 on Build GRCh38
Transcripts
NM_001287583.1 NM_001318098.1 XM_005272145.3 XM_006714739.3 XM_011543759.2 XM_011543760.2 XM_011543761.2 NM_001135647.1

Gene Symbol
FAM53C
Entrez Gene ID
51307
Gene Name
family with sequence similarity 53 member C
Gene Aliases
C5orf6
UniGene
Hs.54056
Gene Chromosome Location
Chr.5: 138337535-138349729 on Build GRCh38
Transcripts
NM_001287583.1 NM_001318098.1 XM_005272145.3 XM_006714739.3 XM_011543759.2 XM_011543760.2 XM_011543761.2 NM_001135647.1

SNP Detail

SNP ID Chromosome Location
rs897908460 Chr.5: 138337472-138337472 on Build GRCh38
rs1002276134 Chr.5: 138337722-138337722 on Build GRCh38
rs979465057 Chr.5: 138337545-138337545 on Build GRCh38
rs985794154 Chr.5: 138337861-138337861 on Build GRCh38
rs1013397920 Chr.5: 138337811-138337811 on Build GRCh38
rs554123884 Chr.5: 138337478-138337478 on Build GRCh38
rs944364228 Chr.5: 138337868-138337868 on Build GRCh38
rs978633665 Chr.5: 138337785-138337785 on Build GRCh38
rs1012341014 Chr.5: 138337895-138337907 on Build GRCh38
rs985851168 Chr.5: 138337896-138337896 on Build GRCh38
rs556955927 Chr.5: 138337701-138337701 on Build GRCh38
rs535482479 Chr.5: 138337664-138337664 on Build GRCh38
rs980758536 Chr.5: 138337488-138337488 on Build GRCh38
rs1015137909 Chr.5: 138337470-138337470 on Build GRCh38
rs925736303 Chr.5: 138337578-138337578 on Build GRCh38
rs959944567 Chr.5: 138337736-138337736 on Build GRCh38
rs918734444 Chr.5: 138337637-138337637 on Build GRCh38
rs978883339 Chr.5: 138337572-138337572 on Build GRCh38
rs954104466 Chr.5: 138337881-138337881 on Build GRCh38
rs755443438 Chr.5: 138337498-138337498 on Build GRCh38
rs17171800 Chr.5: 138337477-138337477 on Build GRCh38
rs1020764367 Chr.5: 138337817-138337817 on Build GRCh38
rs966749354 Chr.5: 138337531-138337531 on Build GRCh38
rs992610904 Chr.5: 138337543-138337543 on Build GRCh38
rs887454635 Chr.5: 138337704-138337704 on Build GRCh38
rs905300577 Chr.5: 138337715-138337715 on Build GRCh38
rs1032385043 Chr.5: 138337848-138337848 on Build GRCh38
rs554190550 Chr.5: 138337728-138337728 on Build GRCh38
rs1036994434 Chr.5: 138337869-138337869 on Build GRCh38
rs959975935 Chr.5: 138337542-138337543 on Build GRCh38
rs887412825 Chr.5: 138337476-138337476 on Build GRCh38
rs902318366 Chr.5: 138337823-138337823 on Build GRCh38
rs890706335 Chr.5: 138337656-138337656 on Build GRCh38
rs1048810036 Chr.5: 138337676-138337676 on Build GRCh38
rs1035017867 Chr.5: 138337734-138337734 on Build GRCh38
rs944981159 Chr.5: 138337662-138337662 on Build GRCh38
rs575532143 Chr.5: 138337711-138337711 on Build GRCh38
rs546131334 Chr.5: 138337746-138337746 on Build GRCh38
rs912798368 Chr.5: 138337580-138337580 on Build GRCh38
rs1037184324 Chr.5: 138337629-138337629 on Build GRCh38
rs867888398 Chr.5: 138337514-138337514 on Build GRCh38
rs954141532 Chr.5: 138337562-138337562 on Build GRCh38
rs183405931 Chr.5: 138337779-138337779 on Build GRCh38
rs3756766 Chr.5: 138337478-138337478 on Build GRCh38
rs897955401 Chr.5: 138337675-138337675 on Build GRCh38
rs1055086150 Chr.5: 138337755-138337755 on Build GRCh38
rs925292479 Chr.5: 138337555-138337555 on Build GRCh38
rs995030662 Chr.5: 138337700-138337700 on Build GRCh38
rs1014086126 Chr.5: 138337741-138337741 on Build GRCh38
rs559836449 Chr.5: 138337829-138337829 on Build GRCh38
rs879834262 Chr.5: 138337477-138337478 on Build GRCh38
rs1036621148 Chr.5: 138337668-138337668 on Build GRCh38
rs777318788 Chr.5: 138337819-138337819 on Build GRCh38
rs929053698 Chr.5: 138337663-138337663 on Build GRCh38
rs945901852 Chr.5: 138337567-138337567 on Build GRCh38
rs911523031 Chr.5: 138337865-138337865 on Build GRCh38
rs944248360 Chr.5: 138337597-138337597 on Build GRCh38
rs960612729 Chr.5: 138337479-138337479 on Build GRCh38
rs1002146652 Chr.5: 138337727-138337727 on Build GRCh38
rs1050118751 Chr.5: 138337616-138337616 on Build GRCh38
rs985768564 Chr.5: 138337577-138337577 on Build GRCh38
rs56366217 Chr.5: 138337826-138337826 on Build GRCh38
rs573076134 Chr.5: 138337805-138337805 on Build GRCh38

View Full Product Details