Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 138465400-138465892 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EGR1
Entrez Gene ID
1958
Gene Name
early growth response 1
Gene Aliases
AT225, G0S30, KROX-24, NGFI-A, TIS8, ZIF-268, ZNF225
UniGene
Hs.326035
Gene Chromosome Location
Chr.5: 138465492-138469315 on Build GRCh38
Transcripts
NM_001964.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6652326 c.71C>T p.P24L Chr.5: 138465832-138465832 on Build GRCh38
7114779 c.48C>T p.I16I Chr.5: 138465809-138465809 on Build GRCh38
3357855 c.103C>T p.L35L Chr.5: 138465864-138465864 on Build GRCh38
145362 c.108G>A p.E36E Chr.5: 138465870-138465870 on Build GRCh38
1486325 c.30G>A p.L10L Chr.5: 138465791-138465791 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs949771592 Chr.5: 138465457-138465457 on Build GRCh38
rs199632450 Chr.5: 138465674-138465674 on Build GRCh38
rs1039410421 Chr.5: 138465548-138465548 on Build GRCh38
rs199968159 Chr.5: 138465751-138465751 on Build GRCh38
rs770687242 Chr.5: 138465660-138465660 on Build GRCh38
rs747647165 Chr.5: 138465509-138465509 on Build GRCh38
rs751640347 Chr.5: 138465817-138465817 on Build GRCh38
rs558452520 Chr.5: 138465864-138465864 on Build GRCh38
rs766886557 Chr.5: 138465786-138465786 on Build GRCh38
rs773869377 Chr.5: 138465784-138465784 on Build GRCh38
rs1033547333 Chr.5: 138465637-138465637 on Build GRCh38
rs761175632 Chr.5: 138465719-138465719 on Build GRCh38
rs962909101 Chr.5: 138465672-138465672 on Build GRCh38
rs762639190 Chr.5: 138465762-138465762 on Build GRCh38
rs768374163 Chr.5: 138465846-138465846 on Build GRCh38
rs944180681 Chr.5: 138465468-138465469 on Build GRCh38
rs754190894 Chr.5: 138465789-138465789 on Build GRCh38
rs769637714 Chr.5: 138465760-138465760 on Build GRCh38
rs901835499 Chr.5: 138465555-138465555 on Build GRCh38
rs1054659794 Chr.5: 138465506-138465506 on Build GRCh38
rs76865845 Chr.5: 138465810-138465810 on Build GRCh38
rs752789997 Chr.5: 138465866-138465868 on Build GRCh38
rs777179772 Chr.5: 138465857-138465857 on Build GRCh38
rs966779941 Chr.5: 138465435-138465435 on Build GRCh38
rs1004424241 Chr.5: 138465599-138465599 on Build GRCh38
rs149170181 Chr.5: 138465837-138465837 on Build GRCh38
rs202023911 Chr.5: 138465785-138465785 on Build GRCh38
rs755238155 Chr.5: 138465721-138465721 on Build GRCh38
rs144720042 Chr.5: 138465520-138465521 on Build GRCh38
rs374447937 Chr.5: 138465831-138465831 on Build GRCh38
rs199735825 Chr.5: 138465724-138465724 on Build GRCh38
rs547507736 Chr.5: 138465832-138465832 on Build GRCh38
rs934814124 Chr.5: 138465477-138465477 on Build GRCh38
rs936884766 Chr.5: 138465455-138465455 on Build GRCh38
rs561524195 Chr.5: 138465754-138465754 on Build GRCh38
rs536129727 Chr.5: 138465448-138465448 on Build GRCh38
rs1042086 Chr.5: 138465688-138465688 on Build GRCh38
rs915582738 Chr.5: 138465438-138465438 on Build GRCh38
rs1048942238 Chr.5: 138465848-138465848 on Build GRCh38
rs993434907 Chr.5: 138465727-138465727 on Build GRCh38
rs13181973 Chr.5: 138465844-138465844 on Build GRCh38
rs745645418 Chr.5: 138465755-138465755 on Build GRCh38
rs547794631 Chr.5: 138465843-138465843 on Build GRCh38
rs765498015 Chr.5: 138465801-138465801 on Build GRCh38
rs543045841 Chr.5: 138465748-138465748 on Build GRCh38
rs578013231 Chr.5: 138465687-138465687 on Build GRCh38
rs6873141 Chr.5: 138465725-138465725 on Build GRCh38
rs911423338 Chr.5: 138465463-138465463 on Build GRCh38
rs924404803 Chr.5: 138465473-138465473 on Build GRCh38
rs199793033 Chr.5: 138465855-138465855 on Build GRCh38
rs758457385 Chr.5: 138465742-138465742 on Build GRCh38
rs751903243 Chr.5: 138465493-138465493 on Build GRCh38
rs536813492 Chr.5: 138465639-138465639 on Build GRCh38
rs759886974 Chr.5: 138465799-138465799 on Build GRCh38
rs888426996 Chr.5: 138465565-138465565 on Build GRCh38
rs28365161 Chr.5: 138465720-138465720 on Build GRCh38
rs554486758 Chr.5: 138465497-138465497 on Build GRCh38
rs764350788 Chr.5: 138465868-138465868 on Build GRCh38
rs866012383 Chr.5: 138465556-138465556 on Build GRCh38
rs142306373 Chr.5: 138465847-138465847 on Build GRCh38
rs200905400 Chr.5: 138465767-138465767 on Build GRCh38
rs1006278463 Chr.5: 138465627-138465627 on Build GRCh38
rs1016121257 Chr.5: 138465653-138465653 on Build GRCh38
rs201288838 Chr.5: 138465836-138465836 on Build GRCh38
rs895031869 Chr.5: 138465870-138465870 on Build GRCh38
rs777766500 Chr.5: 138465745-138465745 on Build GRCh38
rs575937299 Chr.5: 138465525-138465525 on Build GRCh38
rs770857123 Chr.5: 138465752-138465752 on Build GRCh38
rs759975233 Chr.5: 138465860-138465860 on Build GRCh38
rs770821515 Chr.5: 138465719-138465720 on Build GRCh38
rs759486333 Chr.5: 138465805-138465825 on Build GRCh38
rs199695186 Chr.5: 138465788-138465788 on Build GRCh38
rs977751330 Chr.5: 138465442-138465442 on Build GRCh38
rs1026383230 Chr.5: 138465561-138465561 on Build GRCh38
rs200490385 Chr.5: 138465711-138465711 on Build GRCh38
rs754255742 Chr.5: 138465436-138465436 on Build GRCh38
rs565500696 Chr.5: 138465815-138465815 on Build GRCh38
rs550077220 Chr.5: 138465804-138465804 on Build GRCh38
rs990682316 Chr.5: 138465787-138465787 on Build GRCh38
rs1025152422 Chr.5: 138465661-138465661 on Build GRCh38
rs886550961 Chr.5: 138465581-138465581 on Build GRCh38
rs1032030919 Chr.5: 138465713-138465713 on Build GRCh38
rs775269017 Chr.5: 138465761-138465761 on Build GRCh38
rs372611694 Chr.5: 138465866-138465866 on Build GRCh38
rs895021904 Chr.5: 138465657-138465657 on Build GRCh38
rs897200749 Chr.5: 138465517-138465517 on Build GRCh38
rs532091505 Chr.5: 138465820-138465820 on Build GRCh38
rs765297978 Chr.5: 138465864-138465864 on Build GRCh38
rs201629189 Chr.5: 138465729-138465729 on Build GRCh38
rs981149293 Chr.5: 138465462-138465462 on Build GRCh38
rs545513858 Chr.5: 138465714-138465714 on Build GRCh38
rs368477832 Chr.5: 138465863-138465863 on Build GRCh38
rs991540642 Chr.5: 138465430-138465430 on Build GRCh38
rs371476752 Chr.5: 138465805-138465805 on Build GRCh38
rs1049987164 Chr.5: 138465459-138465459 on Build GRCh38
rs768284319 Chr.5: 138465778-138465778 on Build GRCh38
rs563622226 Chr.5: 138465720-138465720 on Build GRCh38
rs978180357 Chr.5: 138465694-138465694 on Build GRCh38
rs962671055 Chr.5: 138465421-138465421 on Build GRCh38

View Full Product Details