Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 139393186-139393679 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PROB1
Entrez Gene ID
389333
Gene Name
proline rich basic protein 1
Gene Aliases
C5orf65
UniGene
Hs.729482
Gene Chromosome Location
Chr.5: 139391946-139395196 on Build GRCh38
Transcripts
NM_001161546.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6301634 c.1679C>T p.T560I Chr.5: 139393403-139393403 on Build GRCh38
5995886 c.1755A>T p.A585A Chr.5: 139393327-139393327 on Build GRCh38
4616019 c.1690_1691delAG p.P565fs*37 Chr.5: 139393391-139393392 on Build GRCh38
7385623 c.1829G>C p.G610A Chr.5: 139393253-139393253 on Build GRCh38
1197455 c.1751T>C p.V584A Chr.5: 139393331-139393331 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781078065 Chr.5: 139393404-139393404 on Build GRCh38
rs904131392 Chr.5: 139393635-139393635 on Build GRCh38
rs369710606 Chr.5: 139393368-139393368 on Build GRCh38
rs921030415 Chr.5: 139393211-139393211 on Build GRCh38
rs552624204 Chr.5: 139393381-139393381 on Build GRCh38
rs553155136 Chr.5: 139393479-139393479 on Build GRCh38
rs1000348803 Chr.5: 139393322-139393322 on Build GRCh38
rs1020407146 Chr.5: 139393296-139393296 on Build GRCh38
rs746808917 Chr.5: 139393375-139393375 on Build GRCh38
rs896542465 Chr.5: 139393606-139393606 on Build GRCh38
rs1044219675 Chr.5: 139393318-139393318 on Build GRCh38
rs980029928 Chr.5: 139393389-139393389 on Build GRCh38
rs772602044 Chr.5: 139393509-139393509 on Build GRCh38
rs968264707 Chr.5: 139393385-139393385 on Build GRCh38
rs912228807 Chr.5: 139393239-139393239 on Build GRCh38
rs939704635 Chr.5: 139393498-139393498 on Build GRCh38
rs111342748 Chr.5: 139393607-139393607 on Build GRCh38
rs753748712 Chr.5: 139393550-139393550 on Build GRCh38
rs535719316 Chr.5: 139393490-139393490 on Build GRCh38
rs183685944 Chr.5: 139393332-139393332 on Build GRCh38
rs764721292 Chr.5: 139393262-139393263 on Build GRCh38
rs926460026 Chr.5: 139393493-139393493 on Build GRCh38
rs1021229114 Chr.5: 139393383-139393383 on Build GRCh38
rs1028956042 Chr.5: 139393649-139393649 on Build GRCh38
rs141056757 Chr.5: 139393575-139393575 on Build GRCh38
rs895829371 Chr.5: 139393275-139393275 on Build GRCh38
rs1000257218 Chr.5: 139393599-139393599 on Build GRCh38
rs904681641 Chr.5: 139393314-139393314 on Build GRCh38
rs773255992 Chr.5: 139393209-139393209 on Build GRCh38
rs931233537 Chr.5: 139393525-139393525 on Build GRCh38
rs1009535627 Chr.5: 139393376-139393376 on Build GRCh38
rs972349959 Chr.5: 139393517-139393517 on Build GRCh38
rs1001114950 Chr.5: 139393638-139393638 on Build GRCh38
rs905137839 Chr.5: 139393223-139393223 on Build GRCh38
rs763116928 Chr.5: 139393312-139393312 on Build GRCh38
rs954248004 Chr.5: 139393653-139393653 on Build GRCh38
rs35571924 Chr.5: 139393434-139393435 on Build GRCh38
rs1010702549 Chr.5: 139393284-139393284 on Build GRCh38
rs151107743 Chr.5: 139393565-139393565 on Build GRCh38
rs1050526864 Chr.5: 139393601-139393601 on Build GRCh38
rs986898644 Chr.5: 139393215-139393215 on Build GRCh38
rs961737016 Chr.5: 139393401-139393401 on Build GRCh38
rs937305971 Chr.5: 139393235-139393235 on Build GRCh38
rs199620338 Chr.5: 139393439-139393439 on Build GRCh38
rs932398788 Chr.5: 139393212-139393212 on Build GRCh38
rs766053899 Chr.5: 139393586-139393586 on Build GRCh38
rs939220451 Chr.5: 139393533-139393533 on Build GRCh38
rs777860665 Chr.5: 139393466-139393466 on Build GRCh38
rs376257702 Chr.5: 139393331-139393331 on Build GRCh38
rs931167385 Chr.5: 139393308-139393308 on Build GRCh38
rs574701701 Chr.5: 139393485-139393485 on Build GRCh38
rs779658454 Chr.5: 139393371-139393371 on Build GRCh38
rs1057130151 Chr.5: 139393272-139393272 on Build GRCh38
rs373584534 Chr.5: 139393435-139393435 on Build GRCh38
rs776114809 Chr.5: 139393524-139393524 on Build GRCh38
rs919868384 Chr.5: 139393518-139393518 on Build GRCh38
rs763971984 Chr.5: 139393541-139393541 on Build GRCh38
rs775037993 Chr.5: 139393402-139393403 on Build GRCh38
rs564225693 Chr.5: 139393406-139393406 on Build GRCh38
rs1042392046 Chr.5: 139393615-139393615 on Build GRCh38
rs1036918622 Chr.5: 139393242-139393242 on Build GRCh38
rs939652877 Chr.5: 139393240-139393240 on Build GRCh38
rs953970607 Chr.5: 139393410-139393410 on Build GRCh38
rs534808227 Chr.5: 139393468-139393468 on Build GRCh38
rs753455003 Chr.5: 139393241-139393241 on Build GRCh38
rs867945272 Chr.5: 139393356-139393356 on Build GRCh38
rs1018995260 Chr.5: 139393398-139393398 on Build GRCh38
rs764777408 Chr.5: 139393547-139393547 on Build GRCh38

View Full Product Details