Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 139394364-139394866 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PROB1
Entrez Gene ID
389333
Gene Name
proline rich basic protein 1
Gene Aliases
C5orf65
UniGene
Hs.729482
Gene Chromosome Location
Chr.5: 139391946-139395196 on Build GRCh38
Transcripts
NM_001161546.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6420645 c.643C>G p.R215G Chr.5: 139394439-139394439 on Build GRCh38
4632956 c.221G>T p.S74I Chr.5: 139394861-139394861 on Build GRCh38
6335941 c.273C>A p.G91G Chr.5: 139394809-139394809 on Build GRCh38
6394758 c.698G>A p.R233H Chr.5: 139394384-139394384 on Build GRCh38
3760938 c.364G>A p.G122S Chr.5: 139394718-139394718 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs918862831 Chr.5: 139394792-139394792 on Build GRCh38
rs938208803 Chr.5: 139394733-139394733 on Build GRCh38
rs910573116 Chr.5: 139394416-139394416 on Build GRCh38
rs760338789 Chr.5: 139394697-139394697 on Build GRCh38
rs950894759 Chr.5: 139394709-139394709 on Build GRCh38
rs888229531 Chr.5: 139394599-139394599 on Build GRCh38
rs781563379 Chr.5: 139394808-139394808 on Build GRCh38
rs750430115 Chr.5: 139394724-139394724 on Build GRCh38
rs897382811 Chr.5: 139394551-139394551 on Build GRCh38
rs976696557 Chr.5: 139394790-139394790 on Build GRCh38
rs557429609 Chr.5: 139394410-139394410 on Build GRCh38
rs555069380 Chr.5: 139394598-139394598 on Build GRCh38
rs771610333 Chr.5: 139394545-139394545 on Build GRCh38
rs35262202 Chr.5: 139394825-139394826 on Build GRCh38
rs965361645 Chr.5: 139394748-139394748 on Build GRCh38
rs1013295904 Chr.5: 139394596-139394596 on Build GRCh38
rs919395125 Chr.5: 139394451-139394451 on Build GRCh38
rs1032204676 Chr.5: 139394645-139394645 on Build GRCh38
rs917586305 Chr.5: 139394817-139394817 on Build GRCh38
rs758533551 Chr.5: 139394730-139394730 on Build GRCh38
rs985513181 Chr.5: 139394421-139394421 on Build GRCh38
rs936739507 Chr.5: 139394414-139394414 on Build GRCh38
rs775097303 Chr.5: 139394584-139394584 on Build GRCh38
rs925416840 Chr.5: 139394401-139394401 on Build GRCh38
rs1021160823 Chr.5: 139394616-139394616 on Build GRCh38
rs771011542 Chr.5: 139394825-139394825 on Build GRCh38
rs7447765 Chr.5: 139394404-139394404 on Build GRCh38
rs776292607 Chr.5: 139394706-139394706 on Build GRCh38
rs1006776469 Chr.5: 139394648-139394648 on Build GRCh38
rs895484047 Chr.5: 139394686-139394686 on Build GRCh38
rs73255289 Chr.5: 139394718-139394718 on Build GRCh38
rs1000686286 Chr.5: 139394636-139394636 on Build GRCh38
rs976581721 Chr.5: 139394442-139394442 on Build GRCh38
rs1042128435 Chr.5: 139394835-139394835 on Build GRCh38
rs979367787 Chr.5: 139394384-139394384 on Build GRCh38
rs929503861 Chr.5: 139394499-139394499 on Build GRCh38
rs371879312 Chr.5: 139394629-139394629 on Build GRCh38
rs1054765878 Chr.5: 139394555-139394555 on Build GRCh38
rs763906406 Chr.5: 139394705-139394705 on Build GRCh38
rs911277279 Chr.5: 139394747-139394747 on Build GRCh38
rs1032257177 Chr.5: 139394396-139394396 on Build GRCh38
rs199886274 Chr.5: 139394501-139394501 on Build GRCh38
rs145817341 Chr.5: 139394847-139394847 on Build GRCh38
rs978570324 Chr.5: 139394394-139394394 on Build GRCh38
rs902333537 Chr.5: 139394619-139394619 on Build GRCh38
rs1017571671 Chr.5: 139394661-139394661 on Build GRCh38
rs752988326 Chr.5: 139394810-139394810 on Build GRCh38
rs751792293 Chr.5: 139394768-139394768 on Build GRCh38
rs985951948 Chr.5: 139394726-139394726 on Build GRCh38
rs558105788 Chr.5: 139394680-139394680 on Build GRCh38
rs751194591 Chr.5: 139394473-139394473 on Build GRCh38
rs1048587905 Chr.5: 139394800-139394800 on Build GRCh38
rs1001682220 Chr.5: 139394604-139394604 on Build GRCh38
rs766588688 Chr.5: 139394754-139394754 on Build GRCh38
rs142054513 Chr.5: 139394542-139394542 on Build GRCh38
rs1049021469 Chr.5: 139394579-139394579 on Build GRCh38
rs542059364 Chr.5: 139394719-139394719 on Build GRCh38
rs748561015 Chr.5: 139394417-139394417 on Build GRCh38
rs986120679 Chr.5: 139394412-139394412 on Build GRCh38
rs953942619 Chr.5: 139394647-139394647 on Build GRCh38

View Full Product Details