Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 139413774-139414267 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC18
Entrez Gene ID
202052
Gene Name
DnaJ heat shock protein family (Hsp40) member C18
Gene Aliases
-
UniGene
Hs.483537
Gene Chromosome Location
Chr.5: 139410203-139439525 on Build GRCh38
Transcripts
NM_152686.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6647288 c.976G>A p.G326R Chr.5: 139414249-139414249 on Build GRCh38
482024 c.974C>T p.A325V Chr.5: 139414251-139414251 on Build GRCh38
1696229 c.1027G>A p.E343K Chr.5: 139414198-139414198 on Build GRCh38
1163380 c.1012G>A p.E338K Chr.5: 139414213-139414213 on Build GRCh38
6647286 c.1009G>A p.A337T Chr.5: 139414216-139414216 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs895585848 Chr.5: 139413959-139413959 on Build GRCh38
rs74411440 Chr.5: 139413877-139413877 on Build GRCh38
rs897956922 Chr.5: 139413953-139413953 on Build GRCh38
rs748334533 Chr.5: 139414184-139414184 on Build GRCh38
rs779145056 Chr.5: 139414160-139414161 on Build GRCh38
rs774034517 Chr.5: 139414174-139414174 on Build GRCh38
rs369684305 Chr.5: 139414162-139414162 on Build GRCh38
rs184705259 Chr.5: 139413971-139413971 on Build GRCh38
rs562355422 Chr.5: 139413870-139413870 on Build GRCh38
rs1032738090 Chr.5: 139413997-139413997 on Build GRCh38
rs760157951 Chr.5: 139413989-139413989 on Build GRCh38
rs944911349 Chr.5: 139413909-139413909 on Build GRCh38
rs767320675 Chr.5: 139414189-139414189 on Build GRCh38
rs377218182 Chr.5: 139414161-139414161 on Build GRCh38
rs904430508 Chr.5: 139413977-139413977 on Build GRCh38
rs14764 Chr.5: 139414193-139414193 on Build GRCh38
rs531443491 Chr.5: 139414144-139414144 on Build GRCh38
rs368192165 Chr.5: 139413972-139413972 on Build GRCh38
rs752639100 Chr.5: 139414191-139414191 on Build GRCh38
rs777689999 Chr.5: 139413863-139413863 on Build GRCh38
rs931616565 Chr.5: 139413858-139413858 on Build GRCh38
rs775871165 Chr.5: 139414152-139414152 on Build GRCh38
rs1056820255 Chr.5: 139414231-139414231 on Build GRCh38
rs146040784 Chr.5: 139414155-139414155 on Build GRCh38
rs771438397 Chr.5: 139413854-139413854 on Build GRCh38
rs745411844 Chr.5: 139414085-139414085 on Build GRCh38
rs111737276 Chr.5: 139414005-139414005 on Build GRCh38
rs978813842 Chr.5: 139413852-139413852 on Build GRCh38
rs756167364 Chr.5: 139414128-139414128 on Build GRCh38
rs549053712 Chr.5: 139414087-139414087 on Build GRCh38
rs757475729 Chr.5: 139414236-139414236 on Build GRCh38
rs772416811 Chr.5: 139414189-139414190 on Build GRCh38
rs1020798249 Chr.5: 139414111-139414111 on Build GRCh38
rs755136782 Chr.5: 139414105-139414105 on Build GRCh38
rs749459331 Chr.5: 139414131-139414131 on Build GRCh38
rs752190190 Chr.5: 139414077-139414077 on Build GRCh38
rs967495595 Chr.5: 139413832-139413832 on Build GRCh38
rs1007322237 Chr.5: 139414031-139414031 on Build GRCh38
rs755874509 Chr.5: 139413850-139413850 on Build GRCh38
rs372536967 Chr.5: 139414130-139414130 on Build GRCh38
rs774814434 Chr.5: 139413911-139413911 on Build GRCh38
rs201783854 Chr.5: 139414208-139414208 on Build GRCh38
rs374143968 Chr.5: 139414238-139414238 on Build GRCh38
rs764365674 Chr.5: 139414230-139414230 on Build GRCh38
rs968262995 Chr.5: 139414125-139414125 on Build GRCh38
rs752628347 Chr.5: 139414120-139414120 on Build GRCh38
rs543958559 Chr.5: 139413816-139413816 on Build GRCh38
rs144028941 Chr.5: 139413890-139413890 on Build GRCh38
rs559024378 Chr.5: 139413932-139413932 on Build GRCh38
rs779332643 Chr.5: 139414149-139414149 on Build GRCh38
rs762583023 Chr.5: 139414163-139414163 on Build GRCh38
rs768112616 Chr.5: 139413998-139413998 on Build GRCh38
rs548941590 Chr.5: 139413950-139413951 on Build GRCh38
rs868460495 Chr.5: 139414213-139414213 on Build GRCh38
rs184813104 Chr.5: 139414159-139414159 on Build GRCh38
rs759306645 Chr.5: 139414180-139414180 on Build GRCh38
rs1034939317 Chr.5: 139414004-139414004 on Build GRCh38
rs746227609 Chr.5: 139414151-139414151 on Build GRCh38
rs764176827 Chr.5: 139414223-139414223 on Build GRCh38
rs142551383 Chr.5: 139414154-139414154 on Build GRCh38
rs367605896 Chr.5: 139414104-139414104 on Build GRCh38
rs770617304 Chr.5: 139413928-139413928 on Build GRCh38
rs1000154802 Chr.5: 139413980-139413980 on Build GRCh38
rs752773591 Chr.5: 139414079-139414079 on Build GRCh38
rs772498479 Chr.5: 139414113-139414113 on Build GRCh38
rs893383522 Chr.5: 139414030-139414030 on Build GRCh38
rs1056853259 Chr.5: 139413899-139413899 on Build GRCh38
rs748906563 Chr.5: 139413914-139413914 on Build GRCh38
rs754834634 Chr.5: 139414107-139414107 on Build GRCh38
rs766015799 Chr.5: 139414165-139414165 on Build GRCh38
rs912143874 Chr.5: 139413898-139413898 on Build GRCh38

View Full Product Details