Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 139448944-139449343 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ECSCR
Entrez Gene ID
641700
Gene Name
endothelial cell surface expressed chemotaxis and apoptosis regulator
Gene Aliases
ARIA, ECSM2
UniGene
Hs.483538
Gene Chromosome Location
Chr.5: 139448554-139462743 on Build GRCh38
Transcripts
NM_001077693.3 NM_001293739.1 NR_121659.1

SNP Detail

SNP ID Chromosome Location
rs373665842 Chr.5: 139449058-139449058 on Build GRCh38
rs573495142 Chr.5: 139449093-139449093 on Build GRCh38
rs756427334 Chr.5: 139449316-139449316 on Build GRCh38
rs137919352 Chr.5: 139449310-139449310 on Build GRCh38
rs951881840 Chr.5: 139449059-139449059 on Build GRCh38
rs1018613945 Chr.5: 139449004-139449004 on Build GRCh38
rs544186967 Chr.5: 139449262-139449262 on Build GRCh38
rs187668113 Chr.5: 139449311-139449311 on Build GRCh38
rs747377280 Chr.5: 139449094-139449094 on Build GRCh38
rs866561958 Chr.5: 139449019-139449019 on Build GRCh38
rs777130666 Chr.5: 139449146-139449146 on Build GRCh38
rs751141700 Chr.5: 139449284-139449284 on Build GRCh38
rs1035511927 Chr.5: 139448998-139448998 on Build GRCh38
rs987960203 Chr.5: 139449035-139449035 on Build GRCh38
rs370560764 Chr.5: 139449042-139449042 on Build GRCh38
rs561747287 Chr.5: 139449193-139449193 on Build GRCh38
rs768923342 Chr.5: 139449145-139449145 on Build GRCh38
rs540528870 Chr.5: 139449114-139449114 on Build GRCh38
rs1027475010 Chr.5: 139449044-139449044 on Build GRCh38
rs146945139 Chr.5: 139449297-139449297 on Build GRCh38
rs552417410 Chr.5: 139449251-139449251 on Build GRCh38
rs1002606816 Chr.5: 139449315-139449315 on Build GRCh38
rs762240757 Chr.5: 139449188-139449188 on Build GRCh38
rs562674138 Chr.5: 139449295-139449295 on Build GRCh38
rs920906644 Chr.5: 139449119-139449119 on Build GRCh38
rs529293034 Chr.5: 139449245-139449245 on Build GRCh38
rs993214063 Chr.5: 139449266-139449266 on Build GRCh38
rs888776424 Chr.5: 139448981-139448981 on Build GRCh38
rs760051078 Chr.5: 139449294-139449294 on Build GRCh38
rs1050824380 Chr.5: 139449160-139449160 on Build GRCh38
rs1008947921 Chr.5: 139449002-139449002 on Build GRCh38
rs1033629494 Chr.5: 139449239-139449239 on Build GRCh38
rs946593676 Chr.5: 139449057-139449057 on Build GRCh38
rs897544016 Chr.5: 139449028-139449028 on Build GRCh38
rs898008814 Chr.5: 139449283-139449283 on Build GRCh38
rs1050676146 Chr.5: 139448972-139448972 on Build GRCh38

View Full Product Details