Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 140558512-140559020 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APBB3
Entrez Gene ID
10307
Gene Name
amyloid beta precursor protein binding family B member 3
Gene Aliases
FE65L2, SRA
UniGene
Hs.529449
Gene Chromosome Location
Chr.5: 140558268-140564604 on Build GRCh38
Transcripts
NM_006051.3 NM_133172.2 NM_133173.2 NM_133174.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5060662 c.1310G>A p.R437H Chr.5: 140558757-140558757 on Build GRCh38
1433632 c.1274C>A p.S425Y Chr.5: 140558793-140558793 on Build GRCh38
1433631 c.1305G>T p.Q435H Chr.5: 140558762-140558762 on Build GRCh38
4524885 c.1298G>A p.G433D Chr.5: 140558769-140558769 on Build GRCh38
7000060 c.1379T>C p.L460P Chr.5: 140558688-140558688 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746025010 Chr.5: 140558599-140558599 on Build GRCh38
rs766408600 Chr.5: 140558839-140558839 on Build GRCh38
rs751180124 Chr.5: 140558627-140558631 on Build GRCh38
rs746337320 Chr.5: 140558745-140558745 on Build GRCh38
rs34926962 Chr.5: 140558785-140558785 on Build GRCh38
rs756002552 Chr.5: 140558781-140558782 on Build GRCh38
rs778410690 Chr.5: 140558763-140558763 on Build GRCh38
rs897346801 Chr.5: 140558968-140558968 on Build GRCh38
rs143491686 Chr.5: 140558657-140558657 on Build GRCh38
rs756057504 Chr.5: 140558556-140558556 on Build GRCh38
rs370414260 Chr.5: 140558641-140558641 on Build GRCh38
rs776792015 Chr.5: 140558918-140558919 on Build GRCh38
rs776227488 Chr.5: 140558797-140558797 on Build GRCh38
rs543273423 Chr.5: 140558543-140558543 on Build GRCh38
rs35131969 Chr.5: 140558965-140558966 on Build GRCh38
rs770340112 Chr.5: 140558700-140558700 on Build GRCh38
rs768520917 Chr.5: 140558729-140558729 on Build GRCh38
rs748777523 Chr.5: 140558638-140558638 on Build GRCh38
rs1007140467 Chr.5: 140558580-140558580 on Build GRCh38
rs146302480 Chr.5: 140558759-140558759 on Build GRCh38
rs764090820 Chr.5: 140558656-140558656 on Build GRCh38
rs757074030 Chr.5: 140558630-140558630 on Build GRCh38
rs781471610 Chr.5: 140558758-140558758 on Build GRCh38
rs961651106 Chr.5: 140558682-140558682 on Build GRCh38
rs994087660 Chr.5: 140558616-140558616 on Build GRCh38
rs773944900 Chr.5: 140558646-140558646 on Build GRCh38
rs767409262 Chr.5: 140558574-140558574 on Build GRCh38
rs761829691 Chr.5: 140558664-140558664 on Build GRCh38
rs145787718 Chr.5: 140558601-140558601 on Build GRCh38
rs529722873 Chr.5: 140558671-140558671 on Build GRCh38
rs759698899 Chr.5: 140558847-140558847 on Build GRCh38
rs764486058 Chr.5: 140558862-140558862 on Build GRCh38
rs760522044 Chr.5: 140558725-140558725 on Build GRCh38
rs537032916 Chr.5: 140558736-140558736 on Build GRCh38
rs377738702 Chr.5: 140558672-140558672 on Build GRCh38
rs755227669 Chr.5: 140558757-140558757 on Build GRCh38
rs958729397 Chr.5: 140558983-140558983 on Build GRCh38
rs532222938 Chr.5: 140558624-140558624 on Build GRCh38
rs548169457 Chr.5: 140558692-140558692 on Build GRCh38
rs760613759 Chr.5: 140558655-140558655 on Build GRCh38
rs150554513 Chr.5: 140558737-140558737 on Build GRCh38
rs778229844 Chr.5: 140558701-140558701 on Build GRCh38
rs765626152 Chr.5: 140558642-140558642 on Build GRCh38
rs1005193166 Chr.5: 140558893-140558893 on Build GRCh38
rs992571578 Chr.5: 140558993-140558993 on Build GRCh38
rs757869143 Chr.5: 140558561-140558570 on Build GRCh38
rs757868663 Chr.5: 140558872-140558872 on Build GRCh38
rs868116147 Chr.5: 140558670-140558670 on Build GRCh38
rs767178967 Chr.5: 140558541-140558541 on Build GRCh38
rs776610034 Chr.5: 140558929-140558929 on Build GRCh38
rs115361603 Chr.5: 140558853-140558853 on Build GRCh38
rs910302628 Chr.5: 140558565-140558565 on Build GRCh38
rs745339001 Chr.5: 140558766-140558766 on Build GRCh38
rs754321148 Chr.5: 140558868-140558868 on Build GRCh38
rs769743683 Chr.5: 140558985-140558985 on Build GRCh38
rs745441682 Chr.5: 140558705-140558705 on Build GRCh38
rs774421031 Chr.5: 140558846-140558846 on Build GRCh38
rs748302965 Chr.5: 140558950-140558950 on Build GRCh38
rs536302777 Chr.5: 140558809-140558809 on Build GRCh38
rs748565423 Chr.5: 140558699-140558699 on Build GRCh38
rs199704720 Chr.5: 140558784-140558784 on Build GRCh38
rs374702672 Chr.5: 140558653-140558653 on Build GRCh38
rs148042615 Chr.5: 140558703-140558703 on Build GRCh38
rs752462282 Chr.5: 140558647-140558647 on Build GRCh38
rs148938296 Chr.5: 140558718-140558718 on Build GRCh38
rs765187519 Chr.5: 140558667-140558667 on Build GRCh38
rs757319109 Chr.5: 140558586-140558586 on Build GRCh38
rs576070968 Chr.5: 140558864-140558864 on Build GRCh38
rs564829716 Chr.5: 140558622-140558622 on Build GRCh38
rs761036999 Chr.5: 140558683-140558685 on Build GRCh38
rs745771417 Chr.5: 140558722-140558723 on Build GRCh38
rs111366985 Chr.5: 140558780-140558780 on Build GRCh38
rs373130164 Chr.5: 140558848-140558848 on Build GRCh38
rs761721268 Chr.5: 140558806-140558806 on Build GRCh38
rs762823479 Chr.5: 140558832-140558832 on Build GRCh38
rs143692635 Chr.5: 140558752-140558752 on Build GRCh38
rs781561764 Chr.5: 140558698-140558698 on Build GRCh38
rs555472778 Chr.5: 140558579-140558580 on Build GRCh38
rs771757069 Chr.5: 140558768-140558768 on Build GRCh38
rs17855437 Chr.5: 140558632-140558632 on Build GRCh38
rs755308507 Chr.5: 140558854-140558854 on Build GRCh38
rs753336111 Chr.5: 140558695-140558695 on Build GRCh38
rs145698446 Chr.5: 140558652-140558652 on Build GRCh38
rs888556291 Chr.5: 140558942-140558943 on Build GRCh38
rs138766975 Chr.5: 140558751-140558751 on Build GRCh38
rs746617921 Chr.5: 140558782-140558782 on Build GRCh38
rs773253585 Chr.5: 140558812-140558812 on Build GRCh38
rs543409719 Chr.5: 140558984-140558984 on Build GRCh38
rs138614728 Chr.5: 140558617-140558617 on Build GRCh38
rs1037207992 Chr.5: 140558961-140558961 on Build GRCh38
rs780325641 Chr.5: 140558687-140558687 on Build GRCh38
rs376331384 Chr.5: 140558589-140558589 on Build GRCh38
rs373326815 Chr.5: 140558539-140558539 on Build GRCh38
rs753000293 Chr.5: 140558851-140558851 on Build GRCh38
rs1006009095 Chr.5: 140558953-140558953 on Build GRCh38
rs753962169 Chr.5: 140558576-140558576 on Build GRCh38
rs758829612 Chr.5: 140558746-140558746 on Build GRCh38

View Full Product Details