Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 140561983-140562477 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APBB3
Entrez Gene ID
10307
Gene Name
amyloid beta precursor protein binding family B member 3
Gene Aliases
FE65L2, SRA
UniGene
Hs.529449
Gene Chromosome Location
Chr.5: 140558268-140564604 on Build GRCh38
Transcripts
NM_006051.3 NM_133172.2 NM_133173.2 NM_133174.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4546314 c.397G>A p.D133N Chr.5: 140562454-140562454 on Build GRCh38
5684894 c.409G>T p.G137W Chr.5: 140562442-140562442 on Build GRCh38
3238933 c.407C>T p.P136L Chr.5: 140562444-140562444 on Build GRCh38
5900713 c.439A>C p.I147L Chr.5: 140562412-140562412 on Build GRCh38
81998 c.561C>T p.H187H Chr.5: 140562165-140562165 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200481375 Chr.5: 140562057-140562057 on Build GRCh38
rs374871689 Chr.5: 140562310-140562310 on Build GRCh38
rs184083845 Chr.5: 140562399-140562399 on Build GRCh38
rs147739115 Chr.5: 140562207-140562207 on Build GRCh38
rs145695941 Chr.5: 140562175-140562175 on Build GRCh38
rs747586649 Chr.5: 140562424-140562424 on Build GRCh38
rs370196973 Chr.5: 140562321-140562321 on Build GRCh38
rs201714439 Chr.5: 140562100-140562100 on Build GRCh38
rs747141139 Chr.5: 140562024-140562024 on Build GRCh38
rs766301321 Chr.5: 140562259-140562259 on Build GRCh38
rs757091523 Chr.5: 140562007-140562007 on Build GRCh38
rs199700470 Chr.5: 140562090-140562090 on Build GRCh38
rs370370773 Chr.5: 140562444-140562444 on Build GRCh38
rs555695006 Chr.5: 140562341-140562341 on Build GRCh38
rs763581838 Chr.5: 140562373-140562374 on Build GRCh38
rs777291861 Chr.5: 140562133-140562133 on Build GRCh38
rs979022363 Chr.5: 140562097-140562097 on Build GRCh38
rs774287378 Chr.5: 140562447-140562447 on Build GRCh38
rs746304138 Chr.5: 140562316-140562316 on Build GRCh38
rs980276299 Chr.5: 140562398-140562398 on Build GRCh38
rs764101333 Chr.5: 140562389-140562389 on Build GRCh38
rs145227435 Chr.5: 140562215-140562215 on Build GRCh38
rs190071068 Chr.5: 140562079-140562079 on Build GRCh38
rs200825219 Chr.5: 140562101-140562101 on Build GRCh38
rs372228585 Chr.5: 140562120-140562120 on Build GRCh38
rs866488878 Chr.5: 140562022-140562022 on Build GRCh38
rs766488336 Chr.5: 140562148-140562148 on Build GRCh38
rs776216532 Chr.5: 140562119-140562119 on Build GRCh38
rs539602385 Chr.5: 140562055-140562058 on Build GRCh38
rs777154368 Chr.5: 140562433-140562433 on Build GRCh38
rs1004746086 Chr.5: 140562339-140562339 on Build GRCh38
rs755243445 Chr.5: 140562040-140562040 on Build GRCh38
rs947496755 Chr.5: 140562236-140562236 on Build GRCh38
rs749729906 Chr.5: 140562186-140562186 on Build GRCh38
rs769543936 Chr.5: 140562238-140562238 on Build GRCh38
rs140778547 Chr.5: 140562391-140562391 on Build GRCh38
rs917152798 Chr.5: 140562126-140562126 on Build GRCh38
rs750845353 Chr.5: 140562407-140562407 on Build GRCh38
rs758901847 Chr.5: 140562409-140562409 on Build GRCh38
rs544391182 Chr.5: 140562128-140562128 on Build GRCh38
rs751655734 Chr.5: 140562149-140562149 on Build GRCh38
rs768114183 Chr.5: 140562219-140562219 on Build GRCh38
rs754132038 Chr.5: 140562309-140562309 on Build GRCh38
rs752904001 Chr.5: 140562170-140562170 on Build GRCh38
rs756086342 Chr.5: 140562110-140562110 on Build GRCh38
rs779936297 Chr.5: 140562020-140562020 on Build GRCh38
rs370757168 Chr.5: 140562384-140562384 on Build GRCh38
rs373407419 Chr.5: 140562331-140562331 on Build GRCh38
rs558152802 Chr.5: 140562385-140562385 on Build GRCh38
rs753597303 Chr.5: 140562005-140562005 on Build GRCh38
rs140070711 Chr.5: 140562130-140562130 on Build GRCh38
rs574279618 Chr.5: 140562333-140562333 on Build GRCh38
rs148789210 Chr.5: 140562220-140562220 on Build GRCh38
rs913269860 Chr.5: 140562429-140562429 on Build GRCh38
rs779278413 Chr.5: 140562311-140562311 on Build GRCh38
rs920129674 Chr.5: 140562076-140562076 on Build GRCh38
rs755612225 Chr.5: 140562425-140562425 on Build GRCh38
rs186001560 Chr.5: 140562364-140562364 on Build GRCh38
rs7715021 Chr.5: 140562358-140562358 on Build GRCh38
rs781616111 Chr.5: 140562054-140562054 on Build GRCh38
rs375776763 Chr.5: 140562277-140562277 on Build GRCh38
rs996930505 Chr.5: 140562294-140562294 on Build GRCh38
rs772877296 Chr.5: 140562243-140562243 on Build GRCh38
rs750242454 Chr.5: 140562127-140562127 on Build GRCh38
rs886693922 Chr.5: 140562035-140562035 on Build GRCh38
rs781456479 Chr.5: 140562156-140562156 on Build GRCh38
rs182314255 Chr.5: 140562270-140562270 on Build GRCh38
rs7734425 Chr.5: 140562275-140562275 on Build GRCh38
rs529364783 Chr.5: 140562420-140562420 on Build GRCh38
rs764874015 Chr.5: 140562122-140562122 on Build GRCh38
rs1047859972 Chr.5: 140562026-140562026 on Build GRCh38
rs900743433 Chr.5: 140562291-140562291 on Build GRCh38
rs1040629907 Chr.5: 140562288-140562288 on Build GRCh38
rs376576611 Chr.5: 140562228-140562228 on Build GRCh38
rs779443597 Chr.5: 140562209-140562209 on Build GRCh38
rs34920898 Chr.5: 140562390-140562390 on Build GRCh38
rs748914035 Chr.5: 140562443-140562443 on Build GRCh38
rs779129894 Chr.5: 140562055-140562055 on Build GRCh38
rs755063242 Chr.5: 140562153-140562153 on Build GRCh38
rs1047891225 Chr.5: 140562245-140562245 on Build GRCh38
rs1027965684 Chr.5: 140562296-140562296 on Build GRCh38
rs759533876 Chr.5: 140562383-140562383 on Build GRCh38
rs955849006 Chr.5: 140562102-140562102 on Build GRCh38
rs759028787 Chr.5: 140562268-140562269 on Build GRCh38
rs368256750 Chr.5: 140562254-140562254 on Build GRCh38
rs745701428 Chr.5: 140562452-140562452 on Build GRCh38
rs763711691 Chr.5: 140562118-140562118 on Build GRCh38
rs373109785 Chr.5: 140562302-140562302 on Build GRCh38
rs750287930 Chr.5: 140562008-140562008 on Build GRCh38
rs904274234 Chr.5: 140562301-140562301 on Build GRCh38
rs774301630 Chr.5: 140562382-140562382 on Build GRCh38
rs777988103 Chr.5: 140562183-140562183 on Build GRCh38
rs746957538 Chr.5: 140562067-140562067 on Build GRCh38

View Full Product Details