Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 140673185-140673691 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DND1
Entrez Gene ID
373863
Gene Name
DND microRNA-mediated repression inhibitor 1
Gene Aliases
RBMS4
UniGene
Hs.591262
Gene Chromosome Location
Chr.5: 140670796-140673586 on Build GRCh38
Transcripts
NM_194249.2 XM_005268469.2 XM_017009600.1

Gene Symbol
WDR55
Entrez Gene ID
54853
Gene Name
WD repeat domain 55
Gene Aliases
-
UniGene
Hs.286261
Gene Chromosome Location
Chr.5: 140664799-140674119 on Build GRCh38
Transcripts
NM_194249.2 XM_005268469.2 XM_017009600.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7271672 c.129C>T p.G43G Chr.5: 140673284-140673284 on Build GRCh38
5367685 c.25-10_25-9insT p.? Chr.5: 140673397-140673398 on Build GRCh38
5419900 c.142G>T p.G48C Chr.5: 140673271-140673271 on Build GRCh38
5421685 c.75G>A p.W25* Chr.5: 140673338-140673338 on Build GRCh38
5503019 c.8C>T p.S3F Chr.5: 140673535-140673535 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757818423 Chr.5: 140673335-140673335 on Build GRCh38
rs745818505 Chr.5: 140673505-140673505 on Build GRCh38
rs376707407 Chr.5: 140673433-140673433 on Build GRCh38
rs766254687 Chr.5: 140673391-140673392 on Build GRCh38
rs902189822 Chr.5: 140673530-140673530 on Build GRCh38
rs1013349980 Chr.5: 140673499-140673499 on Build GRCh38
rs756583223 Chr.5: 140673226-140673226 on Build GRCh38
rs748020236 Chr.5: 140673255-140673255 on Build GRCh38
rs762553823 Chr.5: 140673432-140673470 on Build GRCh38
rs866075591 Chr.5: 140673468-140673468 on Build GRCh38
rs760957572 Chr.5: 140673429-140673429 on Build GRCh38
rs777653293 Chr.5: 140673374-140673374 on Build GRCh38
rs111519226 Chr.5: 140673599-140673599 on Build GRCh38
rs368780113 Chr.5: 140673484-140673484 on Build GRCh38
rs751166591 Chr.5: 140673348-140673348 on Build GRCh38
rs377166872 Chr.5: 140673526-140673526 on Build GRCh38
rs369568831 Chr.5: 140673410-140673410 on Build GRCh38
rs747811391 Chr.5: 140673359-140673359 on Build GRCh38
rs142088963 Chr.5: 140673586-140673586 on Build GRCh38
rs141552022 Chr.5: 140673353-140673353 on Build GRCh38
rs900198266 Chr.5: 140673257-140673257 on Build GRCh38
rs374423440 Chr.5: 140673550-140673550 on Build GRCh38
rs930461743 Chr.5: 140673402-140673402 on Build GRCh38
rs937811673 Chr.5: 140673479-140673479 on Build GRCh38
rs749249714 Chr.5: 140673259-140673259 on Build GRCh38
rs571440734 Chr.5: 140673407-140673407 on Build GRCh38
rs757789509 Chr.5: 140673246-140673246 on Build GRCh38
rs540632131 Chr.5: 140673212-140673212 on Build GRCh38
rs868740958 Chr.5: 140673445-140673445 on Build GRCh38
rs755794543 Chr.5: 140673365-140673365 on Build GRCh38
rs966059279 Chr.5: 140673612-140673612 on Build GRCh38
rs779624497 Chr.5: 140673247-140673247 on Build GRCh38
rs571609012 Chr.5: 140673488-140673488 on Build GRCh38
rs765155320 Chr.5: 140673552-140673552 on Build GRCh38
rs751098806 Chr.5: 140673436-140673436 on Build GRCh38
rs984834568 Chr.5: 140673657-140673657 on Build GRCh38
rs199564657 Chr.5: 140673266-140673266 on Build GRCh38
rs757136151 Chr.5: 140673495-140673495 on Build GRCh38
rs976589897 Chr.5: 140673614-140673614 on Build GRCh38
rs951439350 Chr.5: 140673662-140673663 on Build GRCh38
rs550720780 Chr.5: 140673482-140673482 on Build GRCh38
rs764578051 Chr.5: 140673233-140673233 on Build GRCh38
rs114176478 Chr.5: 140673449-140673449 on Build GRCh38
rs974063166 Chr.5: 140673415-140673415 on Build GRCh38
rs201431369 Chr.5: 140673258-140673258 on Build GRCh38
rs767135434 Chr.5: 140673472-140673472 on Build GRCh38
rs759729985 Chr.5: 140673270-140673270 on Build GRCh38
rs762206339 Chr.5: 140673432-140673432 on Build GRCh38
rs1047231115 Chr.5: 140673224-140673225 on Build GRCh38
rs201389616 Chr.5: 140673274-140673274 on Build GRCh38
rs755812376 Chr.5: 140673481-140673481 on Build GRCh38
rs531936231 Chr.5: 140673461-140673461 on Build GRCh38
rs148614030 Chr.5: 140673582-140673582 on Build GRCh38
rs1007671378 Chr.5: 140673598-140673598 on Build GRCh38
rs753278816 Chr.5: 140673381-140673381 on Build GRCh38
rs566043624 Chr.5: 140673568-140673568 on Build GRCh38
rs753107806 Chr.5: 140673220-140673220 on Build GRCh38
rs759056756 Chr.5: 140673471-140673471 on Build GRCh38
rs201758745 Chr.5: 140673475-140673475 on Build GRCh38
rs547446879 Chr.5: 140673500-140673500 on Build GRCh38
rs1017733675 Chr.5: 140673604-140673604 on Build GRCh38
rs374586539 Chr.5: 140673621-140673621 on Build GRCh38
rs775829419 Chr.5: 140673296-140673296 on Build GRCh38
rs780140549 Chr.5: 140673535-140673535 on Build GRCh38
rs754150314 Chr.5: 140673647-140673647 on Build GRCh38
rs746015671 Chr.5: 140673413-140673413 on Build GRCh38
rs999646443 Chr.5: 140673333-140673333 on Build GRCh38
rs112439761 Chr.5: 140673491-140673491 on Build GRCh38
rs746624191 Chr.5: 140673248-140673248 on Build GRCh38
rs754669893 Chr.5: 140673350-140673350 on Build GRCh38
rs749130710 Chr.5: 140673394-140673394 on Build GRCh38
rs777079971 Chr.5: 140673430-140673430 on Build GRCh38
rs754702646 Chr.5: 140673249-140673249 on Build GRCh38
rs749935965 Chr.5: 140673234-140673234 on Build GRCh38
rs369626937 Chr.5: 140673537-140673537 on Build GRCh38
rs781115264 Chr.5: 140673254-140673254 on Build GRCh38
rs765761367 Chr.5: 140673342-140673342 on Build GRCh38
rs757612991 Chr.5: 140673664-140673664 on Build GRCh38
rs929216982 Chr.5: 140673416-140673416 on Build GRCh38
rs139029027 Chr.5: 140673319-140673319 on Build GRCh38
rs1043418529 Chr.5: 140673531-140673531 on Build GRCh38
rs111650695 Chr.5: 140673253-140673253 on Build GRCh38
rs761004432 Chr.5: 140673307-140673307 on Build GRCh38
rs747153176 Chr.5: 140673538-140673538 on Build GRCh38
rs776725724 Chr.5: 140673220-140673228 on Build GRCh38
rs575880920 Chr.5: 140673589-140673589 on Build GRCh38
rs763840956 Chr.5: 140673496-140673498 on Build GRCh38
rs1013386594 Chr.5: 140673590-140673590 on Build GRCh38
rs772074635 Chr.5: 140673512-140673512 on Build GRCh38
rs764532813 Chr.5: 140673317-140673317 on Build GRCh38
rs1023401870 Chr.5: 140673615-140673615 on Build GRCh38
rs199889319 Chr.5: 140673371-140673371 on Build GRCh38
rs767766110 Chr.5: 140673287-140673287 on Build GRCh38
rs373543064 Chr.5: 140673569-140673569 on Build GRCh38
rs755266345 Chr.5: 140673668-140673669 on Build GRCh38
rs1023496878 Chr.5: 140673395-140673395 on Build GRCh38
rs772270697 Chr.5: 140673422-140673422 on Build GRCh38

View Full Product Details