Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 141644780-141645288 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCHSD1
Entrez Gene ID
89848
Gene Name
FCH and double SH3 domains 1
Gene Aliases
NWK2
UniGene
Hs.591257
Gene Chromosome Location
Chr.5: 141639302-141651453 on Build GRCh38
Transcripts
NM_033449.2 XM_005268524.4 XM_006714803.3 XM_011537698.2 XM_011537700.2 XM_017010013.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6458006 c.1512C>T p.D504D Chr.5: 141644871-141644871 on Build GRCh38
4684215 c.1432C>T p.R478C Chr.5: 141645028-141645028 on Build GRCh38
4816586 c.1504G>A p.D502N Chr.5: 141644879-141644879 on Build GRCh38
3827265 c.1502G>A p.G501E Chr.5: 141644881-141644881 on Build GRCh38
275150 c.1513G>A p.E505K Chr.5: 141644870-141644870 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376157133 Chr.5: 141645027-141645027 on Build GRCh38
rs747429830 Chr.5: 141645160-141645160 on Build GRCh38
rs746301997 Chr.5: 141644914-141644914 on Build GRCh38
rs775777190 Chr.5: 141644813-141644814 on Build GRCh38
rs780531960 Chr.5: 141645143-141645143 on Build GRCh38
rs757271005 Chr.5: 141645037-141645037 on Build GRCh38
rs763424275 Chr.5: 141644930-141644947 on Build GRCh38
rs751435282 Chr.5: 141644890-141644890 on Build GRCh38
rs878966159 Chr.5: 141645002-141645002 on Build GRCh38
rs532328224 Chr.5: 141644841-141644841 on Build GRCh38
rs748390098 Chr.5: 141644895-141644895 on Build GRCh38
rs769500524 Chr.5: 141645063-141645063 on Build GRCh38
rs370137791 Chr.5: 141644913-141644913 on Build GRCh38
rs370399564 Chr.5: 141645081-141645081 on Build GRCh38
rs764634134 Chr.5: 141645076-141645076 on Build GRCh38
rs537218233 Chr.5: 141645007-141645007 on Build GRCh38
rs769018125 Chr.5: 141645161-141645161 on Build GRCh38
rs773363325 Chr.5: 141644820-141644820 on Build GRCh38
rs760217270 Chr.5: 141645167-141645178 on Build GRCh38
rs372761136 Chr.5: 141644871-141644871 on Build GRCh38
rs761387475 Chr.5: 141644846-141644846 on Build GRCh38
rs771296495 Chr.5: 141644831-141644831 on Build GRCh38
rs367794498 Chr.5: 141645104-141645104 on Build GRCh38
rs764846398 Chr.5: 141644936-141644936 on Build GRCh38
rs1018709279 Chr.5: 141645117-141645117 on Build GRCh38
rs971791516 Chr.5: 141645134-141645134 on Build GRCh38
rs201569710 Chr.5: 141645028-141645028 on Build GRCh38
rs763873579 Chr.5: 141644842-141644842 on Build GRCh38
rs772519112 Chr.5: 141645055-141645055 on Build GRCh38
rs999372012 Chr.5: 141645224-141645224 on Build GRCh38
rs199983271 Chr.5: 141644989-141644989 on Build GRCh38
rs762363343 Chr.5: 141644956-141644956 on Build GRCh38
rs749309379 Chr.5: 141644906-141644906 on Build GRCh38
rs556405536 Chr.5: 141645124-141645127 on Build GRCh38
rs772724013 Chr.5: 141645076-141645076 on Build GRCh38
rs764470877 Chr.5: 141645097-141645097 on Build GRCh38
rs371398052 Chr.5: 141645105-141645105 on Build GRCh38
rs753476247 Chr.5: 141644843-141644843 on Build GRCh38
rs764758017 Chr.5: 141644852-141644852 on Build GRCh38
rs754712147 Chr.5: 141644891-141644891 on Build GRCh38
rs766268818 Chr.5: 141645190-141645190 on Build GRCh38
rs1047556971 Chr.5: 141645058-141645058 on Build GRCh38
rs781549146 Chr.5: 141645162-141645162 on Build GRCh38
rs557584724 Chr.5: 141645203-141645203 on Build GRCh38
rs770105525 Chr.5: 141644896-141644896 on Build GRCh38
rs775880422 Chr.5: 141645062-141645062 on Build GRCh38
rs773784917 Chr.5: 141645183-141645183 on Build GRCh38
rs952019268 Chr.5: 141645175-141645175 on Build GRCh38
rs748292285 Chr.5: 141644817-141644817 on Build GRCh38
rs746392666 Chr.5: 141645049-141645049 on Build GRCh38
rs1022250786 Chr.5: 141645132-141645132 on Build GRCh38
rs370915381 Chr.5: 141644955-141644955 on Build GRCh38
rs992162380 Chr.5: 141645239-141645239 on Build GRCh38
rs755971201 Chr.5: 141645084-141645084 on Build GRCh38
rs569678362 Chr.5: 141645213-141645213 on Build GRCh38
rs377063522 Chr.5: 141644903-141644903 on Build GRCh38
rs749885164 Chr.5: 141644870-141644870 on Build GRCh38
rs200412858 Chr.5: 141644982-141644982 on Build GRCh38
rs373942523 Chr.5: 141645077-141645077 on Build GRCh38
rs1001425408 Chr.5: 141645093-141645093 on Build GRCh38
rs781072365 Chr.5: 141644893-141644893 on Build GRCh38
rs117633393 Chr.5: 141645170-141645170 on Build GRCh38
rs745936031 Chr.5: 141645110-141645110 on Build GRCh38
rs766393615 Chr.5: 141644963-141644963 on Build GRCh38
rs752493279 Chr.5: 141644983-141644983 on Build GRCh38
rs376415582 Chr.5: 141644945-141644945 on Build GRCh38
rs751462091 Chr.5: 141644970-141644970 on Build GRCh38
rs749289577 Chr.5: 141644883-141644883 on Build GRCh38
rs537364155 Chr.5: 141645199-141645199 on Build GRCh38
rs1007292840 Chr.5: 141645107-141645107 on Build GRCh38
rs984574891 Chr.5: 141645185-141645185 on Build GRCh38
rs770010813 Chr.5: 141644819-141644819 on Build GRCh38
rs754881036 Chr.5: 141644978-141644978 on Build GRCh38
rs763589187 Chr.5: 141644824-141644824 on Build GRCh38
rs370799818 Chr.5: 141645038-141645038 on Build GRCh38
rs375870031 Chr.5: 141644935-141644935 on Build GRCh38
rs772326688 Chr.5: 141644918-141644918 on Build GRCh38
rs758942806 Chr.5: 141645121-141645121 on Build GRCh38
rs748927477 Chr.5: 141645166-141645166 on Build GRCh38
rs781263173 Chr.5: 141644811-141644811 on Build GRCh38
rs747658757 Chr.5: 141644919-141644919 on Build GRCh38
rs911534518 Chr.5: 141644901-141644901 on Build GRCh38
rs754021283 Chr.5: 141645103-141645103 on Build GRCh38

View Full Product Details