Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 142004778-142005321 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNPDA1
Entrez Gene ID
10007
Gene Name
glucosamine-6-phosphate deaminase 1
Gene Aliases
GNP1, GNPDA, GNPI, GPI, HLN
UniGene
Hs.633853
Gene Chromosome Location
Chr.5: 142000669-142013055 on Build GRCh38
Transcripts
NM_005471.4 XM_005268348.1 XM_006714747.2 XM_011537537.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3852308 c.472C>T p.R158C Chr.5: 142005054-142005054 on Build GRCh38
1208430 c.580A>G p.M194V Chr.5: 142004946-142004946 on Build GRCh38
3852307 c.489C>T p.A163A Chr.5: 142005037-142005037 on Build GRCh38
5996373 c.523G>A p.D175N Chr.5: 142005003-142005003 on Build GRCh38
4890270 c.587C>A p.A196D Chr.5: 142004939-142004939 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775057845 Chr.5: 142005130-142005130 on Build GRCh38
rs758358966 Chr.5: 142005069-142005069 on Build GRCh38
rs892621320 Chr.5: 142004953-142004953 on Build GRCh38
rs577698464 Chr.5: 142005043-142005043 on Build GRCh38
rs752006328 Chr.5: 142004934-142004934 on Build GRCh38
rs780702689 Chr.5: 142005095-142005095 on Build GRCh38
rs751566036 Chr.5: 142004902-142004903 on Build GRCh38
rs2232200 Chr.5: 142004841-142004841 on Build GRCh38
rs11557800 Chr.5: 142005076-142005076 on Build GRCh38
rs909894427 Chr.5: 142004907-142004907 on Build GRCh38
rs545628408 Chr.5: 142005020-142005020 on Build GRCh38
rs375019214 Chr.5: 142004902-142004902 on Build GRCh38
rs141747545 Chr.5: 142005040-142005040 on Build GRCh38
rs763883850 Chr.5: 142004925-142004925 on Build GRCh38
rs757135705 Chr.5: 142004947-142004947 on Build GRCh38
rs368357115 Chr.5: 142004903-142004903 on Build GRCh38
rs370469371 Chr.5: 142004899-142004899 on Build GRCh38
rs556301157 Chr.5: 142005217-142005217 on Build GRCh38
rs778714575 Chr.5: 142004956-142004956 on Build GRCh38
rs374456271 Chr.5: 142004988-142004988 on Build GRCh38
rs767622622 Chr.5: 142005053-142005053 on Build GRCh38
rs947976893 Chr.5: 142005059-142005059 on Build GRCh38
rs139642867 Chr.5: 142005055-142005055 on Build GRCh38
rs900054654 Chr.5: 142005273-142005273 on Build GRCh38
rs139151123 Chr.5: 142004965-142004965 on Build GRCh38
rs2232199 Chr.5: 142004854-142004854 on Build GRCh38
rs749745591 Chr.5: 142005120-142005120 on Build GRCh38
rs753591503 Chr.5: 142004937-142004937 on Build GRCh38
rs765673266 Chr.5: 142004895-142004895 on Build GRCh38
rs746621093 Chr.5: 142004966-142004966 on Build GRCh38
rs542904763 Chr.5: 142005260-142005260 on Build GRCh38
rs1002684083 Chr.5: 142005109-142005109 on Build GRCh38
rs149743845 Chr.5: 142005085-142005085 on Build GRCh38
rs147558028 Chr.5: 142004922-142004922 on Build GRCh38
rs150553968 Chr.5: 142005046-142005046 on Build GRCh38
rs774577541 Chr.5: 142005044-142005044 on Build GRCh38
rs1044291462 Chr.5: 142005263-142005263 on Build GRCh38
rs754572280 Chr.5: 142005086-142005086 on Build GRCh38
rs562774648 Chr.5: 142005294-142005294 on Build GRCh38
rs2232198 Chr.5: 142004959-142004959 on Build GRCh38
rs775556466 Chr.5: 142005163-142005163 on Build GRCh38
rs984699620 Chr.5: 142005247-142005247 on Build GRCh38
rs773560238 Chr.5: 142005036-142005036 on Build GRCh38
rs368639771 Chr.5: 142005003-142005003 on Build GRCh38
rs908660941 Chr.5: 142005042-142005042 on Build GRCh38
rs146216672 Chr.5: 142004960-142004960 on Build GRCh38
rs753033449 Chr.5: 142005054-142005054 on Build GRCh38
rs906961106 Chr.5: 142005100-142005100 on Build GRCh38
rs771579507 Chr.5: 142005129-142005129 on Build GRCh38
rs1034189383 Chr.5: 142005201-142005201 on Build GRCh38
rs201899569 Chr.5: 142005134-142005134 on Build GRCh38
rs772237009 Chr.5: 142005138-142005138 on Build GRCh38
rs202180877 Chr.5: 142005112-142005112 on Build GRCh38
rs1052172483 Chr.5: 142005168-142005168 on Build GRCh38
rs778304597 Chr.5: 142005114-142005114 on Build GRCh38
rs920540561 Chr.5: 142004821-142004821 on Build GRCh38
rs750315322 Chr.5: 142005066-142005066 on Build GRCh38
rs574416056 Chr.5: 142004945-142004945 on Build GRCh38
rs757252728 Chr.5: 142004905-142004906 on Build GRCh38
rs371335616 Chr.5: 142004980-142004980 on Build GRCh38
rs747873200 Chr.5: 142005111-142005111 on Build GRCh38
rs755283952 Chr.5: 142004924-142004924 on Build GRCh38
rs539827966 Chr.5: 142005082-142005082 on Build GRCh38
rs1037567857 Chr.5: 142004801-142004801 on Build GRCh38
rs1016636541 Chr.5: 142005271-142005271 on Build GRCh38
rs1020493579 Chr.5: 142004840-142004840 on Build GRCh38
rs867083889 Chr.5: 142005286-142005286 on Build GRCh38
rs765132687 Chr.5: 142005058-142005058 on Build GRCh38

View Full Product Details