Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 142592086-142592588 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGF1
Entrez Gene ID
2246
Gene Name
fibroblast growth factor 1
Gene Aliases
AFGF, ECGF, ECGF-beta, ECGFA, ECGFB, FGF-1, FGF-alpha, FGFA, GLIO703, HBGF-1, HBGF1
UniGene
Hs.483635
Gene Chromosome Location
Chr.5: 142592178-142698070 on Build GRCh38
Transcripts
NM_000800.4 NM_001144892.2 NM_001144934.1 NM_001144935.1 NM_001257205.1 NM_001257206.1 NM_001257207.1 NM_001257208.1 NM_001257209.1 NM_001257210.1 NM_001257211.1 NM_001257212.1 NM_033136.3 NM_033137.2 XM_005268390.4 XM_011537607.2

SNP Detail

SNP ID Chromosome Location
rs759947308 Chr.5: 142592564-142592564 on Build GRCh38
rs892336489 Chr.5: 142592470-142592470 on Build GRCh38
rs761943239 Chr.5: 142592446-142592446 on Build GRCh38
rs193167271 Chr.5: 142592484-142592484 on Build GRCh38
rs771135391 Chr.5: 142592373-142592373 on Build GRCh38
rs960602189 Chr.5: 142592312-142592312 on Build GRCh38
rs980661798 Chr.5: 142592282-142592282 on Build GRCh38
rs538886886 Chr.5: 142592485-142592485 on Build GRCh38
rs1009778725 Chr.5: 142592227-142592227 on Build GRCh38
rs769754350 Chr.5: 142592516-142592516 on Build GRCh38
rs772039892 Chr.5: 142592237-142592237 on Build GRCh38
rs1002904565 Chr.5: 142592248-142592248 on Build GRCh38
rs1053678006 Chr.5: 142592169-142592169 on Build GRCh38
rs969308352 Chr.5: 142592258-142592258 on Build GRCh38
rs920898844 Chr.5: 142592427-142592427 on Build GRCh38
rs888772539 Chr.5: 142592117-142592117 on Build GRCh38
rs370765374 Chr.5: 142592495-142592495 on Build GRCh38
rs17217653 Chr.5: 142592130-142592130 on Build GRCh38
rs775388010 Chr.5: 142592331-142592331 on Build GRCh38
rs1044436077 Chr.5: 142592524-142592524 on Build GRCh38
rs923457914 Chr.5: 142592422-142592422 on Build GRCh38
rs531884306 Chr.5: 142592565-142592565 on Build GRCh38
rs1016895823 Chr.5: 142592368-142592368 on Build GRCh38
rs188253591 Chr.5: 142592426-142592426 on Build GRCh38
rs556086885 Chr.5: 142592482-142592482 on Build GRCh38
rs773272684 Chr.5: 142592538-142592538 on Build GRCh38
rs972682419 Chr.5: 142592414-142592414 on Build GRCh38
rs766571572 Chr.5: 142592548-142592548 on Build GRCh38
rs566958099 Chr.5: 142592356-142592356 on Build GRCh38
rs755386094 Chr.5: 142592494-142592494 on Build GRCh38
rs62382130 Chr.5: 142592532-142592532 on Build GRCh38
rs932922123 Chr.5: 142592450-142592450 on Build GRCh38
rs907319579 Chr.5: 142592247-142592247 on Build GRCh38
rs17099022 Chr.5: 142592563-142592563 on Build GRCh38
rs892311223 Chr.5: 142592209-142592209 on Build GRCh38
rs371405893 Chr.5: 142592369-142592369 on Build GRCh38
rs995968664 Chr.5: 142592510-142592510 on Build GRCh38
rs910239801 Chr.5: 142592431-142592431 on Build GRCh38
rs1034919251 Chr.5: 142592301-142592301 on Build GRCh38
rs946417386 Chr.5: 142592384-142592384 on Build GRCh38
rs4440401 Chr.5: 142592529-142592529 on Build GRCh38
rs368732568 Chr.5: 142592333-142592333 on Build GRCh38
rs898588981 Chr.5: 142592502-142592502 on Build GRCh38
rs1053709760 Chr.5: 142592453-142592453 on Build GRCh38
rs376382697 Chr.5: 142592124-142592124 on Build GRCh38
rs962238594 Chr.5: 142592405-142592405 on Build GRCh38
rs985861366 Chr.5: 142592429-142592430 on Build GRCh38
rs17217646 Chr.5: 142592332-142592332 on Build GRCh38
rs17224045 Chr.5: 142592539-142592539 on Build GRCh38
rs995523562 Chr.5: 142592216-142592216 on Build GRCh38
rs566227979 Chr.5: 142592252-142592252 on Build GRCh38
rs558491807 Chr.5: 142592493-142592493 on Build GRCh38

View Full Product Details