Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 147400614-147401116 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPYSL3
Entrez Gene ID
1809
Gene Name
dihydropyrimidinase like 3
Gene Aliases
CRMP-4, CRMP4, DRP-3, DRP3, LCRMP, ULIP, ULIP-1
UniGene
Hs.519659
Gene Chromosome Location
Chr.5: 147390808-147510056 on Build GRCh38
Transcripts
NM_001197294.1 NM_001387.2 XM_011537574.1 XM_011537577.2

Gene Symbol
STK32A
Entrez Gene ID
202374
Gene Name
serine/threonine kinase 32A
Gene Aliases
YANK1
UniGene
Hs.585069
Gene Chromosome Location
Chr.5: 147234963-147401996 on Build GRCh38
Transcripts
NM_001197294.1 NM_001387.2 XM_011537574.1 XM_011537577.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6649157 c.1043T>C p.F348S Chr.5: 147400759-147400759 on Build GRCh38
3852482 c.981G>A p.Q327Q Chr.5: 147400821-147400821 on Build GRCh38
6649155 c.1046C>T p.T349I Chr.5: 147400756-147400756 on Build GRCh38
213822 c.1087T>A p.S363T Chr.5: 147400715-147400715 on Build GRCh38
1250693 c.1000T>C p.C334R Chr.5: 147400802-147400802 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs76516681 Chr.5: 147400759-147400759 on Build GRCh38
rs749143796 Chr.5: 147400738-147400738 on Build GRCh38
rs745361456 Chr.5: 147400755-147400755 on Build GRCh38
rs750449334 Chr.5: 147400683-147400683 on Build GRCh38
rs770728163 Chr.5: 147400740-147400740 on Build GRCh38
rs778385459 Chr.5: 147401006-147401006 on Build GRCh38
rs996756723 Chr.5: 147400970-147400970 on Build GRCh38
rs762286701 Chr.5: 147400818-147400818 on Build GRCh38
rs755214812 Chr.5: 147400857-147400857 on Build GRCh38
rs913776154 Chr.5: 147401075-147401075 on Build GRCh38
rs775887640 Chr.5: 147400895-147400895 on Build GRCh38
rs780958701 Chr.5: 147400968-147400968 on Build GRCh38
rs765795345 Chr.5: 147400678-147400678 on Build GRCh38
rs777366989 Chr.5: 147400735-147400735 on Build GRCh38
rs928823013 Chr.5: 147400906-147400906 on Build GRCh38
rs573683483 Chr.5: 147400943-147400943 on Build GRCh38
rs913732310 Chr.5: 147400652-147400652 on Build GRCh38
rs576128168 Chr.5: 147401008-147401008 on Build GRCh38
rs1046190056 Chr.5: 147400697-147400697 on Build GRCh38
rs114150549 Chr.5: 147400896-147400896 on Build GRCh38
rs981919471 Chr.5: 147401089-147401089 on Build GRCh38
rs1028187440 Chr.5: 147401043-147401043 on Build GRCh38
rs1047853680 Chr.5: 147400941-147400941 on Build GRCh38
rs753542128 Chr.5: 147400881-147400881 on Build GRCh38
rs760252867 Chr.5: 147400791-147400791 on Build GRCh38
rs762185887 Chr.5: 147400674-147400674 on Build GRCh38
rs368254542 Chr.5: 147400775-147400775 on Build GRCh38
rs755267708 Chr.5: 147400728-147400728 on Build GRCh38
rs751007269 Chr.5: 147400833-147400833 on Build GRCh38
rs989755177 Chr.5: 147400637-147400637 on Build GRCh38
rs765632189 Chr.5: 147400832-147400832 on Build GRCh38
rs200957398 Chr.5: 147400757-147400757 on Build GRCh38
rs958234306 Chr.5: 147401042-147401042 on Build GRCh38
rs1031479910 Chr.5: 147401082-147401082 on Build GRCh38
rs193040448 Chr.5: 147400668-147400668 on Build GRCh38
rs948973562 Chr.5: 147400873-147400873 on Build GRCh38
rs989415736 Chr.5: 147401059-147401059 on Build GRCh38
rs891006679 Chr.5: 147401047-147401047 on Build GRCh38
rs13168262 Chr.5: 147400958-147400958 on Build GRCh38
rs758591692 Chr.5: 147400684-147400684 on Build GRCh38
rs764382691 Chr.5: 147400658-147400658 on Build GRCh38
rs924697047 Chr.5: 147400708-147400708 on Build GRCh38
rs938840348 Chr.5: 147400939-147400939 on Build GRCh38
rs58562158 Chr.5: 147400843-147400843 on Build GRCh38
rs966549923 Chr.5: 147401078-147401078 on Build GRCh38
rs1038475242 Chr.5: 147400957-147400957 on Build GRCh38
rs776812299 Chr.5: 147400807-147400807 on Build GRCh38
rs201019011 Chr.5: 147400800-147400800 on Build GRCh38
rs370311772 Chr.5: 147400709-147400709 on Build GRCh38
rs547641980 Chr.5: 147400851-147400851 on Build GRCh38
rs1003939912 Chr.5: 147400950-147400950 on Build GRCh38
rs1017597788 Chr.5: 147400982-147400982 on Build GRCh38
rs781502216 Chr.5: 147400877-147400877 on Build GRCh38
rs200870327 Chr.5: 147400707-147400707 on Build GRCh38
rs774990938 Chr.5: 147400785-147400785 on Build GRCh38
rs954773817 Chr.5: 147400955-147400955 on Build GRCh38
rs763113590 Chr.5: 147400837-147400837 on Build GRCh38
rs10515587 Chr.5: 147400710-147400710 on Build GRCh38
rs542693058 Chr.5: 147400947-147400947 on Build GRCh38
rs778946047 Chr.5: 147400743-147400743 on Build GRCh38
rs771625097 Chr.5: 147400774-147400774 on Build GRCh38

View Full Product Details