Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 148383877-148384420 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO38
Entrez Gene ID
81545
Gene Name
F-box protein 38
Gene Aliases
Fbx38, HMN2D, MOKA, SP329
UniGene
Hs.483772
Gene Chromosome Location
Chr.5: 148383935-148442836 on Build GRCh38
Transcripts
NM_001271723.1 NM_030793.4 XM_005268513.1 XM_006714797.1 XM_017009899.1

SNP Detail

SNP ID Chromosome Location
rs544546659 Chr.5: 148383938-148383938 on Build GRCh38
rs1009511218 Chr.5: 148384154-148384154 on Build GRCh38
rs970522739 Chr.5: 148384067-148384067 on Build GRCh38
rs936932134 Chr.5: 148384230-148384230 on Build GRCh38
rs546246574 Chr.5: 148383994-148383994 on Build GRCh38
rs972567766 Chr.5: 148384071-148384071 on Build GRCh38
rs1005714432 Chr.5: 148384207-148384207 on Build GRCh38
rs922877796 Chr.5: 148384290-148384290 on Build GRCh38
rs1024704193 Chr.5: 148384066-148384066 on Build GRCh38
rs1024614524 Chr.5: 148384264-148384264 on Build GRCh38
rs893722931 Chr.5: 148384135-148384135 on Build GRCh38
rs1031975205 Chr.5: 148384032-148384032 on Build GRCh38
rs556483417 Chr.5: 148383948-148383948 on Build GRCh38
rs922636363 Chr.5: 148384052-148384052 on Build GRCh38
rs1017541357 Chr.5: 148384234-148384234 on Build GRCh38
rs906173927 Chr.5: 148384158-148384158 on Build GRCh38
rs531916710 Chr.5: 148384069-148384069 on Build GRCh38
rs1042289462 Chr.5: 148384377-148384377 on Build GRCh38
rs906251635 Chr.5: 148384278-148384278 on Build GRCh38
rs550098670 Chr.5: 148384141-148384141 on Build GRCh38
rs1011897592 Chr.5: 148384244-148384244 on Build GRCh38
rs943424407 Chr.5: 148383946-148383946 on Build GRCh38
rs961595866 Chr.5: 148384245-148384245 on Build GRCh38
rs376773941 Chr.5: 148384171-148384171 on Build GRCh38
rs34664276 Chr.5: 148383956-148383957 on Build GRCh38
rs959414830 Chr.5: 148384197-148384197 on Build GRCh38
rs1049016827 Chr.5: 148383906-148383906 on Build GRCh38
rs529497143 Chr.5: 148384275-148384275 on Build GRCh38
rs1053609135 Chr.5: 148384363-148384363 on Build GRCh38
rs909219501 Chr.5: 148383927-148383927 on Build GRCh38
rs564548678 Chr.5: 148384016-148384016 on Build GRCh38
rs934008637 Chr.5: 148384065-148384065 on Build GRCh38
rs911303458 Chr.5: 148384309-148384309 on Build GRCh38
rs1003212895 Chr.5: 148384162-148384162 on Build GRCh38
rs747753206 Chr.5: 148383997-148383997 on Build GRCh38
rs918418415 Chr.5: 148384076-148384076 on Build GRCh38
rs911301059 Chr.5: 148384088-148384088 on Build GRCh38
rs984701469 Chr.5: 148384003-148384003 on Build GRCh38
rs749587016 Chr.5: 148384345-148384345 on Build GRCh38
rs188298286 Chr.5: 148384077-148384077 on Build GRCh38
rs560479371 Chr.5: 148383980-148383980 on Build GRCh38
rs1054005321 Chr.5: 148384239-148384239 on Build GRCh38
rs909207763 Chr.5: 148384113-148384113 on Build GRCh38
rs868431596 Chr.5: 148384314-148384314 on Build GRCh38
rs372258301 Chr.5: 148384046-148384046 on Build GRCh38
rs955338099 Chr.5: 148384292-148384292 on Build GRCh38
rs1039122258 Chr.5: 148384163-148384163 on Build GRCh38
rs566428423 Chr.5: 148384386-148384386 on Build GRCh38
rs755987861 Chr.5: 148383929-148383929 on Build GRCh38
rs527794216 Chr.5: 148383990-148383990 on Build GRCh38
rs577930895 Chr.5: 148383949-148383949 on Build GRCh38
rs1054062782 Chr.5: 148384093-148384093 on Build GRCh38
rs368618335 Chr.5: 148383989-148383989 on Build GRCh38
rs1020672865 Chr.5: 148383970-148383970 on Build GRCh38
rs933421352 Chr.5: 148384184-148384184 on Build GRCh38
rs771872128 Chr.5: 148384019-148384019 on Build GRCh38
rs542241939 Chr.5: 148383975-148383975 on Build GRCh38
rs961968518 Chr.5: 148384023-148384023 on Build GRCh38
rs866333333 Chr.5: 148384109-148384109 on Build GRCh38
rs959528592 Chr.5: 148384000-148384000 on Build GRCh38
rs115923740 Chr.5: 148383966-148383966 on Build GRCh38
rs1011948422 Chr.5: 148384036-148384036 on Build GRCh38
rs902743095 Chr.5: 148384004-148384004 on Build GRCh38
rs1049779854 Chr.5: 148384073-148384073 on Build GRCh38
rs959065543 Chr.5: 148384033-148384033 on Build GRCh38
rs901992668 Chr.5: 148383979-148383979 on Build GRCh38
rs1009229763 Chr.5: 148383958-148383958 on Build GRCh38
rs943183270 Chr.5: 148384089-148384089 on Build GRCh38
rs545351446 Chr.5: 148384084-148384084 on Build GRCh38
rs1042341263 Chr.5: 148384157-148384157 on Build GRCh38
rs915029806 Chr.5: 148384364-148384364 on Build GRCh38
rs894797776 Chr.5: 148384241-148384241 on Build GRCh38
rs574926713 Chr.5: 148383967-148383967 on Build GRCh38
rs2112761 Chr.5: 148384188-148384188 on Build GRCh38
rs576939531 Chr.5: 148383901-148383901 on Build GRCh38
rs985434195 Chr.5: 148384301-148384301 on Build GRCh38
rs923323939 Chr.5: 148384172-148384172 on Build GRCh38
rs201604998 Chr.5: 148384310-148384310 on Build GRCh38
rs994329932 Chr.5: 148384280-148384280 on Build GRCh38
rs1042964518 Chr.5: 148383995-148383995 on Build GRCh38
rs945209657 Chr.5: 148384368-148384368 on Build GRCh38
rs972931787 Chr.5: 148384251-148384251 on Build GRCh38
rs181803430 Chr.5: 148384361-148384361 on Build GRCh38

View Full Product Details