Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 148408937-148409446 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO38
Entrez Gene ID
81545
Gene Name
F-box protein 38
Gene Aliases
Fbx38, HMN2D, MOKA, SP329
UniGene
Hs.483772
Gene Chromosome Location
Chr.5: 148383935-148442836 on Build GRCh38
Transcripts
NM_001271723.1 NM_030793.4 XM_005268513.1 XM_006714797.1 XM_011537684.2 XM_017009899.1 XM_017009900.1 XM_017009901.1 XM_017009902.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1496004 c.878G>A p.R293K Chr.5: 148409133-148409133 on Build GRCh38
4881069 c.945C>T p.I315I Chr.5: 148409200-148409200 on Build GRCh38
4768985 c.942C>G p.L314L Chr.5: 148409197-148409197 on Build GRCh38
4939990 c.909C>G p.C303W Chr.5: 148409164-148409164 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs889477496 Chr.5: 148409153-148409153 on Build GRCh38
rs10070268 Chr.5: 148409349-148409349 on Build GRCh38
rs927715014 Chr.5: 148409003-148409003 on Build GRCh38
rs1034779876 Chr.5: 148408984-148408984 on Build GRCh38
rs1034344121 Chr.5: 148409320-148409320 on Build GRCh38
rs746189368 Chr.5: 148409221-148409221 on Build GRCh38
rs574595935 Chr.5: 148409401-148409401 on Build GRCh38
rs899840584 Chr.5: 148409051-148409057 on Build GRCh38
rs762106449 Chr.5: 148409124-148409124 on Build GRCh38
rs10045756 Chr.5: 148409023-148409023 on Build GRCh38
rs149020291 Chr.5: 148409147-148409147 on Build GRCh38
rs755714527 Chr.5: 148409197-148409197 on Build GRCh38
rs777036251 Chr.5: 148409116-148409116 on Build GRCh38
rs1010372977 Chr.5: 148409286-148409286 on Build GRCh38
rs373406637 Chr.5: 148409089-148409089 on Build GRCh38
rs972619435 Chr.5: 148409041-148409041 on Build GRCh38
rs142117467 Chr.5: 148409186-148409186 on Build GRCh38
rs922489523 Chr.5: 148409051-148409051 on Build GRCh38
rs962110132 Chr.5: 148409350-148409350 on Build GRCh38
rs779223469 Chr.5: 148409086-148409086 on Build GRCh38
rs950593965 Chr.5: 148409312-148409312 on Build GRCh38
rs972502884 Chr.5: 148409381-148409381 on Build GRCh38
rs1049421856 Chr.5: 148408991-148408991 on Build GRCh38
rs780504525 Chr.5: 148409030-148409030 on Build GRCh38
rs779258103 Chr.5: 148409220-148409220 on Build GRCh38
rs763560734 Chr.5: 148409212-148409212 on Build GRCh38
rs767430187 Chr.5: 148409170-148409170 on Build GRCh38
rs980058570 Chr.5: 148408974-148408974 on Build GRCh38
rs952168682 Chr.5: 148409323-148409323 on Build GRCh38
rs768907362 Chr.5: 148409240-148409240 on Build GRCh38
rs985312459 Chr.5: 148409022-148409022 on Build GRCh38
rs368799795 Chr.5: 148409082-148409082 on Build GRCh38
rs151159697 Chr.5: 148409200-148409200 on Build GRCh38
rs781268841 Chr.5: 148409259-148409260 on Build GRCh38
rs756850214 Chr.5: 148409214-148409214 on Build GRCh38
rs775450074 Chr.5: 148409100-148409100 on Build GRCh38
rs1051600187 Chr.5: 148409396-148409396 on Build GRCh38
rs984664383 Chr.5: 148409316-148409316 on Build GRCh38
rs907428198 Chr.5: 148409308-148409308 on Build GRCh38
rs998230702 Chr.5: 148409059-148409059 on Build GRCh38
rs748412692 Chr.5: 148409255-148409255 on Build GRCh38
rs539737502 Chr.5: 148409143-148409143 on Build GRCh38
rs760869868 Chr.5: 148409107-148409107 on Build GRCh38
rs776804716 Chr.5: 148409253-148409253 on Build GRCh38
rs111309578 Chr.5: 148409096-148409096 on Build GRCh38
rs776265942 Chr.5: 148409391-148409391 on Build GRCh38
rs1029764862 Chr.5: 148409080-148409080 on Build GRCh38
rs537673669 Chr.5: 148409306-148409306 on Build GRCh38
rs35481617 Chr.5: 148409269-148409269 on Build GRCh38
rs1001797363 Chr.5: 148409310-148409310 on Build GRCh38
rs751376784 Chr.5: 148409252-148409255 on Build GRCh38
rs766301076 Chr.5: 148409140-148409140 on Build GRCh38
rs768400203 Chr.5: 148409274-148409274 on Build GRCh38
rs758680515 Chr.5: 148409229-148409229 on Build GRCh38
rs921241247 Chr.5: 148409388-148409388 on Build GRCh38
rs757173882 Chr.5: 148409258-148409259 on Build GRCh38
rs768616588 Chr.5: 148409115-148409115 on Build GRCh38
rs112093603 Chr.5: 148409171-148409171 on Build GRCh38
rs1012527321 Chr.5: 148409210-148409210 on Build GRCh38
rs746380365 Chr.5: 148409087-148409087 on Build GRCh38
rs779869264 Chr.5: 148409213-148409213 on Build GRCh38
rs770074887 Chr.5: 148409261-148409261 on Build GRCh38
rs952132202 Chr.5: 148409014-148409014 on Build GRCh38
rs10077690 Chr.5: 148409235-148409235 on Build GRCh38
rs933881391 Chr.5: 148409421-148409421 on Build GRCh38
rs763693920 Chr.5: 148409080-148409080 on Build GRCh38
rs984891799 Chr.5: 148409347-148409347 on Build GRCh38
rs747445114 Chr.5: 148409236-148409236 on Build GRCh38
rs981577222 Chr.5: 148409254-148409254 on Build GRCh38

View Full Product Details