Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 150220372-150220876 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAMK2A
Entrez Gene ID
815
Gene Name
calcium/calmodulin dependent protein kinase II alpha
Gene Aliases
CAMKA
UniGene
Hs.743976
Gene Chromosome Location
Chr.5: 150219491-150289840 on Build GRCh38
Transcripts
NM_015981.3 NM_171825.2

SNP Detail

SNP ID Chromosome Location
rs576384293 Chr.5: 150220783-150220783 on Build GRCh38
rs916442288 Chr.5: 150220552-150220552 on Build GRCh38
rs536039480 Chr.5: 150220609-150220609 on Build GRCh38
rs558100344 Chr.5: 150220772-150220772 on Build GRCh38
rs939297868 Chr.5: 150220540-150220540 on Build GRCh38
rs548048677 Chr.5: 150220636-150220636 on Build GRCh38
rs1037974565 Chr.5: 150220580-150220580 on Build GRCh38
rs531997567 Chr.5: 150220586-150220586 on Build GRCh38
rs947806324 Chr.5: 150220801-150220801 on Build GRCh38
rs746245033 Chr.5: 150220818-150220818 on Build GRCh38
rs1022286252 Chr.5: 150220695-150220695 on Build GRCh38
rs183021002 Chr.5: 150220604-150220604 on Build GRCh38
rs190785277 Chr.5: 150220460-150220460 on Build GRCh38
rs750759815 Chr.5: 150220714-150220715 on Build GRCh38
rs950978014 Chr.5: 150220694-150220722 on Build GRCh38
rs10038918 Chr.5: 150220513-150220513 on Build GRCh38
rs922744199 Chr.5: 150220452-150220452 on Build GRCh38
rs1044934713 Chr.5: 150220599-150220599 on Build GRCh38
rs960571695 Chr.5: 150220756-150220756 on Build GRCh38
rs759435022 Chr.5: 150220759-150220759 on Build GRCh38
rs964171474 Chr.5: 150220681-150220681 on Build GRCh38
rs914866203 Chr.5: 150220797-150220797 on Build GRCh38
rs563933970 Chr.5: 150220537-150220537 on Build GRCh38
rs547199963 Chr.5: 150220594-150220594 on Build GRCh38
rs548589134 Chr.5: 150220685-150220714 on Build GRCh38
rs975557952 Chr.5: 150220451-150220451 on Build GRCh38
rs924886614 Chr.5: 150220714-150220714 on Build GRCh38
rs559808389 Chr.5: 150220399-150220399 on Build GRCh38
rs769836752 Chr.5: 150220781-150220781 on Build GRCh38
rs1039291172 Chr.5: 150220662-150220662 on Build GRCh38
rs1035302847 Chr.5: 150220752-150220752 on Build GRCh38
rs906395380 Chr.5: 150220823-150220823 on Build GRCh38
rs1053986011 Chr.5: 150220533-150220533 on Build GRCh38
rs964434968 Chr.5: 150220420-150220420 on Build GRCh38
rs935006004 Chr.5: 150220517-150220517 on Build GRCh38
rs966812209 Chr.5: 150220708-150220708 on Build GRCh38
rs900721900 Chr.5: 150220682-150220682 on Build GRCh38
rs539560967 Chr.5: 150220751-150220751 on Build GRCh38
rs34655241 Chr.5: 150220696-150220697 on Build GRCh38
rs191256988 Chr.5: 150220842-150220842 on Build GRCh38
rs777408020 Chr.5: 150220442-150220442 on Build GRCh38
rs1017312269 Chr.5: 150220674-150220674 on Build GRCh38
rs987002884 Chr.5: 150220400-150220400 on Build GRCh38
rs186399006 Chr.5: 150220397-150220397 on Build GRCh38
rs999607514 Chr.5: 150220711-150220711 on Build GRCh38
rs941036595 Chr.5: 150220579-150220579 on Build GRCh38
rs1005552244 Chr.5: 150220665-150220665 on Build GRCh38
rs770372502 Chr.5: 150220546-150220546 on Build GRCh38
rs187985136 Chr.5: 150220667-150220667 on Build GRCh38
rs893559542 Chr.5: 150220590-150220590 on Build GRCh38
rs1010930482 Chr.5: 150220689-150220689 on Build GRCh38
rs530134149 Chr.5: 150220547-150220547 on Build GRCh38
rs977678343 Chr.5: 150220712-150220712 on Build GRCh38
rs906343663 Chr.5: 150220629-150220629 on Build GRCh38
rs890622182 Chr.5: 150220632-150220632 on Build GRCh38
rs939163112 Chr.5: 150220840-150220840 on Build GRCh38
rs918356656 Chr.5: 150220583-150220583 on Build GRCh38
rs1041932568 Chr.5: 150220505-150220505 on Build GRCh38
rs139169139 Chr.5: 150220404-150220404 on Build GRCh38

View Full Product Details