Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 150406177-150406675 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD74
Entrez Gene ID
972
Gene Name
CD74 molecule
Gene Aliases
DHLAG, HLADG, II, Ia-GAMMA
UniGene
Hs.436568
Gene Chromosome Location
Chr.5: 150401637-150412936 on Build GRCh38
Transcripts
NM_001025158.2 NM_001025159.2 NM_004355.3 XM_017010089.1 XM_017010090.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3244273 c.382A>G p.M128V Chr.5: 150406318-150406318 on Build GRCh38
1064368 c.393C>A p.A131A Chr.5: 150406307-150406307 on Build GRCh38
1064367 c.393C>A p.A131A Chr.5: 150406307-150406307 on Build GRCh38
5580579 c.426G>A p.V142V Chr.5: 150406274-150406274 on Build GRCh38
5580578 c.426G>A p.V142V Chr.5: 150406274-150406274 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs971740198 Chr.5: 150406384-150406384 on Build GRCh38
rs978906612 Chr.5: 150406481-150406481 on Build GRCh38
rs918845924 Chr.5: 150406420-150406420 on Build GRCh38
rs375451938 Chr.5: 150406372-150406372 on Build GRCh38
rs368149031 Chr.5: 150406363-150406363 on Build GRCh38
rs750733702 Chr.5: 150406324-150406324 on Build GRCh38
rs77280183 Chr.5: 150406361-150406361 on Build GRCh38
rs570637465 Chr.5: 150406428-150406428 on Build GRCh38
rs759725866 Chr.5: 150406632-150406632 on Build GRCh38
rs142317812 Chr.5: 150406280-150406280 on Build GRCh38
rs574786248 Chr.5: 150406473-150406473 on Build GRCh38
rs912694055 Chr.5: 150406474-150406474 on Build GRCh38
rs11548080 Chr.5: 150406306-150406306 on Build GRCh38
rs760450240 Chr.5: 150406267-150406267 on Build GRCh38
rs192080546 Chr.5: 150406472-150406472 on Build GRCh38
rs950302924 Chr.5: 150406641-150406641 on Build GRCh38
rs370923098 Chr.5: 150406211-150406211 on Build GRCh38
rs1003167720 Chr.5: 150406590-150406590 on Build GRCh38
rs781300005 Chr.5: 150406240-150406240 on Build GRCh38
rs1036525895 Chr.5: 150406643-150406643 on Build GRCh38
rs1042149325 Chr.5: 150406534-150406534 on Build GRCh38
rs751760132 Chr.5: 150406344-150406344 on Build GRCh38
rs143878156 Chr.5: 150406314-150406314 on Build GRCh38
rs762372687 Chr.5: 150406285-150406285 on Build GRCh38
rs79203186 Chr.5: 150406610-150406610 on Build GRCh38
rs1035623703 Chr.5: 150406595-150406595 on Build GRCh38
rs993966828 Chr.5: 150406244-150406244 on Build GRCh38
rs113615775 Chr.5: 150406257-150406257 on Build GRCh38
rs893253471 Chr.5: 150406491-150406491 on Build GRCh38
rs770830127 Chr.5: 150406248-150406248 on Build GRCh38
rs776761159 Chr.5: 150406485-150406485 on Build GRCh38
rs757150024 Chr.5: 150406369-150406369 on Build GRCh38
rs768748299 Chr.5: 150406236-150406236 on Build GRCh38
rs896165781 Chr.5: 150406205-150406205 on Build GRCh38
rs773774903 Chr.5: 150406318-150406318 on Build GRCh38
rs10050491 Chr.5: 150406371-150406371 on Build GRCh38
rs758721454 Chr.5: 150406330-150406330 on Build GRCh38
rs753394801 Chr.5: 150406339-150406340 on Build GRCh38
rs780203298 Chr.5: 150406332-150406332 on Build GRCh38
rs747051901 Chr.5: 150406233-150406233 on Build GRCh38
rs755258543 Chr.5: 150406350-150406350 on Build GRCh38
rs1039793019 Chr.5: 150406488-150406488 on Build GRCh38
rs780260228 Chr.5: 150406225-150406225 on Build GRCh38
rs991500994 Chr.5: 150406588-150406588 on Build GRCh38
rs201263243 Chr.5: 150406242-150406242 on Build GRCh38
rs758592249 Chr.5: 150406219-150406219 on Build GRCh38
rs903184848 Chr.5: 150406559-150406559 on Build GRCh38
rs367553129 Chr.5: 150406283-150406283 on Build GRCh38
rs760048319 Chr.5: 150406247-150406247 on Build GRCh38
rs774137900 Chr.5: 150406250-150406250 on Build GRCh38
rs1027064525 Chr.5: 150406256-150406256 on Build GRCh38
rs997957693 Chr.5: 150406421-150406421 on Build GRCh38
rs947361565 Chr.5: 150406500-150406500 on Build GRCh38
rs903132920 Chr.5: 150406227-150406227 on Build GRCh38
rs765937016 Chr.5: 150406339-150406341 on Build GRCh38
rs763669213 Chr.5: 150406274-150406274 on Build GRCh38
rs771806250 Chr.5: 150406253-150406253 on Build GRCh38
rs760691453 Chr.5: 150406370-150406370 on Build GRCh38
rs538233366 Chr.5: 150406465-150406465 on Build GRCh38
rs112717036 Chr.5: 150406258-150406258 on Build GRCh38
rs893119439 Chr.5: 150406203-150406203 on Build GRCh38
rs759253134 Chr.5: 150406252-150406252 on Build GRCh38
rs377471045 Chr.5: 150406209-150406209 on Build GRCh38

View Full Product Details