Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 150406782-150407286 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD74
Entrez Gene ID
972
Gene Name
CD74 molecule
Gene Aliases
DHLAG, HLADG, II, Ia-GAMMA
UniGene
Hs.436568
Gene Chromosome Location
Chr.5: 150401637-150412936 on Build GRCh38
Transcripts
NM_001025158.2 NM_001025159.2 NM_004355.3 XM_017010089.1 XM_017010090.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4762493 c.338T>C p.L113P Chr.5: 150406921-150406921 on Build GRCh38
1064370 c.281G>A p.R94H Chr.5: 150407169-150407169 on Build GRCh38
5356811 c.183C>T p.L61L Chr.5: 150407267-150407267 on Build GRCh38
1696574 c.349G>T p.A117S Chr.5: 150406910-150406910 on Build GRCh38
5807815 c.224A>T p.Q75L Chr.5: 150407226-150407226 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs552398847 Chr.5: 150407051-150407051 on Build GRCh38
rs766701141 Chr.5: 150406860-150406860 on Build GRCh38
rs752836949 Chr.5: 150406977-150406977 on Build GRCh38
rs377258430 Chr.5: 150407168-150407168 on Build GRCh38
rs924576055 Chr.5: 150406874-150406874 on Build GRCh38
rs116632913 Chr.5: 150406902-150406902 on Build GRCh38
rs766623079 Chr.5: 150406804-150406804 on Build GRCh38
rs747646655 Chr.5: 150406926-150406926 on Build GRCh38
rs139139083 Chr.5: 150406933-150406933 on Build GRCh38
rs754664980 Chr.5: 150407120-150407121 on Build GRCh38
rs371785500 Chr.5: 150406985-150406985 on Build GRCh38
rs375054104 Chr.5: 150406994-150406994 on Build GRCh38
rs548437679 Chr.5: 150406908-150406908 on Build GRCh38
rs200673075 Chr.5: 150407152-150407152 on Build GRCh38
rs1008593615 Chr.5: 150406963-150406963 on Build GRCh38
rs752553953 Chr.5: 150407225-150407227 on Build GRCh38
rs775462951 Chr.5: 150406833-150406833 on Build GRCh38
rs147685850 Chr.5: 150407220-150407220 on Build GRCh38
rs11548093 Chr.5: 150406890-150406890 on Build GRCh38
rs938675800 Chr.5: 150407097-150407097 on Build GRCh38
rs746367511 Chr.5: 150407254-150407254 on Build GRCh38
rs1013045954 Chr.5: 150407108-150407108 on Build GRCh38
rs778431769 Chr.5: 150407249-150407249 on Build GRCh38
rs777087097 Chr.5: 150407114-150407114 on Build GRCh38
rs756396039 Chr.5: 150406982-150406982 on Build GRCh38
rs375570446 Chr.5: 150406850-150406850 on Build GRCh38
rs151232598 Chr.5: 150406923-150406923 on Build GRCh38
rs760797280 Chr.5: 150407148-150407148 on Build GRCh38
rs374658696 Chr.5: 150406845-150406845 on Build GRCh38
rs765736118 Chr.5: 150406846-150406846 on Build GRCh38
rs1026545235 Chr.5: 150407128-150407128 on Build GRCh38
rs754984288 Chr.5: 150407208-150407208 on Build GRCh38
rs370173329 Chr.5: 150406868-150406868 on Build GRCh38
rs1049715496 Chr.5: 150406934-150406934 on Build GRCh38
rs527857815 Chr.5: 150407050-150407050 on Build GRCh38
rs531331869 Chr.5: 150406932-150406932 on Build GRCh38
rs771385899 Chr.5: 150407121-150407121 on Build GRCh38
rs13354393 Chr.5: 150407090-150407090 on Build GRCh38
rs762018826 Chr.5: 150407169-150407169 on Build GRCh38
rs879234501 Chr.5: 150407151-150407151 on Build GRCh38
rs899013309 Chr.5: 150406924-150406924 on Build GRCh38
rs767287388 Chr.5: 150406894-150406894 on Build GRCh38
rs780847683 Chr.5: 150407212-150407212 on Build GRCh38
rs112157732 Chr.5: 150406961-150406961 on Build GRCh38
rs912837055 Chr.5: 150406814-150406814 on Build GRCh38
rs751266362 Chr.5: 150406829-150406829 on Build GRCh38
rs368019468 Chr.5: 150406937-150406937 on Build GRCh38
rs200169434 Chr.5: 150406839-150406839 on Build GRCh38
rs142448212 Chr.5: 150407221-150407221 on Build GRCh38
rs71586172 Chr.5: 150407116-150407116 on Build GRCh38
rs772690375 Chr.5: 150407266-150407266 on Build GRCh38
rs748747022 Chr.5: 150407237-150407237 on Build GRCh38
rs752548987 Chr.5: 150407219-150407219 on Build GRCh38
rs780148338 Chr.5: 150406992-150406992 on Build GRCh38
rs763501745 Chr.5: 150406858-150406858 on Build GRCh38
rs538603918 Chr.5: 150407122-150407138 on Build GRCh38
rs1000442955 Chr.5: 150406816-150406816 on Build GRCh38
rs202092192 Chr.5: 150407167-150407167 on Build GRCh38
rs34845203 Chr.5: 150407001-150407002 on Build GRCh38
rs769558379 Chr.5: 150407012-150407012 on Build GRCh38
rs756167480 Chr.5: 150406893-150406893 on Build GRCh38
rs754032083 Chr.5: 150406987-150406987 on Build GRCh38
rs774860220 Chr.5: 150407122-150407122 on Build GRCh38
rs938712832 Chr.5: 150406885-150406885 on Build GRCh38
rs11548083 Chr.5: 150406895-150406895 on Build GRCh38
rs201039940 Chr.5: 150406838-150406838 on Build GRCh38
rs11548087 Chr.5: 150406886-150406886 on Build GRCh38
rs748799920 Chr.5: 150407120-150407120 on Build GRCh38
rs372583328 Chr.5: 150407137-150407137 on Build GRCh38
rs781091446 Chr.5: 150407005-150407005 on Build GRCh38
rs1033282034 Chr.5: 150407060-150407060 on Build GRCh38
rs777893918 Chr.5: 150407101-150407101 on Build GRCh38
rs779706134 Chr.5: 150406976-150406976 on Build GRCh38
rs11548085 Chr.5: 150407228-150407228 on Build GRCh38
rs766370704 Chr.5: 150407186-150407186 on Build GRCh38
rs758355473 Chr.5: 150406991-150406991 on Build GRCh38
rs774631004 Chr.5: 150406969-150406969 on Build GRCh38
rs373469571 Chr.5: 150407248-150407248 on Build GRCh38
rs772500133 Chr.5: 150407141-150407141 on Build GRCh38
rs751977067 Chr.5: 150406864-150406864 on Build GRCh38
rs977883151 Chr.5: 150407059-150407059 on Build GRCh38
rs766883449 Chr.5: 150406947-150406947 on Build GRCh38
rs1004978 Chr.5: 150407023-150407023 on Build GRCh38
rs751521954 Chr.5: 150407201-150407201 on Build GRCh38
rs145259888 Chr.5: 150406936-150406936 on Build GRCh38
rs767638817 Chr.5: 150406884-150406884 on Build GRCh38
rs200423334 Chr.5: 150407125-150407125 on Build GRCh38
rs184129716 Chr.5: 150406995-150406995 on Build GRCh38
rs111599852 Chr.5: 150407149-150407149 on Build GRCh38
rs746818222 Chr.5: 150406904-150406904 on Build GRCh38
rs760081229 Chr.5: 150407123-150407123 on Build GRCh38
rs747725118 Chr.5: 150407010-150407010 on Build GRCh38

View Full Product Details