Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 150412385-150412899 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD74
Entrez Gene ID
972
Gene Name
CD74 molecule
Gene Aliases
DHLAG, HLADG, II, Ia-GAMMA
UniGene
Hs.436568
Gene Chromosome Location
Chr.5: 150401637-150412936 on Build GRCh38
Transcripts
NM_001025158.2 NM_001025159.2 NM_004355.3 XM_017010089.1 XM_017010090.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1161525 c.98T>A p.L33Q Chr.5: 150412652-150412652 on Build GRCh38
5654045 c.18C>T p.S6S Chr.5: 150412732-150412732 on Build GRCh38
5714682 c.125G>A p.S42N Chr.5: 150412625-150412625 on Build GRCh38
1161527 c.52G>C p.D18H Chr.5: 150412698-150412698 on Build GRCh38
391889 c.12_14delGAG p.R5delR Chr.5: 150412736-150412738 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1006696292 Chr.5: 150412858-150412858 on Build GRCh38
rs1011445159 Chr.5: 150412642-150412642 on Build GRCh38
rs941560349 Chr.5: 150412511-150412511 on Build GRCh38
rs747519987 Chr.5: 150412655-150412655 on Build GRCh38
rs750068714 Chr.5: 150412741-150412741 on Build GRCh38
rs761630646 Chr.5: 150412728-150412728 on Build GRCh38
rs542872004 Chr.5: 150412506-150412506 on Build GRCh38
rs888250424 Chr.5: 150412854-150412854 on Build GRCh38
rs747291913 Chr.5: 150412582-150412582 on Build GRCh38
rs369383358 Chr.5: 150412584-150412584 on Build GRCh38
rs532157011 Chr.5: 150412569-150412569 on Build GRCh38
rs55796634 Chr.5: 150412774-150412774 on Build GRCh38
rs569954321 Chr.5: 150412761-150412762 on Build GRCh38
rs765912608 Chr.5: 150412758-150412758 on Build GRCh38
rs926068655 Chr.5: 150412675-150412675 on Build GRCh38
rs766648871 Chr.5: 150412767-150412767 on Build GRCh38
rs755488280 Chr.5: 150412766-150412766 on Build GRCh38
rs201392115 Chr.5: 150412478-150412478 on Build GRCh38
rs549957995 Chr.5: 150412509-150412514 on Build GRCh38
rs745950543 Chr.5: 150412785-150412785 on Build GRCh38
rs566731061 Chr.5: 150412622-150412622 on Build GRCh38
rs771012514 Chr.5: 150412688-150412688 on Build GRCh38
rs1051059677 Chr.5: 150412568-150412568 on Build GRCh38
rs961615819 Chr.5: 150412500-150412500 on Build GRCh38
rs749520760 Chr.5: 150412773-150412773 on Build GRCh38
rs747677901 Chr.5: 150412505-150412505 on Build GRCh38
rs958381277 Chr.5: 150412757-150412757 on Build GRCh38
rs776339297 Chr.5: 150412722-150412722 on Build GRCh38
rs781680991 Chr.5: 150412670-150412670 on Build GRCh38
rs201173431 Chr.5: 150412789-150412789 on Build GRCh38
rs751092204 Chr.5: 150412765-150412765 on Build GRCh38
rs777177244 Chr.5: 150412594-150412594 on Build GRCh38
rs867187273 Chr.5: 150412729-150412729 on Build GRCh38
rs112138942 Chr.5: 150412624-150412624 on Build GRCh38
rs756522112 Chr.5: 150412768-150412768 on Build GRCh38
rs372726927 Chr.5: 150412769-150412769 on Build GRCh38
rs530225455 Chr.5: 150412596-150412596 on Build GRCh38
rs758768447 Chr.5: 150412575-150412575 on Build GRCh38
rs892948207 Chr.5: 150412638-150412638 on Build GRCh38
rs879134540 Chr.5: 150412623-150412623 on Build GRCh38
rs763122934 Chr.5: 150412608-150412608 on Build GRCh38
rs200332442 Chr.5: 150412647-150412647 on Build GRCh38
rs770099469 Chr.5: 150412682-150412682 on Build GRCh38
rs760644191 Chr.5: 150412697-150412697 on Build GRCh38
rs757242063 Chr.5: 150412762-150412762 on Build GRCh38
rs760098975 Chr.5: 150412820-150412820 on Build GRCh38
rs746987244 Chr.5: 150412736-150412738 on Build GRCh38
rs7709772 Chr.5: 150412546-150412546 on Build GRCh38
rs761788303 Chr.5: 150412799-150412799 on Build GRCh38
rs373360044 Chr.5: 150412618-150412618 on Build GRCh38
rs768591195 Chr.5: 150412715-150412715 on Build GRCh38
rs916711659 Chr.5: 150412508-150412508 on Build GRCh38
rs777511692 Chr.5: 150412686-150412687 on Build GRCh38
rs770201603 Chr.5: 150412600-150412600 on Build GRCh38
rs879161212 Chr.5: 150412621-150412621 on Build GRCh38
rs780645975 Chr.5: 150412578-150412578 on Build GRCh38
rs557237719 Chr.5: 150412456-150412456 on Build GRCh38
rs575494438 Chr.5: 150412504-150412504 on Build GRCh38
rs918767033 Chr.5: 150412876-150412876 on Build GRCh38
rs747833097 Chr.5: 150412787-150412787 on Build GRCh38
rs538528058 Chr.5: 150412435-150412435 on Build GRCh38
rs749863505 Chr.5: 150412632-150412632 on Build GRCh38
rs748539271 Chr.5: 150412677-150412677 on Build GRCh38
rs767307882 Chr.5: 150412611-150412611 on Build GRCh38
rs11739355 Chr.5: 150412550-150412550 on Build GRCh38
rs778992754 Chr.5: 150412780-150412780 on Build GRCh38
rs774357257 Chr.5: 150412693-150412693 on Build GRCh38
rs11739357 Chr.5: 150412572-150412572 on Build GRCh38
rs540334325 Chr.5: 150412558-150412558 on Build GRCh38
rs760565382 Chr.5: 150412616-150412616 on Build GRCh38
rs991098610 Chr.5: 150412496-150412496 on Build GRCh38
rs527730828 Chr.5: 150412631-150412631 on Build GRCh38
rs556953829 Chr.5: 150412786-150412786 on Build GRCh38
rs1013684541 Chr.5: 150412449-150412449 on Build GRCh38
rs773357142 Chr.5: 150412605-150412605 on Build GRCh38
rs779483916 Chr.5: 150412649-150412649 on Build GRCh38
rs181380527 Chr.5: 150412563-150412563 on Build GRCh38
rs773505128 Chr.5: 150412686-150412686 on Build GRCh38
rs147417670 Chr.5: 150412614-150412614 on Build GRCh38
rs186335464 Chr.5: 150412739-150412739 on Build GRCh38
rs552660753 Chr.5: 150412659-150412659 on Build GRCh38

View Full Product Details