Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 151105103-151105600 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANXA6
Entrez Gene ID
309
Gene Name
annexin A6
Gene Aliases
ANX6, CBP68
UniGene
Hs.412117
Gene Chromosome Location
Chr.5: 151100706-151157882 on Build GRCh38
Transcripts
NM_001155.4 NM_001193544.1 XM_005268432.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1064497 c.1973T>C p.M658T Chr.5: 151105249-151105249 on Build GRCh38
1064498 c.1813G>A p.A605T Chr.5: 151105271-151105271 on Build GRCh38
1064499 c.1951G>A p.A651T Chr.5: 151105271-151105271 on Build GRCh38
259456 c.1816G>A p.D606N Chr.5: 151105268-151105268 on Build GRCh38
3852852 c.1817A>G p.D606G Chr.5: 151105267-151105267 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs111968671 Chr.5: 151105445-151105445 on Build GRCh38
rs893107652 Chr.5: 151105404-151105404 on Build GRCh38
rs367989243 Chr.5: 151105368-151105368 on Build GRCh38
rs781316382 Chr.5: 151105565-151105565 on Build GRCh38
rs765630739 Chr.5: 151105189-151105189 on Build GRCh38
rs371870585 Chr.5: 151105403-151105403 on Build GRCh38
rs760598858 Chr.5: 151105321-151105321 on Build GRCh38
rs967756638 Chr.5: 151105157-151105157 on Build GRCh38
rs962435952 Chr.5: 151105327-151105327 on Build GRCh38
rs766404858 Chr.5: 151105340-151105345 on Build GRCh38
rs764443329 Chr.5: 151105335-151105335 on Build GRCh38
rs1054574466 Chr.5: 151105400-151105400 on Build GRCh38
rs767527912 Chr.5: 151105227-151105227 on Build GRCh38
rs532637425 Chr.5: 151105518-151105518 on Build GRCh38
rs377051808 Chr.5: 151105310-151105310 on Build GRCh38
rs773606419 Chr.5: 151105290-151105290 on Build GRCh38
rs948787945 Chr.5: 151105419-151105419 on Build GRCh38
rs373940595 Chr.5: 151105258-151105258 on Build GRCh38
rs778595576 Chr.5: 151105256-151105256 on Build GRCh38
rs756879379 Chr.5: 151105250-151105250 on Build GRCh38
rs373769818 Chr.5: 151105367-151105367 on Build GRCh38
rs918147164 Chr.5: 151105354-151105354 on Build GRCh38
rs911334458 Chr.5: 151105446-151105446 on Build GRCh38
rs571764473 Chr.5: 151105496-151105496 on Build GRCh38
rs771610573 Chr.5: 151105268-151105268 on Build GRCh38
rs762027990 Chr.5: 151105198-151105198 on Build GRCh38
rs753583716 Chr.5: 151105249-151105249 on Build GRCh38
rs962691926 Chr.5: 151105511-151105511 on Build GRCh38
rs763954219 Chr.5: 151105325-151105325 on Build GRCh38
rs1020819493 Chr.5: 151105156-151105156 on Build GRCh38
rs1041090823 Chr.5: 151105521-151105521 on Build GRCh38
rs765389685 Chr.5: 151105216-151105216 on Build GRCh38
rs934759882 Chr.5: 151105393-151105393 on Build GRCh38
rs766514725 Chr.5: 151105308-151105308 on Build GRCh38
rs753156467 Chr.5: 151105269-151105269 on Build GRCh38
rs757236093 Chr.5: 151105433-151105433 on Build GRCh38
rs546787640 Chr.5: 151105495-151105495 on Build GRCh38
rs924652030 Chr.5: 151105391-151105391 on Build GRCh38
rs1028210091 Chr.5: 151105550-151105550 on Build GRCh38
rs982443127 Chr.5: 151105270-151105270 on Build GRCh38
rs770048443 Chr.5: 151105287-151105287 on Build GRCh38
rs776890648 Chr.5: 151105195-151105195 on Build GRCh38
rs959040617 Chr.5: 151105291-151105291 on Build GRCh38
rs955276939 Chr.5: 151105559-151105559 on Build GRCh38
rs1044619311 Chr.5: 151105429-151105429 on Build GRCh38
rs777587858 Chr.5: 151105235-151105235 on Build GRCh38
rs1034695181 Chr.5: 151105192-151105192 on Build GRCh38
rs1016709201 Chr.5: 151105507-151105507 on Build GRCh38
rs764840410 Chr.5: 151105344-151105344 on Build GRCh38
rs944017076 Chr.5: 151105461-151105461 on Build GRCh38
rs886831799 Chr.5: 151105488-151105488 on Build GRCh38
rs752726221 Chr.5: 151105231-151105231 on Build GRCh38
rs1002955294 Chr.5: 151105436-151105436 on Build GRCh38
rs921223166 Chr.5: 151105334-151105334 on Build GRCh38
rs745379492 Chr.5: 151105265-151105265 on Build GRCh38
rs763099195 Chr.5: 151105306-151105306 on Build GRCh38
rs868531 Chr.5: 151105328-151105328 on Build GRCh38
rs884520 Chr.5: 151105226-151105226 on Build GRCh38
rs748518293 Chr.5: 151105283-151105283 on Build GRCh38
rs975111815 Chr.5: 151105545-151105545 on Build GRCh38
rs548558963 Chr.5: 151105414-151105414 on Build GRCh38
rs757068455 Chr.5: 151105338-151105338 on Build GRCh38
rs776994035 Chr.5: 151105282-151105282 on Build GRCh38
rs368489179 Chr.5: 151105330-151105330 on Build GRCh38

View Full Product Details