Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 151182738-151183241 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC69
Entrez Gene ID
26112
Gene Name
coiled-coil domain containing 69
Gene Aliases
-
UniGene
Hs.655336
Gene Chromosome Location
Chr.5: 151181052-151224093 on Build GRCh38
Transcripts
NM_015621.2

SNP Detail

SNP ID Chromosome Location
rs188240405 Chr.5: 151182996-151182996 on Build GRCh38
rs576205711 Chr.5: 151182790-151182790 on Build GRCh38
rs976760321 Chr.5: 151182975-151182975 on Build GRCh38
rs530605440 Chr.5: 151183084-151183084 on Build GRCh38
rs532999352 Chr.5: 151183012-151183012 on Build GRCh38
rs567374355 Chr.5: 151183148-151183148 on Build GRCh38
rs1011929649 Chr.5: 151182775-151182775 on Build GRCh38
rs527776549 Chr.5: 151183136-151183137 on Build GRCh38
rs759676748 Chr.5: 151182990-151182990 on Build GRCh38
rs368768485 Chr.5: 151183139-151183139 on Build GRCh38
rs866064668 Chr.5: 151182900-151182900 on Build GRCh38
rs755206084 Chr.5: 151183075-151183075 on Build GRCh38
rs1022779915 Chr.5: 151182820-151182820 on Build GRCh38
rs1022748646 Chr.5: 151183128-151183128 on Build GRCh38
rs1025868235 Chr.5: 151182875-151182875 on Build GRCh38
rs969759991 Chr.5: 151182827-151182827 on Build GRCh38
rs1039202922 Chr.5: 151182924-151182924 on Build GRCh38
rs752787835 Chr.5: 151183132-151183132 on Build GRCh38
rs116559075 Chr.5: 151183157-151183157 on Build GRCh38
rs544027518 Chr.5: 151182842-151182842 on Build GRCh38
rs981193631 Chr.5: 151182841-151182841 on Build GRCh38
rs888241690 Chr.5: 151183178-151183178 on Build GRCh38
rs13184280 Chr.5: 151182965-151182965 on Build GRCh38
rs969897219 Chr.5: 151183134-151183134 on Build GRCh38
rs546527370 Chr.5: 151183216-151183216 on Build GRCh38
rs201824813 Chr.5: 151183137-151183137 on Build GRCh38
rs975026738 Chr.5: 151183189-151183189 on Build GRCh38
rs563401113 Chr.5: 151183051-151183051 on Build GRCh38
rs1031548576 Chr.5: 151182771-151182771 on Build GRCh38
rs772694172 Chr.5: 151183135-151183136 on Build GRCh38
rs977715481 Chr.5: 151183022-151183022 on Build GRCh38
rs781305595 Chr.5: 151183119-151183119 on Build GRCh38
rs4958903 Chr.5: 151182847-151182847 on Build GRCh38
rs184527148 Chr.5: 151182859-151182859 on Build GRCh38
rs906209800 Chr.5: 151183146-151183146 on Build GRCh38
rs984571756 Chr.5: 151182947-151182947 on Build GRCh38
rs779733995 Chr.5: 151183158-151183158 on Build GRCh38
rs1018771306 Chr.5: 151183199-151183199 on Build GRCh38
rs763656917 Chr.5: 151183087-151183087 on Build GRCh38
rs751758796 Chr.5: 151183061-151183061 on Build GRCh38
rs964004981 Chr.5: 151182966-151182966 on Build GRCh38
rs922134076 Chr.5: 151182946-151182946 on Build GRCh38
rs60344552 Chr.5: 151183136-151183136 on Build GRCh38
rs894766304 Chr.5: 151182773-151182773 on Build GRCh38
rs541362765 Chr.5: 151182928-151182928 on Build GRCh38
rs545124286 Chr.5: 151183030-151183030 on Build GRCh38
rs557875957 Chr.5: 151182766-151182766 on Build GRCh38
rs1005418018 Chr.5: 151183188-151183188 on Build GRCh38
rs906490578 Chr.5: 151182850-151182850 on Build GRCh38
rs964119782 Chr.5: 151183220-151183220 on Build GRCh38
rs559174748 Chr.5: 151182854-151182854 on Build GRCh38
rs201978887 Chr.5: 151183136-151183136 on Build GRCh38
rs1046976123 Chr.5: 151183167-151183167 on Build GRCh38
rs766851207 Chr.5: 151183035-151183035 on Build GRCh38
rs942306440 Chr.5: 151182878-151182878 on Build GRCh38
rs952593198 Chr.5: 151182896-151182896 on Build GRCh38
rs936186735 Chr.5: 151183060-151183060 on Build GRCh38
rs775657853 Chr.5: 151183132-151183132 on Build GRCh38
rs957534144 Chr.5: 151182789-151182789 on Build GRCh38

View Full Product Details