Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 151751625-151752133 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATOX1
Entrez Gene ID
475
Gene Name
antioxidant 1 copper chaperone
Gene Aliases
ATX1, HAH1
UniGene
Hs.125213
Gene Chromosome Location
Chr.5: 151742822-151758649 on Build GRCh38
Transcripts
NM_004045.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7290471 c.21T>C p.S7S Chr.5: 151751765-151751765 on Build GRCh38
3410018 c.71A>G p.N24S Chr.5: 151751715-151751715 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs936817499 Chr.5: 151751867-151751867 on Build GRCh38
rs1005990057 Chr.5: 151751954-151751954 on Build GRCh38
rs1016027081 Chr.5: 151751985-151751985 on Build GRCh38
rs924172111 Chr.5: 151751814-151751814 on Build GRCh38
rs374907691 Chr.5: 151751818-151751818 on Build GRCh38
rs773736280 Chr.5: 151751717-151751717 on Build GRCh38
rs757530682 Chr.5: 151751731-151751731 on Build GRCh38
rs1046943103 Chr.5: 151751690-151751690 on Build GRCh38
rs948263907 Chr.5: 151751897-151751897 on Build GRCh38
rs1026328584 Chr.5: 151751934-151751934 on Build GRCh38
rs774745102 Chr.5: 151751773-151751773 on Build GRCh38
rs1047581656 Chr.5: 151751933-151751933 on Build GRCh38
rs192927907 Chr.5: 151751653-151751653 on Build GRCh38
rs750255249 Chr.5: 151751665-151751665 on Build GRCh38
rs776167055 Chr.5: 151751657-151751657 on Build GRCh38
rs368016346 Chr.5: 151751807-151751807 on Build GRCh38
rs368468342 Chr.5: 151751969-151751969 on Build GRCh38
rs752186915 Chr.5: 151751810-151751810 on Build GRCh38
rs753145473 Chr.5: 151751970-151751970 on Build GRCh38
rs748703620 Chr.5: 151751715-151751715 on Build GRCh38
rs994624158 Chr.5: 151751705-151751705 on Build GRCh38
rs1026147447 Chr.5: 151751724-151751724 on Build GRCh38
rs377364525 Chr.5: 151751797-151751797 on Build GRCh38
rs1054071567 Chr.5: 151751869-151751869 on Build GRCh38
rs780615698 Chr.5: 151751711-151751711 on Build GRCh38
rs534614419 Chr.5: 151752109-151752109 on Build GRCh38
rs757080953 Chr.5: 151751655-151751655 on Build GRCh38
rs759761903 Chr.5: 151751774-151751774 on Build GRCh38
rs754514276 Chr.5: 151751710-151751710 on Build GRCh38
rs185513871 Chr.5: 151751995-151751995 on Build GRCh38
rs1018070760 Chr.5: 151752001-151752001 on Build GRCh38
rs558590378 Chr.5: 151751875-151751875 on Build GRCh38
rs371973236 Chr.5: 151751920-151751920 on Build GRCh38
rs767715613 Chr.5: 151751776-151751776 on Build GRCh38
rs1046399874 Chr.5: 151751901-151751901 on Build GRCh38
rs930113792 Chr.5: 151751919-151751919 on Build GRCh38
rs1055245370 Chr.5: 151751844-151751844 on Build GRCh38
rs758184590 Chr.5: 151751667-151751667 on Build GRCh38
rs564348343 Chr.5: 151752048-151752048 on Build GRCh38
rs886302863 Chr.5: 151751950-151751950 on Build GRCh38
rs996463185 Chr.5: 151752042-151752042 on Build GRCh38
rs371549947 Chr.5: 151751699-151751699 on Build GRCh38
rs1016122065 Chr.5: 151751795-151751795 on Build GRCh38
rs374713540 Chr.5: 151751701-151751701 on Build GRCh38
rs370455762 Chr.5: 151751798-151751798 on Build GRCh38
rs916817703 Chr.5: 151751883-151751883 on Build GRCh38
rs760802363 Chr.5: 151751794-151751794 on Build GRCh38
rs749662160 Chr.5: 151751725-151751725 on Build GRCh38
rs539287199 Chr.5: 151751809-151751809 on Build GRCh38
rs949194172 Chr.5: 151751888-151751888 on Build GRCh38
rs775487140 Chr.5: 151751786-151751786 on Build GRCh38
rs751218130 Chr.5: 151751808-151751808 on Build GRCh38
rs922901705 Chr.5: 151751801-151751801 on Build GRCh38
rs770264182 Chr.5: 151751716-151751716 on Build GRCh38
rs1006307986 Chr.5: 151751971-151751971 on Build GRCh38
rs75682555 Chr.5: 151751915-151751915 on Build GRCh38
rs936454865 Chr.5: 151751825-151751825 on Build GRCh38
rs547419437 Chr.5: 151751761-151751761 on Build GRCh38
rs571657964 Chr.5: 151751794-151751794 on Build GRCh38
rs925051927 Chr.5: 151751821-151751821 on Build GRCh38
rs964808669 Chr.5: 151751793-151751793 on Build GRCh38
rs755626530 Chr.5: 151751817-151751817 on Build GRCh38

View Full Product Details