Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 151800307-151800847 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
G3BP1
Entrez Gene ID
10146
Gene Name
G3BP stress granule assembly factor 1
Gene Aliases
G3BP, HDH-VIII
UniGene
Hs.3353
Gene Chromosome Location
Chr.5: 151771915-151811352 on Build GRCh38
Transcripts
NM_005754.2 NM_198395.1 XM_006714749.3 XM_006714750.3 XM_017008922.1 XM_017008923.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5585286 c.1077C>T p.F359F Chr.5: 151800339-151800339 on Build GRCh38
7341309 c.1116C>A p.N372K Chr.5: 151800791-151800791 on Build GRCh38
6362327 c.1075T>A p.F359I Chr.5: 151800337-151800337 on Build GRCh38
6356126 c.1167G>A p.E389E Chr.5: 151800842-151800842 on Build GRCh38
3410020 c.1107G>T p.L369F Chr.5: 151800782-151800782 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1014506405 Chr.5: 151800714-151800714 on Build GRCh38
rs778141231 Chr.5: 151800753-151800753 on Build GRCh38
rs534421928 Chr.5: 151800694-151800694 on Build GRCh38
rs200923414 Chr.5: 151800745-151800745 on Build GRCh38
rs201690048 Chr.5: 151800770-151800770 on Build GRCh38
rs911418105 Chr.5: 151800525-151800525 on Build GRCh38
rs909383779 Chr.5: 151800519-151800519 on Build GRCh38
rs372020718 Chr.5: 151800459-151800459 on Build GRCh38
rs767828974 Chr.5: 151800697-151800697 on Build GRCh38
rs917744464 Chr.5: 151800569-151800569 on Build GRCh38
rs927421529 Chr.5: 151800533-151800533 on Build GRCh38
rs757851083 Chr.5: 151800390-151800390 on Build GRCh38
rs1026736532 Chr.5: 151800409-151800409 on Build GRCh38
rs751022249 Chr.5: 151800725-151800725 on Build GRCh38
rs765858765 Chr.5: 151800721-151800721 on Build GRCh38
rs745694171 Chr.5: 151800726-151800727 on Build GRCh38
rs547724565 Chr.5: 151800510-151800510 on Build GRCh38
rs573639517 Chr.5: 151800689-151800689 on Build GRCh38
rs761573350 Chr.5: 151800371-151800371 on Build GRCh38
rs1036915331 Chr.5: 151800583-151800583 on Build GRCh38
rs919896165 Chr.5: 151800458-151800459 on Build GRCh38
rs3033225 Chr.5: 151800429-151800433 on Build GRCh38
rs551643401 Chr.5: 151800602-151800602 on Build GRCh38
rs368169057 Chr.5: 151800741-151800741 on Build GRCh38
rs529381425 Chr.5: 151800469-151800469 on Build GRCh38
rs530706989 Chr.5: 151800387-151800387 on Build GRCh38
rs750038506 Chr.5: 151800381-151800381 on Build GRCh38
rs953905729 Chr.5: 151800416-151800416 on Build GRCh38
rs780880879 Chr.5: 151800385-151800386 on Build GRCh38
rs913271524 Chr.5: 151800529-151800529 on Build GRCh38
rs555581937 Chr.5: 151800676-151800676 on Build GRCh38
rs1020553532 Chr.5: 151800683-151800683 on Build GRCh38
rs1014205735 Chr.5: 151800408-151800408 on Build GRCh38
rs961948384 Chr.5: 151800439-151800440 on Build GRCh38
rs1054989224 Chr.5: 151800551-151800551 on Build GRCh38
rs115448423 Chr.5: 151800468-151800468 on Build GRCh38
rs995150642 Chr.5: 151800594-151800594 on Build GRCh38
rs978800613 Chr.5: 151800700-151800700 on Build GRCh38
rs768119751 Chr.5: 151800471-151800471 on Build GRCh38
rs184148769 Chr.5: 151800377-151800377 on Build GRCh38
rs765135383 Chr.5: 151800784-151800784 on Build GRCh38
rs962282847 Chr.5: 151800737-151800737 on Build GRCh38
rs1055546414 Chr.5: 151800658-151800658 on Build GRCh38
rs57076413 Chr.5: 151800534-151800534 on Build GRCh38
rs776448858 Chr.5: 151800351-151800351 on Build GRCh38
rs745902782 Chr.5: 151800806-151800806 on Build GRCh38
rs549223894 Chr.5: 151800419-151800419 on Build GRCh38
rs367985973 Chr.5: 151800345-151800345 on Build GRCh38
rs904172138 Chr.5: 151800621-151800621 on Build GRCh38
rs889571491 Chr.5: 151800604-151800604 on Build GRCh38
rs774554057 Chr.5: 151800776-151800776 on Build GRCh38
rs965641859 Chr.5: 151800505-151800505 on Build GRCh38
rs755577956 Chr.5: 151800732-151800732 on Build GRCh38
rs187489514 Chr.5: 151800532-151800532 on Build GRCh38
rs1010376123 Chr.5: 151800644-151800644 on Build GRCh38
rs770829365 Chr.5: 151800717-151800717 on Build GRCh38
rs200853817 Chr.5: 151800760-151800760 on Build GRCh38
rs968716468 Chr.5: 151800531-151800531 on Build GRCh38
rs1015252234 Chr.5: 151800414-151800414 on Build GRCh38
rs781523306 Chr.5: 151800738-151800738 on Build GRCh38
rs1007034036 Chr.5: 151800619-151800619 on Build GRCh38
rs41290589 Chr.5: 151800579-151800579 on Build GRCh38
rs562275195 Chr.5: 151800710-151800710 on Build GRCh38
rs897037011 Chr.5: 151800588-151800588 on Build GRCh38
rs201241693 Chr.5: 151800822-151800822 on Build GRCh38
rs769676238 Chr.5: 151800781-151800782 on Build GRCh38
rs879219472 Chr.5: 151800684-151800684 on Build GRCh38
rs570042843 Chr.5: 151800638-151800638 on Build GRCh38
rs143691130 Chr.5: 151800708-151800708 on Build GRCh38
rs748836518 Chr.5: 151800449-151800449 on Build GRCh38
rs370209054 Chr.5: 151800640-151800640 on Build GRCh38
rs995099815 Chr.5: 151800403-151800403 on Build GRCh38
rs762265266 Chr.5: 151800625-151800625 on Build GRCh38

View Full Product Details