Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 151859692-151860236 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLRA1
Entrez Gene ID
2741
Gene Name
glycine receptor alpha 1
Gene Aliases
HKPX1, STHE
UniGene
Hs.121490
Gene Chromosome Location
Chr.5: 151820798-151924836 on Build GRCh38
Transcripts
NM_000171.3 NM_001146040.1 NM_001292000.1 XM_017009349.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6470098 c.395A>G p.K132R Chr.5: 151859866-151859866 on Build GRCh38
1162701 c.299G>A p.R100H Chr.5: 151859962-151859962 on Build GRCh38
1064664 c.277C>T p.R93W Chr.5: 151859984-151859984 on Build GRCh38
737153 c.437T>A p.L146Q Chr.5: 151859824-151859824 on Build GRCh38
1064663 c.408C>A p.F136L Chr.5: 151859853-151859853 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779404967 Chr.5: 151859742-151859742 on Build GRCh38
rs775236352 Chr.5: 151859965-151859965 on Build GRCh38
rs763726037 Chr.5: 151859974-151859974 on Build GRCh38
rs890639262 Chr.5: 151860173-151860173 on Build GRCh38
rs867853750 Chr.5: 151859876-151859876 on Build GRCh38
rs938809250 Chr.5: 151859720-151859720 on Build GRCh38
rs762489154 Chr.5: 151859937-151859937 on Build GRCh38
rs753405761 Chr.5: 151859978-151859978 on Build GRCh38
rs370889887 Chr.5: 151860175-151860175 on Build GRCh38
rs374949730 Chr.5: 151859951-151859951 on Build GRCh38
rs763601365 Chr.5: 151859958-151859958 on Build GRCh38
rs751965923 Chr.5: 151860032-151860032 on Build GRCh38
rs759191307 Chr.5: 151859787-151859787 on Build GRCh38
rs1050569733 Chr.5: 151860160-151860160 on Build GRCh38
rs202233543 Chr.5: 151859923-151859923 on Build GRCh38
rs907846992 Chr.5: 151859987-151859987 on Build GRCh38
rs375840161 Chr.5: 151859970-151859970 on Build GRCh38
rs776829161 Chr.5: 151859777-151859777 on Build GRCh38
rs553096050 Chr.5: 151860059-151860059 on Build GRCh38
rs557783325 Chr.5: 151859775-151859775 on Build GRCh38
rs955880571 Chr.5: 151859723-151859723 on Build GRCh38
rs561848502 Chr.5: 151859812-151859812 on Build GRCh38
rs543660999 Chr.5: 151859808-151859808 on Build GRCh38
rs932099226 Chr.5: 151859915-151859915 on Build GRCh38
rs775032574 Chr.5: 151859914-151859914 on Build GRCh38
rs138293464 Chr.5: 151860204-151860204 on Build GRCh38
rs959728098 Chr.5: 151860120-151860120 on Build GRCh38
rs760390019 Chr.5: 151859969-151859969 on Build GRCh38
rs117996103 Chr.5: 151860178-151860178 on Build GRCh38
rs779234204 Chr.5: 151859870-151859870 on Build GRCh38
rs752007761 Chr.5: 151860128-151860128 on Build GRCh38
rs940690636 Chr.5: 151860015-151860015 on Build GRCh38
rs371331129 Chr.5: 151859788-151859788 on Build GRCh38
rs199639315 Chr.5: 151859967-151859967 on Build GRCh38
rs539581717 Chr.5: 151859746-151859746 on Build GRCh38
rs758917994 Chr.5: 151860017-151860017 on Build GRCh38
rs748259870 Chr.5: 151860052-151860052 on Build GRCh38
rs6870491 Chr.5: 151859745-151859745 on Build GRCh38
rs769717473 Chr.5: 151860060-151860060 on Build GRCh38
rs866694759 Chr.5: 151859805-151859805 on Build GRCh38
rs757745596 Chr.5: 151860205-151860205 on Build GRCh38
rs1057116813 Chr.5: 151859727-151859727 on Build GRCh38
rs762986466 Chr.5: 151859779-151859779 on Build GRCh38
rs921998828 Chr.5: 151859906-151859906 on Build GRCh38
rs545897916 Chr.5: 151860132-151860132 on Build GRCh38
rs973462811 Chr.5: 151860061-151860061 on Build GRCh38
rs281864914 Chr.5: 151859962-151859962 on Build GRCh38
rs747286269 Chr.5: 151859883-151859883 on Build GRCh38
rs774085354 Chr.5: 151859955-151859955 on Build GRCh38
rs7729026 Chr.5: 151859729-151859729 on Build GRCh38
rs376001592 Chr.5: 151859764-151859764 on Build GRCh38
rs750848435 Chr.5: 151860016-151860016 on Build GRCh38
rs199547699 Chr.5: 151859984-151859984 on Build GRCh38
rs750107200 Chr.5: 151859883-151859883 on Build GRCh38
rs970190041 Chr.5: 151860064-151860064 on Build GRCh38
rs371496582 Chr.5: 151859828-151859828 on Build GRCh38
rs281864915 Chr.5: 151859963-151859963 on Build GRCh38
rs376416020 Chr.5: 151860024-151860026 on Build GRCh38
rs973540634 Chr.5: 151860199-151860199 on Build GRCh38
rs1035733049 Chr.5: 151860100-151860100 on Build GRCh38
rs369500388 Chr.5: 151859964-151859964 on Build GRCh38
rs374834686 Chr.5: 151859813-151859813 on Build GRCh38
rs781487245 Chr.5: 151859933-151859933 on Build GRCh38
rs143836612 Chr.5: 151860206-151860206 on Build GRCh38
rs764921483 Chr.5: 151859765-151859765 on Build GRCh38
rs148952106 Chr.5: 151860003-151860003 on Build GRCh38
rs774260091 Chr.5: 151859781-151859781 on Build GRCh38
rs983155451 Chr.5: 151860121-151860121 on Build GRCh38
rs960531612 Chr.5: 151859854-151859854 on Build GRCh38
rs758650282 Chr.5: 151859874-151859874 on Build GRCh38
rs752746595 Chr.5: 151860017-151860024 on Build GRCh38
rs374319634 Chr.5: 151859922-151859922 on Build GRCh38
rs143377552 Chr.5: 151859826-151859826 on Build GRCh38
rs781505806 Chr.5: 151860048-151860048 on Build GRCh38
rs780446903 Chr.5: 151859881-151859881 on Build GRCh38
rs755308567 Chr.5: 151860041-151860041 on Build GRCh38
rs766221222 Chr.5: 151859780-151859780 on Build GRCh38
rs757843415 Chr.5: 151859862-151859862 on Build GRCh38
rs185811794 Chr.5: 151860095-151860095 on Build GRCh38
rs112970419 Chr.5: 151859871-151859871 on Build GRCh38
rs757677190 Chr.5: 151859737-151859737 on Build GRCh38
rs908106565 Chr.5: 151860133-151860133 on Build GRCh38
rs575951950 Chr.5: 151859802-151859802 on Build GRCh38
rs780604003 Chr.5: 151860022-151860022 on Build GRCh38
rs281864912 Chr.5: 151859973-151859973 on Build GRCh38
rs1008065494 Chr.5: 151859966-151859966 on Build GRCh38

View Full Product Details