Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 153992693-153993240 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM114A2
Entrez Gene ID
10827
Gene Name
family with sequence similarity 114 member A2
Gene Aliases
133K02, C5orf3
UniGene
Hs.166551
Gene Chromosome Location
Chr.5: 153990131-154038938 on Build GRCh38
Transcripts
NM_001317993.1 NM_001317994.1 NM_001317995.1 NM_018691.3 XM_005268360.4 XM_005268361.4 XM_011537540.2 XM_011537541.2 XM_011537542.2 XM_017008967.1 XM_017008968.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
482388 c.1428C>G p.L476L Chr.5: 153993066-153993066 on Build GRCh38
4819704 c.1452C>G p.L484L Chr.5: 153993042-153993042 on Build GRCh38
449159 c.1483G>A p.E495K Chr.5: 153993011-153993011 on Build GRCh38
6432246 c.1414G>T p.A472S Chr.5: 153993080-153993080 on Build GRCh38
1328963 c.1509A>G p.L503L Chr.5: 153992985-153992985 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1048761134 Chr.5: 153992884-153992884 on Build GRCh38
rs762342947 Chr.5: 153992998-153992998 on Build GRCh38
rs370219072 Chr.5: 153993114-153993114 on Build GRCh38
rs934061294 Chr.5: 153993165-153993165 on Build GRCh38
rs753991388 Chr.5: 153993145-153993145 on Build GRCh38
rs929865599 Chr.5: 153992856-153992856 on Build GRCh38
rs949123446 Chr.5: 153992748-153992748 on Build GRCh38
rs981152427 Chr.5: 153993033-153993033 on Build GRCh38
rs76890735 Chr.5: 153993193-153993193 on Build GRCh38
rs577855017 Chr.5: 153992935-153992935 on Build GRCh38
rs1033654892 Chr.5: 153992978-153992978 on Build GRCh38
rs900359043 Chr.5: 153993206-153993206 on Build GRCh38
rs764847212 Chr.5: 153992980-153992980 on Build GRCh38
rs767891601 Chr.5: 153993138-153993138 on Build GRCh38
rs747422633 Chr.5: 153993122-153993123 on Build GRCh38
rs376900940 Chr.5: 153993069-153993069 on Build GRCh38
rs112040862 Chr.5: 153993004-153993004 on Build GRCh38
rs778104090 Chr.5: 153993143-153993143 on Build GRCh38
rs990857464 Chr.5: 153992989-153992989 on Build GRCh38
rs370402893 Chr.5: 153992860-153992860 on Build GRCh38
rs888389629 Chr.5: 153992889-153992889 on Build GRCh38
rs375885200 Chr.5: 153993187-153993187 on Build GRCh38
rs763978420 Chr.5: 153992962-153992962 on Build GRCh38
rs181593064 Chr.5: 153993203-153993203 on Build GRCh38
rs1046255451 Chr.5: 153992797-153992797 on Build GRCh38
rs758802296 Chr.5: 153993017-153993018 on Build GRCh38
rs937647005 Chr.5: 153992736-153992736 on Build GRCh38
rs918447493 Chr.5: 153992836-153992836 on Build GRCh38
rs528100856 Chr.5: 153993090-153993090 on Build GRCh38
rs749424650 Chr.5: 153993035-153993035 on Build GRCh38
rs770891795 Chr.5: 153993036-153993036 on Build GRCh38
rs760560154 Chr.5: 153992957-153992957 on Build GRCh38
rs759979323 Chr.5: 153993133-153993133 on Build GRCh38
rs981886756 Chr.5: 153993038-153993038 on Build GRCh38
rs559466789 Chr.5: 153992912-153992912 on Build GRCh38
rs909758019 Chr.5: 153993094-153993094 on Build GRCh38
rs1020389523 Chr.5: 153992946-153992946 on Build GRCh38
rs546557317 Chr.5: 153993122-153993122 on Build GRCh38
rs6866471 Chr.5: 153993183-153993183 on Build GRCh38
rs188786544 Chr.5: 153992936-153992936 on Build GRCh38
rs551072940 Chr.5: 153992955-153992955 on Build GRCh38
rs147853164 Chr.5: 153993081-153993081 on Build GRCh38
rs780300198 Chr.5: 153992933-153992933 on Build GRCh38
rs1041754035 Chr.5: 153993181-153993181 on Build GRCh38
rs927536702 Chr.5: 153992727-153992727 on Build GRCh38
rs114815094 Chr.5: 153993202-153993202 on Build GRCh38
rs561638008 Chr.5: 153993005-153993005 on Build GRCh38
rs542828691 Chr.5: 153993176-153993176 on Build GRCh38
rs199650757 Chr.5: 153993142-153993142 on Build GRCh38
rs371479761 Chr.5: 153992952-153992952 on Build GRCh38
rs373727986 Chr.5: 153993037-153993037 on Build GRCh38
rs916276521 Chr.5: 153992745-153992745 on Build GRCh38
rs745823152 Chr.5: 153992947-153992947 on Build GRCh38
rs369423237 Chr.5: 153993028-153993028 on Build GRCh38
rs768572365 Chr.5: 153993067-153993067 on Build GRCh38
rs2560049 Chr.5: 153993016-153993016 on Build GRCh38
rs761576819 Chr.5: 153993071-153993071 on Build GRCh38
rs369207860 Chr.5: 153992943-153992943 on Build GRCh38
rs1008738900 Chr.5: 153992890-153992890 on Build GRCh38
rs202197693 Chr.5: 153992986-153992986 on Build GRCh38
rs752113768 Chr.5: 153993129-153993129 on Build GRCh38
rs762585292 Chr.5: 153993113-153993113 on Build GRCh38
rs891183175 Chr.5: 153992822-153992822 on Build GRCh38
rs778371483 Chr.5: 153993062-153993063 on Build GRCh38
rs146086196 Chr.5: 153993010-153993010 on Build GRCh38
rs748354543 Chr.5: 153992938-153992938 on Build GRCh38
rs757423481 Chr.5: 153993146-153993146 on Build GRCh38
rs201693208 Chr.5: 153993012-153993012 on Build GRCh38
rs1057772 Chr.5: 153992965-153992965 on Build GRCh38
rs746913963 Chr.5: 153993049-153993049 on Build GRCh38
rs375972557 Chr.5: 153992942-153992942 on Build GRCh38
rs946019105 Chr.5: 153992764-153992764 on Build GRCh38
rs766962060 Chr.5: 153993003-153993003 on Build GRCh38
rs990517666 Chr.5: 153992718-153992718 on Build GRCh38
rs942669796 Chr.5: 153993110-153993110 on Build GRCh38
rs776486256 Chr.5: 153992964-153992964 on Build GRCh38

View Full Product Details