Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 154865082-154865500 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNOT8
Entrez Gene ID
9337
Gene Name
CCR4-NOT transcription complex subunit 8
Gene Aliases
CAF1, CALIF, Caf1b, POP2, hCAF1
UniGene
Hs.26703
Gene Chromosome Location
Chr.5: 154857952-154876792 on Build GRCh38
Transcripts
NM_001301073.1 NM_001301074.1 NM_001301075.1 NM_001301077.1 NM_001301080.1 NM_001301082.1 NM_001301083.1 NM_001301086.1 NM_004779.5 XM_005268527.2 XM_011537706.1 XM_011537708.2 XM_017010048.1 XM_017010049.1 XM_017010050.1 XM_017010051.1 XM_017010052.1 XM_017010053.1 XM_017010054.1 XM_017010055.1 XM_017010056.1 XM_017010057.1 XM_017010058.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6631563 c.164G>A p.R55H Chr.5: 154865238-154865238 on Build GRCh38
3827516 c.205G>A p.V69I Chr.5: 154865279-154865279 on Build GRCh38
1064760 c.250G>A p.E84K Chr.5: 154865324-154865324 on Build GRCh38
6437710 c.274A>C p.I92L Chr.5: 154865348-154865348 on Build GRCh38
7165972 c.297C>G p.F99L Chr.5: 154865371-154865371 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773036044 Chr.5: 154865151-154865151 on Build GRCh38
rs774037667 Chr.5: 154865162-154865162 on Build GRCh38
rs773063803 Chr.5: 154865146-154865146 on Build GRCh38
rs771635799 Chr.5: 154865359-154865359 on Build GRCh38
rs1026909104 Chr.5: 154865241-154865241 on Build GRCh38
rs750700906 Chr.5: 154865238-154865238 on Build GRCh38
rs746875605 Chr.5: 154865363-154865363 on Build GRCh38
rs899265035 Chr.5: 154865197-154865197 on Build GRCh38
rs763685629 Chr.5: 154865386-154865386 on Build GRCh38
rs759324217 Chr.5: 154865352-154865352 on Build GRCh38
rs375825981 Chr.5: 154865154-154865154 on Build GRCh38
rs755591818 Chr.5: 154865189-154865189 on Build GRCh38
rs201058556 Chr.5: 154865408-154865408 on Build GRCh38
rs976161339 Chr.5: 154865379-154865379 on Build GRCh38
rs777741922 Chr.5: 154865278-154865278 on Build GRCh38
rs549871153 Chr.5: 154865149-154865149 on Build GRCh38
rs371364378 Chr.5: 154865335-154865335 on Build GRCh38
rs775140622 Chr.5: 154865367-154865367 on Build GRCh38
rs765802788 Chr.5: 154865417-154865417 on Build GRCh38
rs990502494 Chr.5: 154865459-154865459 on Build GRCh38
rs200072532 Chr.5: 154865336-154865336 on Build GRCh38
rs915564667 Chr.5: 154865460-154865460 on Build GRCh38
rs544730710 Chr.5: 154865409-154865409 on Build GRCh38
rs965095664 Chr.5: 154865305-154865305 on Build GRCh38
rs752170679 Chr.5: 154865187-154865187 on Build GRCh38
rs759078726 Chr.5: 154865164-154865164 on Build GRCh38
rs773112070 Chr.5: 154865469-154865469 on Build GRCh38
rs1030453960 Chr.5: 154865394-154865394 on Build GRCh38
rs185861935 Chr.5: 154865454-154865454 on Build GRCh38
rs760985144 Chr.5: 154865474-154865474 on Build GRCh38
rs758735466 Chr.5: 154865436-154865436 on Build GRCh38
rs770986870 Chr.5: 154865411-154865420 on Build GRCh38
rs1017589710 Chr.5: 154865136-154865136 on Build GRCh38
rs1006895217 Chr.5: 154865121-154865121 on Build GRCh38
rs768773408 Chr.5: 154865272-154865272 on Build GRCh38
rs766938064 Chr.5: 154865311-154865312 on Build GRCh38
rs764703838 Chr.5: 154865205-154865205 on Build GRCh38
rs747075581 Chr.5: 154865250-154865250 on Build GRCh38
rs569679779 Chr.5: 154865246-154865246 on Build GRCh38
rs757756259 Chr.5: 154865225-154865225 on Build GRCh38
rs779400145 Chr.5: 154865230-154865230 on Build GRCh38
rs758582468 Chr.5: 154865239-154865239 on Build GRCh38
rs749199415 Chr.5: 154865311-154865311 on Build GRCh38
rs754392052 Chr.5: 154865219-154865219 on Build GRCh38
rs980192051 Chr.5: 154865456-154865456 on Build GRCh38
rs767156186 Chr.5: 154865172-154865172 on Build GRCh38
rs977213205 Chr.5: 154865421-154865421 on Build GRCh38
rs750779265 Chr.5: 154865430-154865430 on Build GRCh38
rs149227642 Chr.5: 154865446-154865446 on Build GRCh38
rs943394560 Chr.5: 154865150-154865150 on Build GRCh38
rs532346031 Chr.5: 154865116-154865116 on Build GRCh38

View Full Product Details