Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 154901009-154901558 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GEMIN5
Entrez Gene ID
25929
Gene Name
gem nuclear organelle associated protein 5
Gene Aliases
GEMIN-5
UniGene
Hs.483921
Gene Chromosome Location
Chr.5: 154887416-154938216 on Build GRCh38
Transcripts
NM_001252156.1 NM_015465.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3735700 c.2942C>T p.A981V Chr.5: 154901411-154901411 on Build GRCh38
1435451 c.3005A>G p.H1002R Chr.5: 154901348-154901348 on Build GRCh38
3373757 c.2962A>T p.I988F Chr.5: 154901391-154901391 on Build GRCh38
3784672 c.2883G>T p.W961C Chr.5: 154901470-154901470 on Build GRCh38
6529669 c.2876A>G p.H959R Chr.5: 154901477-154901477 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373378229 Chr.5: 154901359-154901359 on Build GRCh38
rs74803058 Chr.5: 154901508-154901508 on Build GRCh38
rs183066599 Chr.5: 154901101-154901101 on Build GRCh38
rs1005990717 Chr.5: 154901068-154901068 on Build GRCh38
rs1020581613 Chr.5: 154901141-154901141 on Build GRCh38
rs1010395553 Chr.5: 154901127-154901127 on Build GRCh38
rs190972976 Chr.5: 154901099-154901099 on Build GRCh38
rs916613628 Chr.5: 154901154-154901154 on Build GRCh38
rs777940320 Chr.5: 154901296-154901296 on Build GRCh38
rs777275234 Chr.5: 154901496-154901496 on Build GRCh38
rs1048084723 Chr.5: 154901226-154901226 on Build GRCh38
rs776310731 Chr.5: 154901339-154901339 on Build GRCh38
rs144546733 Chr.5: 154901162-154901162 on Build GRCh38
rs1032976763 Chr.5: 154901529-154901529 on Build GRCh38
rs768016683 Chr.5: 154901403-154901403 on Build GRCh38
rs929233679 Chr.5: 154901223-154901223 on Build GRCh38
rs369151963 Chr.5: 154901288-154901288 on Build GRCh38
rs746870507 Chr.5: 154901332-154901332 on Build GRCh38
rs529777236 Chr.5: 154901477-154901477 on Build GRCh38
rs565205023 Chr.5: 154901377-154901377 on Build GRCh38
rs754010625 Chr.5: 154901430-154901430 on Build GRCh38
rs749260647 Chr.5: 154901313-154901313 on Build GRCh38
rs1051909479 Chr.5: 154901423-154901423 on Build GRCh38
rs1001598182 Chr.5: 154901106-154901106 on Build GRCh38
rs142807733 Chr.5: 154901066-154901066 on Build GRCh38
rs768077387 Chr.5: 154901098-154901098 on Build GRCh38
rs941660291 Chr.5: 154901316-154901316 on Build GRCh38
rs900297407 Chr.5: 154901327-154901327 on Build GRCh38
rs773950112 Chr.5: 154901525-154901525 on Build GRCh38
rs1032366448 Chr.5: 154901114-154901114 on Build GRCh38
rs769499034 Chr.5: 154901479-154901479 on Build GRCh38
rs186687071 Chr.5: 154901109-154901109 on Build GRCh38
rs929899283 Chr.5: 154901059-154901059 on Build GRCh38
rs774969772 Chr.5: 154901372-154901372 on Build GRCh38
rs751473417 Chr.5: 154901453-154901453 on Build GRCh38
rs191440006 Chr.5: 154901200-154901200 on Build GRCh38
rs761144694 Chr.5: 154901113-154901113 on Build GRCh38
rs908893405 Chr.5: 154901249-154901249 on Build GRCh38
rs748871139 Chr.5: 154901503-154901503 on Build GRCh38
rs760959818 Chr.5: 154901420-154901420 on Build GRCh38
rs752967948 Chr.5: 154901411-154901411 on Build GRCh38
rs760029026 Chr.5: 154901391-154901391 on Build GRCh38
rs758370707 Chr.5: 154901322-154901322 on Build GRCh38
rs986465130 Chr.5: 154901156-154901156 on Build GRCh38
rs764167437 Chr.5: 154901425-154901425 on Build GRCh38
rs11167703 Chr.5: 154901163-154901163 on Build GRCh38
rs928440232 Chr.5: 154901181-154901181 on Build GRCh38
rs772782258 Chr.5: 154901369-154901369 on Build GRCh38
rs140416900 Chr.5: 154901433-154901433 on Build GRCh38
rs919650397 Chr.5: 154901216-154901216 on Build GRCh38
rs958255766 Chr.5: 154901115-154901115 on Build GRCh38
rs866457096 Chr.5: 154901354-154901354 on Build GRCh38
rs1017745717 Chr.5: 154901096-154901096 on Build GRCh38
rs754856830 Chr.5: 154901460-154901460 on Build GRCh38
rs902670653 Chr.5: 154901506-154901506 on Build GRCh38
rs990933298 Chr.5: 154901123-154901123 on Build GRCh38
rs760885717 Chr.5: 154901105-154901105 on Build GRCh38
rs768406354 Chr.5: 154901336-154901336 on Build GRCh38
rs779955359 Chr.5: 154901324-154901324 on Build GRCh38
rs769290970 Chr.5: 154901365-154901365 on Build GRCh38
rs758978840 Chr.5: 154901323-154901324 on Build GRCh38

View Full Product Details