Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 157285251-157285753 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYFIP2
Entrez Gene ID
26999
Gene Name
cytoplasmic FMR1 interacting protein 2
Gene Aliases
PIR121
UniGene
Hs.519702
Gene Chromosome Location
Chr.5: 157266080-157395598 on Build GRCh38
Transcripts
NM_001037333.2 NM_001291721.1 NM_001291722.1 NM_014376.3 XM_011534516.2 XM_017009341.1 XM_017009342.1

SNP Detail

SNP ID Chromosome Location
rs771763167 Chr.5: 157285318-157285318 on Build GRCh38
rs141082748 Chr.5: 157285614-157285614 on Build GRCh38
rs940319794 Chr.5: 157285600-157285600 on Build GRCh38
rs188567647 Chr.5: 157285275-157285275 on Build GRCh38
rs1005466588 Chr.5: 157285695-157285695 on Build GRCh38
rs376687042 Chr.5: 157285400-157285400 on Build GRCh38
rs979678083 Chr.5: 157285608-157285608 on Build GRCh38
rs1019609938 Chr.5: 157285651-157285651 on Build GRCh38
rs968323449 Chr.5: 157285545-157285545 on Build GRCh38
rs371357141 Chr.5: 157285674-157285674 on Build GRCh38
rs377540260 Chr.5: 157285612-157285612 on Build GRCh38
rs903088370 Chr.5: 157285644-157285644 on Build GRCh38
rs750421212 Chr.5: 157285403-157285403 on Build GRCh38
rs78461959 Chr.5: 157285540-157285540 on Build GRCh38
rs761596947 Chr.5: 157285391-157285391 on Build GRCh38
rs757626231 Chr.5: 157285463-157285463 on Build GRCh38
rs371578292 Chr.5: 157285370-157285370 on Build GRCh38
rs749370451 Chr.5: 157285606-157285606 on Build GRCh38
rs775257977 Chr.5: 157285349-157285349 on Build GRCh38
rs112527842 Chr.5: 157285724-157285724 on Build GRCh38
rs1021631734 Chr.5: 157285341-157285341 on Build GRCh38
rs778978138 Chr.5: 157285649-157285649 on Build GRCh38
rs776506102 Chr.5: 157285373-157285373 on Build GRCh38
rs907046718 Chr.5: 157285276-157285276 on Build GRCh38
rs752822676 Chr.5: 157285430-157285430 on Build GRCh38
rs894433345 Chr.5: 157285331-157285331 on Build GRCh38
rs151110759 Chr.5: 157285535-157285535 on Build GRCh38
rs1002688153 Chr.5: 157285277-157285277 on Build GRCh38
rs879875568 Chr.5: 157285406-157285406 on Build GRCh38
rs145170129 Chr.5: 157285661-157285661 on Build GRCh38
rs755029730 Chr.5: 157285409-157285409 on Build GRCh38
rs777945352 Chr.5: 157285433-157285433 on Build GRCh38
rs760411094 Chr.5: 157285360-157285360 on Build GRCh38
rs563688080 Chr.5: 157285728-157285728 on Build GRCh38
rs908895113 Chr.5: 157285573-157285573 on Build GRCh38
rs1055563913 Chr.5: 157285615-157285615 on Build GRCh38
rs568084859 Chr.5: 157285300-157285300 on Build GRCh38
rs916715778 Chr.5: 157285543-157285543 on Build GRCh38
rs760770557 Chr.5: 157285594-157285594 on Build GRCh38
rs386694005 Chr.5: 157285727-157285728 on Build GRCh38
rs571089246 Chr.5: 157285528-157285528 on Build GRCh38
rs369640959 Chr.5: 157285472-157285472 on Build GRCh38
rs1028497949 Chr.5: 157285703-157285703 on Build GRCh38
rs1011416477 Chr.5: 157285338-157285338 on Build GRCh38
rs767625240 Chr.5: 157285412-157285412 on Build GRCh38
rs397765583 Chr.5: 157285726-157285727 on Build GRCh38
rs377704391 Chr.5: 157285273-157285273 on Build GRCh38
rs371710290 Chr.5: 157285470-157285470 on Build GRCh38
rs367998009 Chr.5: 157285442-157285442 on Build GRCh38
rs921423992 Chr.5: 157285482-157285482 on Build GRCh38
rs1011468753 Chr.5: 157285633-157285633 on Build GRCh38
rs555405104 Chr.5: 157285619-157285619 on Build GRCh38
rs116130695 Chr.5: 157285642-157285642 on Build GRCh38
rs998040599 Chr.5: 157285301-157285301 on Build GRCh38
rs779546387 Chr.5: 157285508-157285508 on Build GRCh38

View Full Product Details