Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 157286928-157287425 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYFIP2
Entrez Gene ID
26999
Gene Name
cytoplasmic FMR1 interacting protein 2
Gene Aliases
PIR121
UniGene
Hs.519702
Gene Chromosome Location
Chr.5: 157266080-157395598 on Build GRCh38
Transcripts
NM_001037333.2 NM_001291721.1 NM_001291722.1 NM_014376.3 XM_011534516.2 XM_017009341.1 XM_017009342.1

SNP Detail

SNP ID Chromosome Location
rs933100955 Chr.5: 157287396-157287397 on Build GRCh38
rs953759441 Chr.5: 157287274-157287274 on Build GRCh38
rs1014332278 Chr.5: 157287345-157287345 on Build GRCh38
rs771673397 Chr.5: 157286985-157286985 on Build GRCh38
rs373637240 Chr.5: 157287125-157287125 on Build GRCh38
rs1037902511 Chr.5: 157287230-157287230 on Build GRCh38
rs1052792290 Chr.5: 157287244-157287244 on Build GRCh38
rs891426858 Chr.5: 157287246-157287246 on Build GRCh38
rs765066355 Chr.5: 157286999-157286999 on Build GRCh38
rs1052078915 Chr.5: 157287178-157287178 on Build GRCh38
rs904142495 Chr.5: 157287212-157287212 on Build GRCh38
rs1000176577 Chr.5: 157287240-157287240 on Build GRCh38
rs375350435 Chr.5: 157287063-157287063 on Build GRCh38
rs757670706 Chr.5: 157287072-157287072 on Build GRCh38
rs752839483 Chr.5: 157287014-157287014 on Build GRCh38
rs769539457 Chr.5: 157287127-157287127 on Build GRCh38
rs371433380 Chr.5: 157286996-157286996 on Build GRCh38
rs557059352 Chr.5: 157287176-157287176 on Build GRCh38
rs2288068 Chr.5: 157287128-157287128 on Build GRCh38
rs1006765578 Chr.5: 157287275-157287275 on Build GRCh38
rs997950346 Chr.5: 157287258-157287258 on Build GRCh38
rs375599223 Chr.5: 157287279-157287279 on Build GRCh38
rs933048695 Chr.5: 157287212-157287212 on Build GRCh38
rs149452827 Chr.5: 157287354-157287354 on Build GRCh38
rs17655440 Chr.5: 157287198-157287198 on Build GRCh38
rs766961343 Chr.5: 157287075-157287075 on Build GRCh38
rs913535866 Chr.5: 157287390-157287390 on Build GRCh38
rs369941644 Chr.5: 157287099-157287099 on Build GRCh38
rs534811222 Chr.5: 157287005-157287005 on Build GRCh38
rs370614335 Chr.5: 157287142-157287142 on Build GRCh38
rs372488845 Chr.5: 157287124-157287124 on Build GRCh38
rs779373166 Chr.5: 157287105-157287105 on Build GRCh38
rs889276207 Chr.5: 157287219-157287219 on Build GRCh38
rs774223297 Chr.5: 157287145-157287145 on Build GRCh38
rs762823896 Chr.5: 157287141-157287141 on Build GRCh38
rs777602957 Chr.5: 157287380-157287380 on Build GRCh38
rs138001953 Chr.5: 157287165-157287165 on Build GRCh38
rs748285949 Chr.5: 157287378-157287378 on Build GRCh38
rs762910079 Chr.5: 157287002-157287002 on Build GRCh38
rs768366793 Chr.5: 157286994-157286994 on Build GRCh38
rs948250670 Chr.5: 157287194-157287194 on Build GRCh38
rs993623483 Chr.5: 157287004-157287004 on Build GRCh38
rs775170340 Chr.5: 157286990-157286990 on Build GRCh38
rs760792343 Chr.5: 157287150-157287150 on Build GRCh38
rs189060554 Chr.5: 157287167-157287167 on Build GRCh38
rs367682322 Chr.5: 157286992-157286992 on Build GRCh38
rs143013338 Chr.5: 157287359-157287359 on Build GRCh38
rs372917531 Chr.5: 157287164-157287164 on Build GRCh38
rs939441424 Chr.5: 157287153-157287153 on Build GRCh38
rs916761102 Chr.5: 157287179-157287179 on Build GRCh38
rs764126246 Chr.5: 157287152-157287152 on Build GRCh38
rs987219094 Chr.5: 157287397-157287397 on Build GRCh38
rs1007787398 Chr.5: 157287224-157287224 on Build GRCh38
rs746422045 Chr.5: 157286974-157286974 on Build GRCh38
rs756226998 Chr.5: 157287029-157287029 on Build GRCh38
rs746532639 Chr.5: 157287119-157287119 on Build GRCh38
rs746333698 Chr.5: 157287078-157287078 on Build GRCh38
rs988217041 Chr.5: 157287387-157287387 on Build GRCh38
rs895895386 Chr.5: 157287336-157287336 on Build GRCh38
rs572240200 Chr.5: 157287361-157287361 on Build GRCh38
rs555499512 Chr.5: 157287261-157287261 on Build GRCh38
rs955806468 Chr.5: 157287054-157287054 on Build GRCh38
rs565892801 Chr.5: 157286957-157286957 on Build GRCh38
rs761657699 Chr.5: 157286997-157286997 on Build GRCh38
rs780125706 Chr.5: 157287364-157287364 on Build GRCh38

View Full Product Details