Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 158730984-158731493 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EBF1
Entrez Gene ID
1879
Gene Name
early B-cell factor 1
Gene Aliases
COE1, EBF, O/E-1, OLF1
UniGene
Hs.573143
Gene Chromosome Location
Chr.5: 158695915-159099762 on Build GRCh38
Transcripts
NM_001290360.2 NM_001324101.1 NM_001324103.1 NM_001324106.1 NM_001324107.1 NM_001324108.1 NM_001324109.1 NM_001324111.1 NM_024007.4 NM_182708.2 XM_017009192.1 XM_017009193.1 XM_017009194.1 XM_017009195.1 XM_017009196.1 XM_017009197.1 XM_017009198.1 XM_017009199.1 XM_017009200.1 XM_017009201.1 XM_017009204.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1619957 c.1125+6G>C p.? Chr.5: 158731063-158731063 on Build GRCh38
379719 c.1098C>A p.H366Q Chr.5: 158731096-158731096 on Build GRCh38
6170392 c.1125G>T p.K375N Chr.5: 158731069-158731069 on Build GRCh38
6651129 c.1116T>C p.R372R Chr.5: 158731078-158731078 on Build GRCh38
4935437 c.1045G>C p.E349Q Chr.5: 158731149-158731149 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs921012466 Chr.5: 158731356-158731356 on Build GRCh38
rs377318158 Chr.5: 158731463-158731463 on Build GRCh38
rs765704187 Chr.5: 158731061-158731062 on Build GRCh38
rs532751199 Chr.5: 158731164-158731164 on Build GRCh38
rs767428260 Chr.5: 158731096-158731096 on Build GRCh38
rs896839297 Chr.5: 158731010-158731010 on Build GRCh38
rs973920875 Chr.5: 158731309-158731309 on Build GRCh38
rs554035459 Chr.5: 158731345-158731345 on Build GRCh38
rs1033178505 Chr.5: 158731422-158731422 on Build GRCh38
rs186847533 Chr.5: 158731184-158731184 on Build GRCh38
rs559063255 Chr.5: 158731430-158731430 on Build GRCh38
rs539657616 Chr.5: 158731421-158731421 on Build GRCh38
rs751908502 Chr.5: 158731386-158731386 on Build GRCh38
rs1004160351 Chr.5: 158731099-158731099 on Build GRCh38
rs1056090414 Chr.5: 158731349-158731349 on Build GRCh38
rs990719934 Chr.5: 158731265-158731265 on Build GRCh38
rs977620747 Chr.5: 158731293-158731293 on Build GRCh38
rs780062562 Chr.5: 158731049-158731049 on Build GRCh38
rs561604777 Chr.5: 158731023-158731023 on Build GRCh38
rs947830249 Chr.5: 158731354-158731354 on Build GRCh38
rs762837314 Chr.5: 158731168-158731168 on Build GRCh38
rs764996146 Chr.5: 158731165-158731165 on Build GRCh38
rs1034814061 Chr.5: 158731219-158731219 on Build GRCh38
rs772422154 Chr.5: 158731129-158731129 on Build GRCh38
rs765978409 Chr.5: 158731056-158731056 on Build GRCh38
rs753917018 Chr.5: 158731198-158731198 on Build GRCh38
rs568337073 Chr.5: 158731326-158731326 on Build GRCh38
rs777823795 Chr.5: 158731185-158731185 on Build GRCh38
rs531188238 Chr.5: 158731267-158731267 on Build GRCh38
rs752679304 Chr.5: 158731052-158731052 on Build GRCh38
rs993822307 Chr.5: 158731016-158731016 on Build GRCh38
rs1002876205 Chr.5: 158731157-158731157 on Build GRCh38
rs761138916 Chr.5: 158731140-158731140 on Build GRCh38
rs751490606 Chr.5: 158731175-158731175 on Build GRCh38
rs202004918 Chr.5: 158731055-158731055 on Build GRCh38
rs200547458 Chr.5: 158731079-158731079 on Build GRCh38
rs771206001 Chr.5: 158731100-158731100 on Build GRCh38
rs1024908567 Chr.5: 158731114-158731114 on Build GRCh38
rs568634470 Chr.5: 158731210-158731210 on Build GRCh38
rs199832129 Chr.5: 158731031-158731031 on Build GRCh38
rs776038203 Chr.5: 158731138-158731138 on Build GRCh38
rs1023139441 Chr.5: 158731033-158731033 on Build GRCh38
rs768270338 Chr.5: 158731147-158731147 on Build GRCh38
rs533890731 Chr.5: 158731340-158731340 on Build GRCh38
rs753294053 Chr.5: 158731248-158731248 on Build GRCh38
rs570510614 Chr.5: 158731401-158731401 on Build GRCh38
rs530360531 Chr.5: 158731030-158731030 on Build GRCh38
rs146276323 Chr.5: 158731150-158731150 on Build GRCh38
rs922103670 Chr.5: 158731341-158731341 on Build GRCh38
rs764573079 Chr.5: 158731355-158731355 on Build GRCh38
rs886910374 Chr.5: 158731438-158731438 on Build GRCh38
rs988852327 Chr.5: 158731215-158731215 on Build GRCh38
rs1000798711 Chr.5: 158731390-158731390 on Build GRCh38
rs374747069 Chr.5: 158731024-158731024 on Build GRCh38
rs575836054 Chr.5: 158731435-158731435 on Build GRCh38
rs985909797 Chr.5: 158731282-158731282 on Build GRCh38
rs773026096 Chr.5: 158731290-158731290 on Build GRCh38
rs766126972 Chr.5: 158731173-158731173 on Build GRCh38
rs191703921 Chr.5: 158731276-158731276 on Build GRCh38
rs911076577 Chr.5: 158731283-158731283 on Build GRCh38
rs746252760 Chr.5: 158731111-158731111 on Build GRCh38
rs1029662879 Chr.5: 158731275-158731275 on Build GRCh38
rs750136811 Chr.5: 158731166-158731166 on Build GRCh38
rs779178390 Chr.5: 158731101-158731101 on Build GRCh38
rs896120250 Chr.5: 158731350-158731350 on Build GRCh38
rs958852514 Chr.5: 158731223-158731223 on Build GRCh38
rs757031018 Chr.5: 158731029-158731029 on Build GRCh38
rs183534125 Chr.5: 158731331-158731331 on Build GRCh38

View Full Product Details