Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 160563798-160564322 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP10B
Entrez Gene ID
23120
Gene Name
ATPase phospholipid transporting 10B (putative)
Gene Aliases
ATPVB
UniGene
Hs.109358
Gene Chromosome Location
Chr.5: 160563120-160929411 on Build GRCh38
Transcripts
NM_025153.2 XM_006714833.2 XM_011534468.2 XM_011534469.1 XM_011534472.2 XM_017009252.1 XM_017009253.1

SNP Detail

SNP ID Chromosome Location
rs568112843 Chr.5: 160564063-160564063 on Build GRCh38
rs916333657 Chr.5: 160564215-160564215 on Build GRCh38
rs949539501 Chr.5: 160564022-160564022 on Build GRCh38
rs1040380871 Chr.5: 160564059-160564059 on Build GRCh38
rs1008061504 Chr.5: 160564183-160564183 on Build GRCh38
rs768848477 Chr.5: 160564229-160564229 on Build GRCh38
rs559180683 Chr.5: 160564235-160564235 on Build GRCh38
rs562586168 Chr.5: 160563970-160563970 on Build GRCh38
rs747274652 Chr.5: 160564226-160564226 on Build GRCh38
rs901947966 Chr.5: 160563825-160563825 on Build GRCh38
rs939824819 Chr.5: 160563951-160563951 on Build GRCh38
rs1042565519 Chr.5: 160564043-160564043 on Build GRCh38
rs942113797 Chr.5: 160563902-160563902 on Build GRCh38
rs918191427 Chr.5: 160564016-160564016 on Build GRCh38
rs1034709301 Chr.5: 160564163-160564163 on Build GRCh38
rs757286788 Chr.5: 160564054-160564054 on Build GRCh38
rs902595217 Chr.5: 160564051-160564051 on Build GRCh38
rs762296996 Chr.5: 160563877-160563877 on Build GRCh38
rs962251176 Chr.5: 160563913-160563913 on Build GRCh38
rs1015484613 Chr.5: 160564136-160564136 on Build GRCh38
rs777043518 Chr.5: 160564265-160564265 on Build GRCh38
rs186342339 Chr.5: 160564029-160564029 on Build GRCh38
rs919560440 Chr.5: 160564294-160564294 on Build GRCh38
rs867459366 Chr.5: 160564262-160564262 on Build GRCh38
rs986656381 Chr.5: 160563909-160563909 on Build GRCh38
rs919609096 Chr.5: 160563982-160563982 on Build GRCh38
rs1028226516 Chr.5: 160564152-160564152 on Build GRCh38
rs201810779 Chr.5: 160564107-160564107 on Build GRCh38
rs916432255 Chr.5: 160563984-160563984 on Build GRCh38
rs938232605 Chr.5: 160563985-160563985 on Build GRCh38
rs927587983 Chr.5: 160563947-160563947 on Build GRCh38
rs902444836 Chr.5: 160563991-160563991 on Build GRCh38
rs771997549 Chr.5: 160564133-160564133 on Build GRCh38
rs935219717 Chr.5: 160564038-160564038 on Build GRCh38
rs539387341 Chr.5: 160564169-160564169 on Build GRCh38
rs183299655 Chr.5: 160563861-160563861 on Build GRCh38
rs1029069963 Chr.5: 160564085-160564085 on Build GRCh38
rs147384461 Chr.5: 160564156-160564156 on Build GRCh38
rs1029598350 Chr.5: 160563880-160563880 on Build GRCh38
rs191198750 Chr.5: 160564261-160564261 on Build GRCh38
rs528336424 Chr.5: 160563894-160563894 on Build GRCh38
rs975336775 Chr.5: 160563838-160563838 on Build GRCh38
rs990855727 Chr.5: 160564207-160564207 on Build GRCh38
rs981680465 Chr.5: 160564143-160564143 on Build GRCh38
rs996256193 Chr.5: 160564076-160564076 on Build GRCh38
rs979301135 Chr.5: 160564280-160564280 on Build GRCh38
rs973862116 Chr.5: 160564245-160564245 on Build GRCh38
rs946433730 Chr.5: 160564232-160564232 on Build GRCh38
rs996743337 Chr.5: 160564129-160564130 on Build GRCh38
rs371014607 Chr.5: 160564174-160564174 on Build GRCh38
rs990580126 Chr.5: 160563977-160563977 on Build GRCh38
rs1043909764 Chr.5: 160563990-160563990 on Build GRCh38
rs1015430060 Chr.5: 160564206-160564206 on Build GRCh38
rs559700552 Chr.5: 160563922-160563922 on Build GRCh38
rs762095036 Chr.5: 160564295-160564295 on Build GRCh38
rs143164900 Chr.5: 160564118-160564118 on Build GRCh38
rs373945454 Chr.5: 160563885-160563885 on Build GRCh38
rs547376535 Chr.5: 160564138-160564138 on Build GRCh38
rs6895540 Chr.5: 160563987-160563987 on Build GRCh38

View Full Product Details