Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 161288026-161288524 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRB2
Entrez Gene ID
2561
Gene Name
gamma-aminobutyric acid type A receptor beta2 subunit
Gene Aliases
-
UniGene
Hs.303527
Gene Chromosome Location
Chr.5: 161288429-161548124 on Build GRCh38
Transcripts
NM_000813.2 NM_021911.2

SNP Detail

SNP ID Chromosome Location
rs1037237745 Chr.5: 161288068-161288068 on Build GRCh38
rs13189983 Chr.5: 161288433-161288433 on Build GRCh38
rs976585504 Chr.5: 161288197-161288197 on Build GRCh38
rs966818642 Chr.5: 161288164-161288164 on Build GRCh38
rs149204805 Chr.5: 161288386-161288386 on Build GRCh38
rs959586816 Chr.5: 161288088-161288088 on Build GRCh38
rs13156134 Chr.5: 161288434-161288434 on Build GRCh38
rs576610890 Chr.5: 161288052-161288052 on Build GRCh38
rs78792815 Chr.5: 161288269-161288269 on Build GRCh38
rs983187734 Chr.5: 161288289-161288289 on Build GRCh38
rs755515164 Chr.5: 161288133-161288133 on Build GRCh38
rs1001277672 Chr.5: 161288502-161288502 on Build GRCh38
rs559716560 Chr.5: 161288243-161288243 on Build GRCh38
rs963406474 Chr.5: 161288349-161288349 on Build GRCh38
rs377616810 Chr.5: 161288216-161288216 on Build GRCh38
rs868040449 Chr.5: 161288497-161288497 on Build GRCh38
rs17458157 Chr.5: 161288492-161288492 on Build GRCh38
rs924491140 Chr.5: 161288184-161288184 on Build GRCh38
rs548709490 Chr.5: 161288360-161288360 on Build GRCh38
rs772966351 Chr.5: 161288335-161288337 on Build GRCh38
rs909547155 Chr.5: 161288266-161288266 on Build GRCh38
rs777357506 Chr.5: 161288209-161288209 on Build GRCh38
rs544018980 Chr.5: 161288065-161288065 on Build GRCh38
rs577103216 Chr.5: 161288190-161288190 on Build GRCh38
rs252948 Chr.5: 161288084-161288084 on Build GRCh38
rs917111980 Chr.5: 161288461-161288461 on Build GRCh38
rs369136546 Chr.5: 161288278-161288278 on Build GRCh38
rs113205936 Chr.5: 161288134-161288134 on Build GRCh38
rs745616833 Chr.5: 161288489-161288489 on Build GRCh38
rs902543306 Chr.5: 161288194-161288194 on Build GRCh38
rs186745300 Chr.5: 161288067-161288067 on Build GRCh38
rs937121497 Chr.5: 161288371-161288371 on Build GRCh38
rs1055257745 Chr.5: 161288352-161288352 on Build GRCh38
rs191256505 Chr.5: 161288478-161288478 on Build GRCh38
rs1010912485 Chr.5: 161288121-161288121 on Build GRCh38
rs939814217 Chr.5: 161288268-161288268 on Build GRCh38
rs1011154802 Chr.5: 161288416-161288416 on Build GRCh38
rs1055173262 Chr.5: 161288115-161288115 on Build GRCh38
rs949437449 Chr.5: 161288487-161288487 on Build GRCh38
rs902509673 Chr.5: 161288500-161288500 on Build GRCh38
rs1034358248 Chr.5: 161288087-161288087 on Build GRCh38
rs907522229 Chr.5: 161288306-161288306 on Build GRCh38
rs138275079 Chr.5: 161288219-161288219 on Build GRCh38
rs906251860 Chr.5: 161288293-161288293 on Build GRCh38
rs921198836 Chr.5: 161288237-161288237 on Build GRCh38
rs983761557 Chr.5: 161288255-161288255 on Build GRCh38
rs761424061 Chr.5: 161288283-161288283 on Build GRCh38
rs939531562 Chr.5: 161288321-161288321 on Build GRCh38
rs575939144 Chr.5: 161288338-161288338 on Build GRCh38
rs978577647 Chr.5: 161288165-161288165 on Build GRCh38
rs888199719 Chr.5: 161288053-161288053 on Build GRCh38
rs906693014 Chr.5: 161288072-161288072 on Build GRCh38
rs13159923 Chr.5: 161288440-161288440 on Build GRCh38
rs752463240 Chr.5: 161288094-161288094 on Build GRCh38

View Full Product Details