Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 161291382-161291884 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRB2
Entrez Gene ID
2561
Gene Name
gamma-aminobutyric acid type A receptor beta2 subunit
Gene Aliases
-
UniGene
Hs.303527
Gene Chromosome Location
Chr.5: 161288429-161548124 on Build GRCh38
Transcripts
NM_000813.2 NM_021911.2

SNP Detail

SNP ID Chromosome Location
rs896066937 Chr.5: 161291766-161291766 on Build GRCh38
rs1014509353 Chr.5: 161291772-161291772 on Build GRCh38
rs1044532115 Chr.5: 161291698-161291698 on Build GRCh38
rs539702536 Chr.5: 161291501-161291501 on Build GRCh38
rs34190633 Chr.5: 161291832-161291832 on Build GRCh38
rs879723744 Chr.5: 161291667-161291667 on Build GRCh38
rs970166948 Chr.5: 161291559-161291559 on Build GRCh38
rs1047854987 Chr.5: 161291745-161291745 on Build GRCh38
rs910216108 Chr.5: 161291680-161291680 on Build GRCh38
rs1022992934 Chr.5: 161291786-161291786 on Build GRCh38
rs534042984 Chr.5: 161291625-161291625 on Build GRCh38
rs544448902 Chr.5: 161291668-161291668 on Build GRCh38
rs748387934 Chr.5: 161291666-161291666 on Build GRCh38
rs898886516 Chr.5: 161291692-161291692 on Build GRCh38
rs931129628 Chr.5: 161291623-161291623 on Build GRCh38
rs573895982 Chr.5: 161291662-161291662 on Build GRCh38
rs1003293595 Chr.5: 161291759-161291759 on Build GRCh38
rs1026997445 Chr.5: 161291460-161291460 on Build GRCh38
rs578214058 Chr.5: 161291701-161291701 on Build GRCh38
rs200095996 Chr.5: 161291751-161291751 on Build GRCh38
rs978820526 Chr.5: 161291574-161291574 on Build GRCh38
rs912223718 Chr.5: 161291520-161291520 on Build GRCh38
rs778024516 Chr.5: 161291836-161291836 on Build GRCh38
rs563086259 Chr.5: 161291676-161291676 on Build GRCh38
rs1014269015 Chr.5: 161291515-161291515 on Build GRCh38
rs956108973 Chr.5: 161291856-161291856 on Build GRCh38
rs905848564 Chr.5: 161291813-161291813 on Build GRCh38
rs76427125 Chr.5: 161291842-161291842 on Build GRCh38
rs913810383 Chr.5: 161291669-161291669 on Build GRCh38
rs896047881 Chr.5: 161291475-161291475 on Build GRCh38
rs1037401774 Chr.5: 161291683-161291683 on Build GRCh38
rs1011855478 Chr.5: 161291835-161291835 on Build GRCh38
rs985208128 Chr.5: 161291444-161291444 on Build GRCh38
rs557893455 Chr.5: 161291529-161291529 on Build GRCh38
rs115591351 Chr.5: 161291432-161291432 on Build GRCh38
rs1033528553 Chr.5: 161291848-161291848 on Build GRCh38
rs940995322 Chr.5: 161291753-161291753 on Build GRCh38
rs933217521 Chr.5: 161291673-161291673 on Build GRCh38
rs1021307252 Chr.5: 161291422-161291422 on Build GRCh38
rs555399112 Chr.5: 161291661-161291661 on Build GRCh38
rs1048273753 Chr.5: 161291660-161291660 on Build GRCh38
rs748703147 Chr.5: 161291856-161291857 on Build GRCh38
rs1022940608 Chr.5: 161291526-161291526 on Build GRCh38
rs1032203781 Chr.5: 161291593-161291593 on Build GRCh38
rs943146674 Chr.5: 161291682-161291682 on Build GRCh38
rs965458762 Chr.5: 161291550-161291550 on Build GRCh38
rs573162654 Chr.5: 161291543-161291543 on Build GRCh38
rs369482102 Chr.5: 161291795-161291795 on Build GRCh38
rs754787833 Chr.5: 161291433-161291433 on Build GRCh38

View Full Product Details