Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 161293336-161293845 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRB2
Entrez Gene ID
2561
Gene Name
gamma-aminobutyric acid type A receptor beta2 subunit
Gene Aliases
-
UniGene
Hs.303527
Gene Chromosome Location
Chr.5: 161288429-161548124 on Build GRCh38
Transcripts
NM_000813.2 NM_021911.2

SNP Detail

SNP ID Chromosome Location
rs113926219 Chr.5: 161293548-161293548 on Build GRCh38
rs559898394 Chr.5: 161293701-161293701 on Build GRCh38
rs964355416 Chr.5: 161293504-161293504 on Build GRCh38
rs957324543 Chr.5: 161293377-161293377 on Build GRCh38
rs976791935 Chr.5: 161293506-161293506 on Build GRCh38
rs143967869 Chr.5: 161293654-161293654 on Build GRCh38
rs555422740 Chr.5: 161293432-161293432 on Build GRCh38
rs372791346 Chr.5: 161293477-161293477 on Build GRCh38
rs772907429 Chr.5: 161293460-161293460 on Build GRCh38
rs931376634 Chr.5: 161293599-161293599 on Build GRCh38
rs942708761 Chr.5: 161293532-161293532 on Build GRCh38
rs543980542 Chr.5: 161293535-161293535 on Build GRCh38
rs975363654 Chr.5: 161293536-161293536 on Build GRCh38
rs907079848 Chr.5: 161293700-161293700 on Build GRCh38
rs1030100845 Chr.5: 161293531-161293531 on Build GRCh38
rs781099267 Chr.5: 161293478-161293478 on Build GRCh38
rs1034102617 Chr.5: 161293369-161293369 on Build GRCh38
rs912594141 Chr.5: 161293503-161293503 on Build GRCh38
rs910049673 Chr.5: 161293663-161293663 on Build GRCh38
rs59737038 Chr.5: 161293460-161293462 on Build GRCh38
rs940623393 Chr.5: 161293636-161293636 on Build GRCh38
rs1011721714 Chr.5: 161293728-161293728 on Build GRCh38
rs182141806 Chr.5: 161293538-161293538 on Build GRCh38
rs1020488921 Chr.5: 161293729-161293729 on Build GRCh38
rs967677323 Chr.5: 161293436-161293436 on Build GRCh38
rs750564358 Chr.5: 161293666-161293666 on Build GRCh38
rs990346076 Chr.5: 161293381-161293381 on Build GRCh38
rs573644009 Chr.5: 161293468-161293468 on Build GRCh38
rs987008904 Chr.5: 161293488-161293488 on Build GRCh38
rs907699433 Chr.5: 161293621-161293621 on Build GRCh38
rs527611001 Chr.5: 161293712-161293712 on Build GRCh38
rs967939218 Chr.5: 161293738-161293738 on Build GRCh38
rs796440688 Chr.5: 161293463-161293465 on Build GRCh38
rs1055839559 Chr.5: 161293702-161293702 on Build GRCh38
rs78568422 Chr.5: 161293603-161293603 on Build GRCh38
rs1045686036 Chr.5: 161293678-161293678 on Build GRCh38

View Full Product Details