Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 162101107-162101625 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRG2
Entrez Gene ID
2566
Gene Name
gamma-aminobutyric acid type A receptor gamma2 subunit
Gene Aliases
CAE2, ECA2, GEFSP3
UniGene
Hs.7195
Gene Chromosome Location
Chr.5: 162067642-162155539 on Build GRCh38
Transcripts
NM_000816.3 NM_198903.2 NM_198904.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3247952 c.618G>C p.L206F Chr.5: 162101304-162101304 on Build GRCh38
1065342 c.629G>A p.S210N Chr.5: 162101315-162101315 on Build GRCh38
6987965 c.549-1G>T p.? Chr.5: 162101234-162101234 on Build GRCh38
3247950 c.565G>C p.E189Q Chr.5: 162101251-162101251 on Build GRCh38
1065341 c.600T>C p.D200D Chr.5: 162101286-162101286 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771888226 Chr.5: 162101298-162101298 on Build GRCh38
rs764971510 Chr.5: 162101204-162101204 on Build GRCh38
rs906971953 Chr.5: 162101316-162101316 on Build GRCh38
rs970912162 Chr.5: 162101536-162101536 on Build GRCh38
rs981863613 Chr.5: 162101269-162101269 on Build GRCh38
rs909136816 Chr.5: 162101565-162101565 on Build GRCh38
rs62384032 Chr.5: 162101581-162101581 on Build GRCh38
rs758280295 Chr.5: 162101214-162101227 on Build GRCh38
rs34875047 Chr.5: 162101581-162101581 on Build GRCh38
rs211037 Chr.5: 162101274-162101274 on Build GRCh38
rs750459631 Chr.5: 162101232-162101232 on Build GRCh38
rs760578797 Chr.5: 162101333-162101333 on Build GRCh38
rs986318925 Chr.5: 162101426-162101426 on Build GRCh38
rs548306755 Chr.5: 162101358-162101358 on Build GRCh38
rs764144533 Chr.5: 162101215-162101215 on Build GRCh38
rs17855004 Chr.5: 162101313-162101313 on Build GRCh38
rs929252404 Chr.5: 162101188-162101188 on Build GRCh38
rs764235541 Chr.5: 162101504-162101504 on Build GRCh38
rs968588789 Chr.5: 162101393-162101393 on Build GRCh38
rs752486094 Chr.5: 162101211-162101212 on Build GRCh38
rs758574181 Chr.5: 162101238-162101238 on Build GRCh38
rs775399703 Chr.5: 162101329-162101329 on Build GRCh38
rs751723547 Chr.5: 162101265-162101265 on Build GRCh38
rs869285825 Chr.5: 162101591-162101592 on Build GRCh38
rs926261472 Chr.5: 162101584-162101584 on Build GRCh38
rs534873454 Chr.5: 162101527-162101527 on Build GRCh38
rs1041029648 Chr.5: 162101431-162101431 on Build GRCh38
rs188278790 Chr.5: 162101483-162101483 on Build GRCh38
rs370315018 Chr.5: 162101359-162101359 on Build GRCh38
rs770506471 Chr.5: 162101282-162101282 on Build GRCh38
rs966112801 Chr.5: 162101471-162101471 on Build GRCh38
rs756432734 Chr.5: 162101369-162101369 on Build GRCh38
rs868460983 Chr.5: 162101253-162101253 on Build GRCh38
rs756375646 Chr.5: 162101409-162101409 on Build GRCh38
rs764024986 Chr.5: 162101326-162101327 on Build GRCh38
rs755215653 Chr.5: 162101266-162101266 on Build GRCh38
rs1027590415 Chr.5: 162101473-162101473 on Build GRCh38
rs981748988 Chr.5: 162101545-162101545 on Build GRCh38
rs937739985 Chr.5: 162101299-162101299 on Build GRCh38
rs996916558 Chr.5: 162101451-162101451 on Build GRCh38
rs71587179 Chr.5: 162101592-162101592 on Build GRCh38
rs749764060 Chr.5: 162101281-162101281 on Build GRCh38
rs145582456 Chr.5: 162101335-162101335 on Build GRCh38
rs761672853 Chr.5: 162101339-162101339 on Build GRCh38
rs529462959 Chr.5: 162101349-162101349 on Build GRCh38
rs781498456 Chr.5: 162101268-162101268 on Build GRCh38
rs778408584 Chr.5: 162101280-162101280 on Build GRCh38
rs774016949 Chr.5: 162101292-162101292 on Build GRCh38
rs1004905517 Chr.5: 162101491-162101491 on Build GRCh38
rs1015794365 Chr.5: 162101510-162101510 on Build GRCh38
rs759221112 Chr.5: 162101295-162101295 on Build GRCh38
rs766398270 Chr.5: 162101341-162101341 on Build GRCh38
rs537071801 Chr.5: 162101404-162101404 on Build GRCh38
rs150134470 Chr.5: 162101271-162101271 on Build GRCh38
rs983036380 Chr.5: 162101168-162101168 on Build GRCh38
rs55712126 Chr.5: 162101372-162101372 on Build GRCh38
rs558559594 Chr.5: 162101440-162101440 on Build GRCh38
rs368429259 Chr.5: 162101338-162101338 on Build GRCh38
rs908244332 Chr.5: 162101183-162101183 on Build GRCh38
rs564464466 Chr.5: 162101256-162101256 on Build GRCh38
rs752566687 Chr.5: 162101204-162101204 on Build GRCh38
rs367737806 Chr.5: 162101184-162101184 on Build GRCh38
rs79110808 Chr.5: 162101579-162101579 on Build GRCh38
rs751670193 Chr.5: 162101347-162101347 on Build GRCh38
rs760493990 Chr.5: 162101205-162101205 on Build GRCh38

View Full Product Details