Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 163443904-163444408 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNG1
Entrez Gene ID
900
Gene Name
cyclin G1
Gene Aliases
CCNG
UniGene
Hs.79101
Gene Chromosome Location
Chr.5: 163437567-163445016 on Build GRCh38
Transcripts
NM_004060.3 NM_199246.1

SNP Detail

SNP ID Chromosome Location
rs1055356076 Chr.5: 163443986-163443986 on Build GRCh38
rs376581128 Chr.5: 163444299-163444299 on Build GRCh38
rs754246739 Chr.5: 163444361-163444361 on Build GRCh38
rs754546531 Chr.5: 163444058-163444058 on Build GRCh38
rs759039482 Chr.5: 163443928-163443928 on Build GRCh38
rs574467199 Chr.5: 163444308-163444312 on Build GRCh38
rs1021533521 Chr.5: 163444157-163444157 on Build GRCh38
rs767105467 Chr.5: 163443969-163443969 on Build GRCh38
rs908093092 Chr.5: 163444374-163444374 on Build GRCh38
rs931164033 Chr.5: 163443982-163443982 on Build GRCh38
rs1037754610 Chr.5: 163443943-163443943 on Build GRCh38
rs1029570505 Chr.5: 163444316-163444316 on Build GRCh38
rs955015812 Chr.5: 163444346-163444346 on Build GRCh38
rs1002125349 Chr.5: 163443952-163443952 on Build GRCh38
rs371481198 Chr.5: 163444296-163444296 on Build GRCh38
rs940659906 Chr.5: 163443933-163443933 on Build GRCh38
rs17883523 Chr.5: 163444317-163444317 on Build GRCh38
rs1032106676 Chr.5: 163444246-163444251 on Build GRCh38
rs943617302 Chr.5: 163444226-163444226 on Build GRCh38
rs957761278 Chr.5: 163444289-163444289 on Build GRCh38
rs565847062 Chr.5: 163444150-163444150 on Build GRCh38
rs756560845 Chr.5: 163444287-163444287 on Build GRCh38
rs1013918775 Chr.5: 163444089-163444090 on Build GRCh38
rs898419201 Chr.5: 163443950-163443950 on Build GRCh38
rs766423181 Chr.5: 163444050-163444050 on Build GRCh38
rs2291790 Chr.5: 163444179-163444179 on Build GRCh38
rs992278729 Chr.5: 163444027-163444027 on Build GRCh38
rs980013359 Chr.5: 163444192-163444192 on Build GRCh38
rs988134922 Chr.5: 163444360-163444360 on Build GRCh38
rs149348306 Chr.5: 163444204-163444204 on Build GRCh38
rs1050896730 Chr.5: 163444332-163444332 on Build GRCh38
rs960754561 Chr.5: 163443974-163443974 on Build GRCh38
rs2069369 Chr.5: 163443979-163443979 on Build GRCh38
rs895087859 Chr.5: 163444062-163444062 on Build GRCh38

View Full Product Details