Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 177514784-177515286 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX41
Entrez Gene ID
51428
Gene Name
DEAD-box helicase 41
Gene Aliases
ABS, MPLPF
UniGene
Hs.484288
Gene Chromosome Location
Chr.5: 177511577-177517326 on Build GRCh38
Transcripts
NM_001321732.1 NM_001321830.1 NM_016222.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1066414 c.844C>T p.R282C Chr.5: 177514792-177514792 on Build GRCh38
6495027 c.770G>A p.G257E Chr.5: 177514944-177514944 on Build GRCh38
7208614 c.601A>T p.I201F Chr.5: 177515229-177515229 on Build GRCh38
7203436 c.590A>G p.K197R Chr.5: 177515240-177515240 on Build GRCh38
6495026 c.804delG p.E268fs*36 Chr.5: 177514832-177514832 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs551630412 Chr.5: 177514981-177514981 on Build GRCh38
rs751293126 Chr.5: 177514978-177514978 on Build GRCh38
rs755198680 Chr.5: 177515088-177515088 on Build GRCh38
rs778216275 Chr.5: 177514992-177514992 on Build GRCh38
rs767782519 Chr.5: 177514879-177514879 on Build GRCh38
rs371633755 Chr.5: 177515093-177515093 on Build GRCh38
rs773904073 Chr.5: 177515119-177515119 on Build GRCh38
rs571753440 Chr.5: 177515000-177515000 on Build GRCh38
rs771510721 Chr.5: 177515247-177515247 on Build GRCh38
rs761188003 Chr.5: 177514903-177514903 on Build GRCh38
rs747935808 Chr.5: 177515018-177515018 on Build GRCh38
rs1013249905 Chr.5: 177515087-177515087 on Build GRCh38
rs775258340 Chr.5: 177514881-177514881 on Build GRCh38
rs773536438 Chr.5: 177515263-177515263 on Build GRCh38
rs773819724 Chr.5: 177515041-177515041 on Build GRCh38
rs376574336 Chr.5: 177515115-177515115 on Build GRCh38
rs143274979 Chr.5: 177514973-177514973 on Build GRCh38
rs139780256 Chr.5: 177514938-177514938 on Build GRCh38
rs748590587 Chr.5: 177514837-177514837 on Build GRCh38
rs376093707 Chr.5: 177515002-177515002 on Build GRCh38
rs375274366 Chr.5: 177515254-177515254 on Build GRCh38
rs760334582 Chr.5: 177515261-177515261 on Build GRCh38
rs146175373 Chr.5: 177514974-177514974 on Build GRCh38
rs759325258 Chr.5: 177515236-177515236 on Build GRCh38
rs769733379 Chr.5: 177515107-177515116 on Build GRCh38
rs200122668 Chr.5: 177514815-177514815 on Build GRCh38
rs747842313 Chr.5: 177514963-177514963 on Build GRCh38
rs778379292 Chr.5: 177514825-177514825 on Build GRCh38
rs745800400 Chr.5: 177515066-177515067 on Build GRCh38
rs749282766 Chr.5: 177514849-177514849 on Build GRCh38
rs149122003 Chr.5: 177515239-177515239 on Build GRCh38
rs776481635 Chr.5: 177515015-177515015 on Build GRCh38
rs768526031 Chr.5: 177514953-177514953 on Build GRCh38
rs759005362 Chr.5: 177515135-177515135 on Build GRCh38
rs781318967 Chr.5: 177515089-177515089 on Build GRCh38
rs144003034 Chr.5: 177515024-177515024 on Build GRCh38
rs958202796 Chr.5: 177514868-177514868 on Build GRCh38
rs762515434 Chr.5: 177515026-177515026 on Build GRCh38
rs73804366 Chr.5: 177514873-177514873 on Build GRCh38
rs753835830 Chr.5: 177514899-177514899 on Build GRCh38
rs754196793 Chr.5: 177515058-177515058 on Build GRCh38
rs756840368 Chr.5: 177514813-177514813 on Build GRCh38
rs755920162 Chr.5: 177515231-177515231 on Build GRCh38
rs376867329 Chr.5: 177515065-177515065 on Build GRCh38
rs778514273 Chr.5: 177514961-177514961 on Build GRCh38
rs749438678 Chr.5: 177515177-177515178 on Build GRCh38
rs371392769 Chr.5: 177514895-177514895 on Build GRCh38
rs780209663 Chr.5: 177515003-177515003 on Build GRCh38
rs541153895 Chr.5: 177515197-177515197 on Build GRCh38
rs756369893 Chr.5: 177514984-177514984 on Build GRCh38
rs910481563 Chr.5: 177515027-177515027 on Build GRCh38
rs758406210 Chr.5: 177515066-177515066 on Build GRCh38
rs745544108 Chr.5: 177514827-177514827 on Build GRCh38
rs747256985 Chr.5: 177515007-177515007 on Build GRCh38
rs765680048 Chr.5: 177515062-177515062 on Build GRCh38
rs145085939 Chr.5: 177515233-177515233 on Build GRCh38
rs769832683 Chr.5: 177515019-177515019 on Build GRCh38
rs778947112 Chr.5: 177514942-177514942 on Build GRCh38
rs371460149 Chr.5: 177515061-177515061 on Build GRCh38
rs749253420 Chr.5: 177515111-177515111 on Build GRCh38
rs150888965 Chr.5: 177514835-177514835 on Build GRCh38
rs200056121 Chr.5: 177515162-177515162 on Build GRCh38
rs780120166 Chr.5: 177515079-177515079 on Build GRCh38
rs905341773 Chr.5: 177515222-177515222 on Build GRCh38
rs775024380 Chr.5: 177515166-177515166 on Build GRCh38
rs773822170 Chr.5: 177514845-177514845 on Build GRCh38
rs750013130 Chr.5: 177514916-177514916 on Build GRCh38
rs200707924 Chr.5: 177514966-177514966 on Build GRCh38
rs181004515 Chr.5: 177514928-177514928 on Build GRCh38
rs962423100 Chr.5: 177515191-177515191 on Build GRCh38
rs920927202 Chr.5: 177515244-177515244 on Build GRCh38
rs771077918 Chr.5: 177514851-177514851 on Build GRCh38
rs1004241531 Chr.5: 177515253-177515253 on Build GRCh38
rs890612475 Chr.5: 177515004-177515004 on Build GRCh38
rs139112542 Chr.5: 177514996-177514996 on Build GRCh38
rs747126487 Chr.5: 177515080-177515080 on Build GRCh38
rs775458261 Chr.5: 177515113-177515115 on Build GRCh38
rs114125242 Chr.5: 177515090-177515090 on Build GRCh38
rs773221580 Chr.5: 177515098-177515098 on Build GRCh38
rs756526975 Chr.5: 177514931-177514931 on Build GRCh38
rs764806939 Chr.5: 177514909-177514909 on Build GRCh38
rs745329544 Chr.5: 177515246-177515246 on Build GRCh38
rs949946325 Chr.5: 177514845-177514846 on Build GRCh38
rs768353496 Chr.5: 177515262-177515262 on Build GRCh38
rs367840907 Chr.5: 177515042-177515042 on Build GRCh38
rs762124039 Chr.5: 177515176-177515176 on Build GRCh38
rs1000039111 Chr.5: 177514836-177514836 on Build GRCh38
rs765156652 Chr.5: 177514967-177514967 on Build GRCh38
rs781436670 Chr.5: 177515203-177515203 on Build GRCh38
rs568464531 Chr.5: 177514807-177514807 on Build GRCh38
rs772246357 Chr.5: 177514949-177514949 on Build GRCh38
rs778763328 Chr.5: 177515245-177515245 on Build GRCh38
rs775735135 Chr.5: 177514950-177514950 on Build GRCh38
rs935314360 Chr.5: 177515118-177515118 on Build GRCh38
rs535988198 Chr.5: 177514832-177514832 on Build GRCh38
rs765996540 Chr.5: 177515033-177515033 on Build GRCh38
rs1009033631 Chr.5: 177515005-177515005 on Build GRCh38
rs774371727 Chr.5: 177514870-177514870 on Build GRCh38
rs369566052 Chr.5: 177515186-177515186 on Build GRCh38
rs760338178 Chr.5: 177514951-177514951 on Build GRCh38
rs768277082 Chr.5: 177515009-177515009 on Build GRCh38
rs770169990 Chr.5: 177514841-177514841 on Build GRCh38
rs755036127 Chr.5: 177515200-177515200 on Build GRCh38
rs34567318 Chr.5: 177515203-177515204 on Build GRCh38
rs780979459 Chr.5: 177514832-177514832 on Build GRCh38
rs982554644 Chr.5: 177515137-177515137 on Build GRCh38
rs760498538 Chr.5: 177514894-177514894 on Build GRCh38
rs373575560 Chr.5: 177515214-177515214 on Build GRCh38
rs758121960 Chr.5: 177514917-177514917 on Build GRCh38
rs746011550 Chr.5: 177514948-177514948 on Build GRCh38
rs970017153 Chr.5: 177514919-177514919 on Build GRCh38
rs901158764 Chr.5: 177514834-177514834 on Build GRCh38
rs749923024 Chr.5: 177514968-177514968 on Build GRCh38
rs770860996 Chr.5: 177514940-177514940 on Build GRCh38
rs139845884 Chr.5: 177514937-177514937 on Build GRCh38
rs920310571 Chr.5: 177515006-177515006 on Build GRCh38
rs766913824 Chr.5: 177515192-177515192 on Build GRCh38
rs573545872 Chr.5: 177515187-177515187 on Build GRCh38

View Full Product Details