Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 179698776-179699287 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CANX
Entrez Gene ID
821
Gene Name
calnexin
Gene Aliases
CNX, IP90, P90
UniGene
Hs.567968
Gene Chromosome Location
Chr.5: 179678631-179731641 on Build GRCh38
Transcripts
NM_001024649.1 NM_001746.3 XM_011534664.1 XM_011534665.2

SNP Detail

SNP ID Chromosome Location
rs932043393 Chr.5: 179698815-179698815 on Build GRCh38
rs995800616 Chr.5: 179698911-179698911 on Build GRCh38
rs548613789 Chr.5: 179698868-179698868 on Build GRCh38
rs1007634689 Chr.5: 179699241-179699241 on Build GRCh38
rs944885440 Chr.5: 179699163-179699163 on Build GRCh38
rs1036523978 Chr.5: 179699237-179699237 on Build GRCh38
rs1023992996 Chr.5: 179699075-179699075 on Build GRCh38
rs896214150 Chr.5: 179699243-179699243 on Build GRCh38
rs924774752 Chr.5: 179698968-179698968 on Build GRCh38
rs966247751 Chr.5: 179698964-179698964 on Build GRCh38
rs1003486609 Chr.5: 179698936-179698936 on Build GRCh38
rs887437015 Chr.5: 179699228-179699228 on Build GRCh38
rs944967258 Chr.5: 179698847-179698847 on Build GRCh38
rs980965558 Chr.5: 179699172-179699172 on Build GRCh38
rs576989362 Chr.5: 179698991-179698991 on Build GRCh38
rs1019486394 Chr.5: 179699242-179699242 on Build GRCh38
rs936211802 Chr.5: 179698977-179698977 on Build GRCh38
rs185536392 Chr.5: 179699069-179699069 on Build GRCh38
rs115239999 Chr.5: 179698850-179698850 on Build GRCh38
rs977640593 Chr.5: 179699130-179699130 on Build GRCh38
rs924805726 Chr.5: 179698865-179698865 on Build GRCh38
rs761392597 Chr.5: 179699056-179699056 on Build GRCh38
rs756252202 Chr.5: 179699070-179699070 on Build GRCh38
rs1134924 Chr.5: 179699089-179699089 on Build GRCh38
rs548756803 Chr.5: 179699227-179699227 on Build GRCh38
rs1053203215 Chr.5: 179698860-179698860 on Build GRCh38
rs1011873470 Chr.5: 179698914-179698914 on Build GRCh38
rs917238243 Chr.5: 179698823-179698823 on Build GRCh38
rs977671563 Chr.5: 179698967-179698967 on Build GRCh38
rs536615868 Chr.5: 179698866-179698866 on Build GRCh38
rs376470095 Chr.5: 179699178-179699178 on Build GRCh38
rs911899931 Chr.5: 179698962-179698962 on Build GRCh38
rs1003455368 Chr.5: 179699033-179699033 on Build GRCh38
rs961908722 Chr.5: 179698938-179698938 on Build GRCh38
rs953594761 Chr.5: 179698955-179698955 on Build GRCh38
rs1036322199 Chr.5: 179699045-179699045 on Build GRCh38
rs912113734 Chr.5: 179698841-179698841 on Build GRCh38
rs566715079 Chr.5: 179698874-179698874 on Build GRCh38
rs897716781 Chr.5: 179699199-179699199 on Build GRCh38
rs1054552596 Chr.5: 179698982-179698982 on Build GRCh38
rs926002026 Chr.5: 179698843-179698843 on Build GRCh38
rs938963271 Chr.5: 179699191-179699191 on Build GRCh38
rs193279323 Chr.5: 179698833-179698833 on Build GRCh38
rs1027701158 Chr.5: 179698941-179698941 on Build GRCh38
rs4701196 Chr.5: 179698934-179698934 on Build GRCh38
rs926053012 Chr.5: 179698951-179698951 on Build GRCh38
rs969249110 Chr.5: 179699077-179699077 on Build GRCh38
rs998776360 Chr.5: 179699255-179699255 on Build GRCh38
rs986121202 Chr.5: 179698961-179698961 on Build GRCh38
rs897750866 Chr.5: 179699047-179699047 on Build GRCh38
rs532636785 Chr.5: 179698805-179698805 on Build GRCh38
rs1014797163 Chr.5: 179699026-179699026 on Build GRCh38
rs540604433 Chr.5: 179699230-179699230 on Build GRCh38
rs573473085 Chr.5: 179699207-179699207 on Build GRCh38
rs1036037591 Chr.5: 179699193-179699193 on Build GRCh38
rs900729964 Chr.5: 179698965-179698965 on Build GRCh38
rs1044711138 Chr.5: 179699016-179699016 on Build GRCh38
rs914959743 Chr.5: 179699005-179699005 on Build GRCh38
rs373052316 Chr.5: 179699149-179699149 on Build GRCh38
rs957486043 Chr.5: 179699148-179699148 on Build GRCh38
rs989031669 Chr.5: 179698959-179698959 on Build GRCh38
rs112694584 Chr.5: 179699247-179699247 on Build GRCh38
rs558700034 Chr.5: 179698975-179698975 on Build GRCh38
rs893457853 Chr.5: 179698903-179698903 on Build GRCh38
rs953466193 Chr.5: 179699097-179699097 on Build GRCh38
rs372071727 Chr.5: 179698856-179698856 on Build GRCh38
rs980049257 Chr.5: 179698946-179698946 on Build GRCh38
rs1027422765 Chr.5: 179698917-179698917 on Build GRCh38
rs1014754304 Chr.5: 179698937-179698937 on Build GRCh38
rs1004633075 Chr.5: 179699015-179699015 on Build GRCh38
rs1045135733 Chr.5: 179698926-179698926 on Build GRCh38
rs1004749235 Chr.5: 179698931-179698931 on Build GRCh38
rs73338133 Chr.5: 179699264-179699264 on Build GRCh38
rs1008082207 Chr.5: 179699098-179699098 on Build GRCh38
rs976246932 Chr.5: 179699169-179699169 on Build GRCh38
rs994923037 Chr.5: 179698940-179698940 on Build GRCh38
rs973514562 Chr.5: 179698802-179698802 on Build GRCh38
rs947818625 Chr.5: 179699154-179699154 on Build GRCh38
rs924824090 Chr.5: 179699144-179699144 on Build GRCh38
rs755273025 Chr.5: 179699150-179699150 on Build GRCh38
rs906221839 Chr.5: 179699024-179699024 on Build GRCh38
rs537670398 Chr.5: 179699009-179699009 on Build GRCh38
rs966049916 Chr.5: 179699114-179699114 on Build GRCh38
rs922206013 Chr.5: 179699203-179699203 on Build GRCh38
rs900172919 Chr.5: 179698878-179698878 on Build GRCh38
rs990303620 Chr.5: 179699148-179699148 on Build GRCh38
rs113745096 Chr.5: 179699231-179699231 on Build GRCh38
rs957623322 Chr.5: 179699145-179699145 on Build GRCh38
rs1018933393 Chr.5: 179699105-179699105 on Build GRCh38
rs773181581 Chr.5: 179698830-179698830 on Build GRCh38
rs753364875 Chr.5: 179699013-179699013 on Build GRCh38
rs541377664 Chr.5: 179699095-179699095 on Build GRCh38
rs1044468915 Chr.5: 179698842-179698842 on Build GRCh38
rs951744844 Chr.5: 179698925-179698925 on Build GRCh38

View Full Product Details