Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 150283464-150283985 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CIART
Entrez Gene ID
148523
Gene Name
circadian associated repressor of transcription
Gene Aliases
C1orf51, CHRONO, GM129
UniGene
Hs.54680
Gene Chromosome Location
Chr.1: 150282533-150287093 on Build GRCh38
Transcripts
NM_001300838.1 NM_001300839.1 NM_001300840.1 NM_001300841.1 NM_144697.3 XM_017000378.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5965452 c.403G>A p.D135N Chr.1: 150283841-150283841 on Build GRCh38
7291166 c.247C>T p.R83W Chr.1: 150283514-150283514 on Build GRCh38
4667400 c.229C>T p.P77S Chr.1: 150283496-150283496 on Build GRCh38
1160857 c.204G>A p.S68S Chr.1: 150283471-150283471 on Build GRCh38
4667399 c.200G>A p.R67Q Chr.1: 150283467-150283467 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782396736 Chr.1: 150283663-150283663 on Build GRCh38
rs145863368 Chr.1: 150283521-150283521 on Build GRCh38
rs781802341 Chr.1: 150283525-150283525 on Build GRCh38
rs782663456 Chr.1: 150283897-150283897 on Build GRCh38
rs782108251 Chr.1: 150283777-150283777 on Build GRCh38
rs969778711 Chr.1: 150283790-150283790 on Build GRCh38
rs782548414 Chr.1: 150283845-150283845 on Build GRCh38
rs145324799 Chr.1: 150283626-150283626 on Build GRCh38
rs889189366 Chr.1: 150283690-150283690 on Build GRCh38
rs782699353 Chr.1: 150283793-150283793 on Build GRCh38
rs199853192 Chr.1: 150283683-150283683 on Build GRCh38
rs782432969 Chr.1: 150283576-150283576 on Build GRCh38
rs113957787 Chr.1: 150283691-150283691 on Build GRCh38
rs782540243 Chr.1: 150283532-150283532 on Build GRCh38
rs981378388 Chr.1: 150283791-150283791 on Build GRCh38
rs946678261 Chr.1: 150283673-150283673 on Build GRCh38
rs587726852 Chr.1: 150283890-150283890 on Build GRCh38
rs781963912 Chr.1: 150283486-150283486 on Build GRCh38
rs781811485 Chr.1: 150283814-150283814 on Build GRCh38
rs782061165 Chr.1: 150283652-150283652 on Build GRCh38
rs781829032 Chr.1: 150283757-150283757 on Build GRCh38
rs782384319 Chr.1: 150283928-150283928 on Build GRCh38
rs200664734 Chr.1: 150283540-150283540 on Build GRCh38
rs587723252 Chr.1: 150283729-150283729 on Build GRCh38
rs969579527 Chr.1: 150283536-150283536 on Build GRCh38
rs939607694 Chr.1: 150283581-150283581 on Build GRCh38
rs782376273 Chr.1: 150283597-150283597 on Build GRCh38
rs782597616 Chr.1: 150283578-150283578 on Build GRCh38
rs782114444 Chr.1: 150283496-150283496 on Build GRCh38
rs782358787 Chr.1: 150283624-150283624 on Build GRCh38
rs781841039 Chr.1: 150283573-150283573 on Build GRCh38
rs782312659 Chr.1: 150283765-150283765 on Build GRCh38
rs993441445 Chr.1: 150283743-150283743 on Build GRCh38
rs1022760711 Chr.1: 150283773-150283773 on Build GRCh38
rs587593879 Chr.1: 150283745-150283745 on Build GRCh38
rs2298164 Chr.1: 150283594-150283594 on Build GRCh38
rs781867035 Chr.1: 150283866-150283866 on Build GRCh38
rs913923803 Chr.1: 150283665-150283665 on Build GRCh38
rs782528971 Chr.1: 150283553-150283553 on Build GRCh38
rs372326987 Chr.1: 150283746-150283746 on Build GRCh38
rs782463177 Chr.1: 150283873-150283873 on Build GRCh38
rs782028788 Chr.1: 150283667-150283667 on Build GRCh38
rs988410776 Chr.1: 150283939-150283939 on Build GRCh38
rs143005306 Chr.1: 150283557-150283557 on Build GRCh38
rs144282615 Chr.1: 150283610-150283610 on Build GRCh38
rs376071716 Chr.1: 150283664-150283664 on Build GRCh38
rs377608254 Chr.1: 150283554-150283554 on Build GRCh38
rs201427036 Chr.1: 150283886-150283886 on Build GRCh38
rs1053088522 Chr.1: 150283621-150283621 on Build GRCh38
rs755990671 Chr.1: 150283877-150283877 on Build GRCh38
rs782214203 Chr.1: 150283613-150283613 on Build GRCh38
rs928230974 Chr.1: 150283563-150283563 on Build GRCh38
rs782797983 Chr.1: 150283684-150283684 on Build GRCh38
rs961138582 Chr.1: 150283926-150283926 on Build GRCh38
rs782285950 Chr.1: 150283920-150283920 on Build GRCh38
rs142518963 Chr.1: 150283515-150283515 on Build GRCh38
rs901785875 Chr.1: 150283741-150283741 on Build GRCh38
rs782677729 Chr.1: 150283851-150283851 on Build GRCh38
rs587673232 Chr.1: 150283749-150283749 on Build GRCh38

View Full Product Details