Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 150554260-150554754 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTSL4
Entrez Gene ID
54507
Gene Name
ADAMTS like 4
Gene Aliases
ADAMTSL-4, ECTOL2, TSRC1
UniGene
Hs.516243
Gene Chromosome Location
Chr.1: 150549369-150560937 on Build GRCh38
Transcripts
NM_001288607.1 NM_001288608.1 NM_019032.5 NM_025008.4 XM_011509644.2 XM_011509645.2 XM_011509648.2 XM_011509649.2 XM_011509650.2 XM_011509651.2 XM_011509652.1 XM_017001506.1

Gene Symbol
LOC100289061
Entrez Gene ID
100289061
Gene Name
uncharacterized LOC100289061
Gene Aliases
-
UniGene
Hs.677561
Gene Chromosome Location
Chr.1: 150548564-150556700 on Build GRCh38
Transcripts
NM_001288607.1 NM_001288608.1 NM_019032.5 NM_025008.4 XM_011509644.2 XM_011509645.2 XM_011509648.2 XM_011509649.2 XM_011509650.2 XM_011509651.2 XM_011509652.1 XM_017001506.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4435984 c.1209delG p.Y404fs*40 Chr.1: 150554442-150554442 on Build GRCh38
6051388 c.1149G>T p.Q383H Chr.1: 150554382-150554382 on Build GRCh38
6051389 c.1149G>T p.Q383H Chr.1: 150554382-150554382 on Build GRCh38
4460163 c.1155C>T p.D385D Chr.1: 150554388-150554388 on Build GRCh38
6594511 c.1222C>T p.P408S Chr.1: 150554455-150554455 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752257967 Chr.1: 150554334-150554334 on Build GRCh38
rs773709711 Chr.1: 150554479-150554479 on Build GRCh38
rs923985258 Chr.1: 150554555-150554555 on Build GRCh38
rs760448489 Chr.1: 150554511-150554511 on Build GRCh38
rs1031222221 Chr.1: 150554590-150554590 on Build GRCh38
rs970577109 Chr.1: 150554287-150554287 on Build GRCh38
rs145639662 Chr.1: 150554375-150554375 on Build GRCh38
rs748247749 Chr.1: 150554467-150554467 on Build GRCh38
rs371019983 Chr.1: 150554671-150554673 on Build GRCh38
rs1048298617 Chr.1: 150554699-150554699 on Build GRCh38
rs773514905 Chr.1: 150554384-150554384 on Build GRCh38
rs953041467 Chr.1: 150554675-150554675 on Build GRCh38
rs587758769 Chr.1: 150554509-150554509 on Build GRCh38
rs111260629 Chr.1: 150554648-150554648 on Build GRCh38
rs587766449 Chr.1: 150554352-150554352 on Build GRCh38
rs41317523 Chr.1: 150554311-150554311 on Build GRCh38
rs989202763 Chr.1: 150554338-150554338 on Build GRCh38
rs587732229 Chr.1: 150554285-150554285 on Build GRCh38
rs372010340 Chr.1: 150554350-150554350 on Build GRCh38
rs749389911 Chr.1: 150554634-150554634 on Build GRCh38
rs755914142 Chr.1: 150554531-150554531 on Build GRCh38
rs371339040 Chr.1: 150554406-150554406 on Build GRCh38
rs948194489 Chr.1: 150554603-150554603 on Build GRCh38
rs888433095 Chr.1: 150554494-150554494 on Build GRCh38
rs752928902 Chr.1: 150554315-150554315 on Build GRCh38
rs753373488 Chr.1: 150554408-150554408 on Build GRCh38
rs762772611 Chr.1: 150554557-150554557 on Build GRCh38
rs755252678 Chr.1: 150554577-150554577 on Build GRCh38
rs774550931 Chr.1: 150554442-150554443 on Build GRCh38
rs182053027 Chr.1: 150554412-150554412 on Build GRCh38
rs755627505 Chr.1: 150554343-150554343 on Build GRCh38
rs199960526 Chr.1: 150554359-150554359 on Build GRCh38
rs587640119 Chr.1: 150554573-150554573 on Build GRCh38
rs150241051 Chr.1: 150554407-150554407 on Build GRCh38
rs764148151 Chr.1: 150554347-150554347 on Build GRCh38
rs780280898 Chr.1: 150554430-150554430 on Build GRCh38
rs766202192 Chr.1: 150554570-150554570 on Build GRCh38
rs770204797 Chr.1: 150554471-150554471 on Build GRCh38
rs768429793 Chr.1: 150554640-150554640 on Build GRCh38
rs757184334 Chr.1: 150554355-150554355 on Build GRCh38
rs756169149 Chr.1: 150554598-150554598 on Build GRCh38
rs936943617 Chr.1: 150554371-150554371 on Build GRCh38
rs745659924 Chr.1: 150554358-150554358 on Build GRCh38
rs1003823157 Chr.1: 150554296-150554296 on Build GRCh38
rs138209983 Chr.1: 150554305-150554305 on Build GRCh38
rs1048529390 Chr.1: 150554493-150554493 on Build GRCh38
rs587686142 Chr.1: 150554420-150554420 on Build GRCh38
rs995155497 Chr.1: 150554694-150554694 on Build GRCh38
rs776069420 Chr.1: 150554523-150554523 on Build GRCh38
rs995446810 Chr.1: 150554472-150554472 on Build GRCh38
rs757197289 Chr.1: 150554410-150554410 on Build GRCh38
rs781299982 Chr.1: 150554461-150554461 on Build GRCh38
rs376066523 Chr.1: 150554441-150554441 on Build GRCh38
rs978460115 Chr.1: 150554548-150554548 on Build GRCh38
rs771243690 Chr.1: 150554387-150554388 on Build GRCh38
rs775105543 Chr.1: 150554419-150554419 on Build GRCh38
rs770313672 Chr.1: 150554380-150554380 on Build GRCh38
rs151226442 Chr.1: 150554374-150554374 on Build GRCh38
rs185118272 Chr.1: 150554583-150554583 on Build GRCh38
rs755162489 Chr.1: 150554455-150554455 on Build GRCh38
rs758617597 Chr.1: 150554360-150554360 on Build GRCh38
rs368855050 Chr.1: 150554499-150554499 on Build GRCh38
rs775233529 Chr.1: 150554396-150554396 on Build GRCh38
rs750330260 Chr.1: 150554542-150554542 on Build GRCh38
rs367543737 Chr.1: 150554361-150554361 on Build GRCh38
rs765103386 Chr.1: 150554539-150554539 on Build GRCh38
rs749402444 Chr.1: 150554369-150554370 on Build GRCh38
rs986240919 Chr.1: 150554720-150554720 on Build GRCh38
rs745985357 Chr.1: 150554594-150554595 on Build GRCh38
rs1002582786 Chr.1: 150554298-150554298 on Build GRCh38
rs778620331 Chr.1: 150554356-150554356 on Build GRCh38
rs779474970 Chr.1: 150554637-150554637 on Build GRCh38
rs1046608087 Chr.1: 150554625-150554625 on Build GRCh38
rs767290674 Chr.1: 150554395-150554395 on Build GRCh38
rs1015918624 Chr.1: 150554305-150554309 on Build GRCh38
rs771381358 Chr.1: 150554491-150554491 on Build GRCh38
rs763308439 Chr.1: 150554392-150554392 on Build GRCh38
rs778174725 Chr.1: 150554600-150554600 on Build GRCh38
rs113553233 Chr.1: 150554584-150554584 on Build GRCh38
rs12095798 Chr.1: 150554633-150554633 on Build GRCh38
rs749636083 Chr.1: 150554486-150554486 on Build GRCh38
rs587603507 Chr.1: 150554654-150554654 on Build GRCh38
rs587646093 Chr.1: 150554663-150554665 on Build GRCh38
rs763822682 Chr.1: 150554515-150554515 on Build GRCh38
rs906984629 Chr.1: 150554393-150554393 on Build GRCh38

View Full Product Details