Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 150554954-150555496 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTSL4
Entrez Gene ID
54507
Gene Name
ADAMTS like 4
Gene Aliases
ADAMTSL-4, ECTOL2, TSRC1
UniGene
Hs.516243
Gene Chromosome Location
Chr.1: 150549369-150560937 on Build GRCh38
Transcripts
NM_001288607.1 NM_001288608.1 NM_019032.5 NM_025008.4 XM_011509644.2 XM_011509645.2 XM_011509648.2 XM_011509649.2 XM_011509650.2 XM_011509651.2 XM_011509652.1 XM_017001506.1 XM_017001507.1

Gene Symbol
LOC100289061
Entrez Gene ID
100289061
Gene Name
uncharacterized LOC100289061
Gene Aliases
-
UniGene
Hs.677561
Gene Chromosome Location
Chr.1: 150548564-150556700 on Build GRCh38
Transcripts
NM_001288607.1 NM_001288608.1 NM_019032.5 NM_025008.4 XM_011509644.2 XM_011509645.2 XM_011509648.2 XM_011509649.2 XM_011509650.2 XM_011509651.2 XM_011509652.1 XM_017001506.1 XM_017001507.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6562289 c.1311C>T p.G437G Chr.1: 150555436-150555436 on Build GRCh38
4524846 c.1295G>A p.R432H Chr.1: 150555489-150555489 on Build GRCh38
6594550 c.1284C>T p.R428R Chr.1: 150555478-150555478 on Build GRCh38
4771834 c.1363C>T p.R455C Chr.1: 150555488-150555488 on Build GRCh38
6259434 c.1283G>T p.R428L Chr.1: 150555477-150555477 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1031075714 Chr.1: 150555235-150555235 on Build GRCh38
rs113869234 Chr.1: 150555268-150555268 on Build GRCh38
rs751424524 Chr.1: 150555417-150555417 on Build GRCh38
rs910901867 Chr.1: 150555404-150555404 on Build GRCh38
rs200905514 Chr.1: 150555461-150555461 on Build GRCh38
rs753846444 Chr.1: 150555456-150555456 on Build GRCh38
rs752923405 Chr.1: 150555443-150555443 on Build GRCh38
rs972677770 Chr.1: 150555315-150555315 on Build GRCh38
rs370209728 Chr.1: 150555391-150555391 on Build GRCh38
rs180670920 Chr.1: 150555349-150555349 on Build GRCh38
rs587680953 Chr.1: 150555002-150555002 on Build GRCh38
rs776094670 Chr.1: 150555459-150555461 on Build GRCh38
rs112255887 Chr.1: 150555053-150555053 on Build GRCh38
rs111465316 Chr.1: 150555246-150555246 on Build GRCh38
rs910057660 Chr.1: 150555210-150555210 on Build GRCh38
rs185549053 Chr.1: 150555444-150555444 on Build GRCh38
rs761255647 Chr.1: 150555378-150555378 on Build GRCh38
rs771126937 Chr.1: 150554991-150554991 on Build GRCh38
rs747879178 Chr.1: 150555476-150555476 on Build GRCh38
rs1041249171 Chr.1: 150555178-150555178 on Build GRCh38
rs767653546 Chr.1: 150555434-150555434 on Build GRCh38
rs112312596 Chr.1: 150555239-150555239 on Build GRCh38
rs587608615 Chr.1: 150555097-150555097 on Build GRCh38
rs868090460 Chr.1: 150555102-150555102 on Build GRCh38
rs762928526 Chr.1: 150555408-150555408 on Build GRCh38
rs764869132 Chr.1: 150555146-150555146 on Build GRCh38
rs1007318345 Chr.1: 150555052-150555052 on Build GRCh38
rs370381933 Chr.1: 150555462-150555462 on Build GRCh38
rs111428433 Chr.1: 150555153-150555153 on Build GRCh38
rs892421777 Chr.1: 150555284-150555284 on Build GRCh38
rs889662689 Chr.1: 150555115-150555115 on Build GRCh38
rs587724675 Chr.1: 150555222-150555222 on Build GRCh38
rs587646671 Chr.1: 150555192-150555192 on Build GRCh38
rs757903395 Chr.1: 150555459-150555459 on Build GRCh38
rs368613246 Chr.1: 150555309-150555309 on Build GRCh38
rs780743473 Chr.1: 150555467-150555467 on Build GRCh38
rs1025932741 Chr.1: 150555147-150555147 on Build GRCh38
rs901124929 Chr.1: 150555453-150555453 on Build GRCh38
rs984247104 Chr.1: 150555191-150555191 on Build GRCh38
rs1007699595 Chr.1: 150555343-150555343 on Build GRCh38
rs948218830 Chr.1: 150555006-150555006 on Build GRCh38
rs993214415 Chr.1: 150555369-150555369 on Build GRCh38
rs375860019 Chr.1: 150555424-150555424 on Build GRCh38
rs948104477 Chr.1: 150555221-150555221 on Build GRCh38
rs587743882 Chr.1: 150555353-150555353 on Build GRCh38
rs1023658383 Chr.1: 150555263-150555263 on Build GRCh38
rs918372921 Chr.1: 150554985-150554987 on Build GRCh38
rs766291988 Chr.1: 150555416-150555416 on Build GRCh38
rs1025985332 Chr.1: 150555414-150555414 on Build GRCh38
rs370084822 Chr.1: 150555291-150555291 on Build GRCh38
rs772566618 Chr.1: 150555405-150555405 on Build GRCh38
rs891485383 Chr.1: 150555279-150555279 on Build GRCh38
rs916804525 Chr.1: 150554988-150554988 on Build GRCh38
rs930974621 Chr.1: 150555025-150555025 on Build GRCh38
rs189743555 Chr.1: 150555109-150555109 on Build GRCh38
rs1043507733 Chr.1: 150555301-150555301 on Build GRCh38

View Full Product Details