Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 150813033-150813530 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARNT
Entrez Gene ID
405
Gene Name
aryl hydrocarbon receptor nuclear translocator
Gene Aliases
HIF-1-beta, HIF-1beta, HIF1-beta, HIF1B, HIF1BETA, TANGO, bHLHe2
UniGene
Hs.632446
Gene Chromosome Location
Chr.1: 150809705-150876768 on Build GRCh38
Transcripts
NM_001197325.1 NM_001286035.1 NM_001286036.1 NM_001668.3 NM_178427.2 XM_005245151.1 XM_005245153.1 XM_005245154.3 XM_005245157.1 XM_011509542.2 XM_011509543.2 XM_011509544.2 XM_011509545.2 XM_011509546.1 XM_011509547.2 XM_017001288.1 XM_017001289.1 XM_017001290.1 XM_017001291.1 XM_017001292.1 XM_017001293.1 XM_017001294.1 XM_017001295.1 XM_017001296.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4968538 c.2251C>G p.Q751E Chr.1: 150813201-150813201 on Build GRCh38
6245289 c.2193C>A p.G731G Chr.1: 150813259-150813259 on Build GRCh38
7417977 c.2269G>T p.E757* Chr.1: 150813183-150813183 on Build GRCh38
4535713 c.2222G>A p.S741N Chr.1: 150813230-150813230 on Build GRCh38
6846675 c.2236C>G p.Q746E Chr.1: 150813216-150813216 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754795169 Chr.1: 150813364-150813364 on Build GRCh38
rs761276305 Chr.1: 150813315-150813315 on Build GRCh38
rs201586805 Chr.1: 150813338-150813338 on Build GRCh38
rs759158743 Chr.1: 150813080-150813080 on Build GRCh38
rs587599025 Chr.1: 150813428-150813428 on Build GRCh38
rs768879651 Chr.1: 150813143-150813143 on Build GRCh38
rs996069567 Chr.1: 150813071-150813071 on Build GRCh38
rs370493512 Chr.1: 150813288-150813288 on Build GRCh38
rs982837315 Chr.1: 150813248-150813248 on Build GRCh38
rs587733705 Chr.1: 150813299-150813299 on Build GRCh38
rs747257592 Chr.1: 150813129-150813129 on Build GRCh38
rs1027444624 Chr.1: 150813072-150813072 on Build GRCh38
rs1054492347 Chr.1: 150813337-150813337 on Build GRCh38
rs587704498 Chr.1: 150813179-150813179 on Build GRCh38
rs763745736 Chr.1: 150813229-150813229 on Build GRCh38
rs587623384 Chr.1: 150813196-150813196 on Build GRCh38
rs1007233409 Chr.1: 150813181-150813181 on Build GRCh38
rs773231217 Chr.1: 150813284-150813284 on Build GRCh38
rs866800266 Chr.1: 150813254-150813259 on Build GRCh38
rs775902596 Chr.1: 150813122-150813122 on Build GRCh38
rs765720006 Chr.1: 150813184-150813184 on Build GRCh38
rs756789458 Chr.1: 150813237-150813237 on Build GRCh38
rs587669786 Chr.1: 150813453-150813453 on Build GRCh38
rs748240537 Chr.1: 150813263-150813263 on Build GRCh38
rs763360757 Chr.1: 150813205-150813205 on Build GRCh38
rs995438061 Chr.1: 150813470-150813470 on Build GRCh38
rs759303852 Chr.1: 150813112-150813112 on Build GRCh38
rs183129107 Chr.1: 150813064-150813064 on Build GRCh38
rs587742698 Chr.1: 150813427-150813427 on Build GRCh38
rs587668426 Chr.1: 150813411-150813411 on Build GRCh38
rs748207091 Chr.1: 150813242-150813242 on Build GRCh38
rs587641102 Chr.1: 150813091-150813091 on Build GRCh38
rs945731876 Chr.1: 150813370-150813370 on Build GRCh38
rs764861223 Chr.1: 150813328-150813328 on Build GRCh38
rs758361425 Chr.1: 150813250-150813250 on Build GRCh38
rs2275237 Chr.1: 150813335-150813335 on Build GRCh38
rs374110573 Chr.1: 150813374-150813374 on Build GRCh38
rs763722704 Chr.1: 150813310-150813310 on Build GRCh38
rs892816049 Chr.1: 150813341-150813341 on Build GRCh38
rs758410825 Chr.1: 150813361-150813361 on Build GRCh38
rs894360513 Chr.1: 150813454-150813454 on Build GRCh38
rs587695675 Chr.1: 150813397-150813397 on Build GRCh38
rs756666812 Chr.1: 150813443-150813444 on Build GRCh38
rs754047400 Chr.1: 150813161-150813161 on Build GRCh38
rs776575226 Chr.1: 150813150-150813150 on Build GRCh38
rs73010816 Chr.1: 150813271-150813271 on Build GRCh38
rs771672355 Chr.1: 150813145-150813145 on Build GRCh38
rs1003107245 Chr.1: 150813395-150813395 on Build GRCh38
rs201828588 Chr.1: 150813211-150813211 on Build GRCh38
rs200025645 Chr.1: 150813259-150813259 on Build GRCh38
rs780025415 Chr.1: 150813255-150813255 on Build GRCh38
rs1002751282 Chr.1: 150813062-150813062 on Build GRCh38
rs753348131 Chr.1: 150813236-150813236 on Build GRCh38
rs753495822 Chr.1: 150813312-150813312 on Build GRCh38
rs751366135 Chr.1: 150813362-150813362 on Build GRCh38
rs745777019 Chr.1: 150813249-150813249 on Build GRCh38
rs766756351 Chr.1: 150813209-150813209 on Build GRCh38
rs373498149 Chr.1: 150813208-150813209 on Build GRCh38
rs768770073 Chr.1: 150813257-150813257 on Build GRCh38
rs761891026 Chr.1: 150813151-150813151 on Build GRCh38
rs769870604 Chr.1: 150813390-150813390 on Build GRCh38
rs746464139 Chr.1: 150813361-150813361 on Build GRCh38
rs374226509 Chr.1: 150813381-150813381 on Build GRCh38
rs779585971 Chr.1: 150813243-150813245 on Build GRCh38
rs765586486 Chr.1: 150813286-150813286 on Build GRCh38
rs774712704 Chr.1: 150813295-150813295 on Build GRCh38
rs186737810 Chr.1: 150813078-150813078 on Build GRCh38
rs951846297 Chr.1: 150813089-150813089 on Build GRCh38

View Full Product Details