Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 150984189-150984698 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANXA9
Entrez Gene ID
8416
Gene Name
annexin A9
Gene Aliases
ANX31
UniGene
Hs.3346
Gene Chromosome Location
Chr.1: 150982023-150995638 on Build GRCh38
Transcripts
NM_003568.2 XM_011510058.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4662175 c.361G>T p.E121* Chr.1: 150984374-150984374 on Build GRCh38
3473547 c.339C>T p.P113P Chr.1: 150984352-150984352 on Build GRCh38
6601347 c.272T>C p.L91P Chr.1: 150984285-150984285 on Build GRCh38
4629738 c.403G>A p.V135M Chr.1: 150984607-150984607 on Build GRCh38
2182864 c.402C>T p.D134D Chr.1: 150984606-150984606 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773735585 Chr.1: 150984607-150984607 on Build GRCh38
rs191544976 Chr.1: 150984314-150984314 on Build GRCh38
rs947143987 Chr.1: 150984483-150984483 on Build GRCh38
rs761956643 Chr.1: 150984631-150984631 on Build GRCh38
rs769503984 Chr.1: 150984285-150984285 on Build GRCh38
rs745346533 Chr.1: 150984613-150984613 on Build GRCh38
rs748876350 Chr.1: 150984605-150984605 on Build GRCh38
rs776969564 Chr.1: 150984628-150984628 on Build GRCh38
rs200970169 Chr.1: 150984640-150984640 on Build GRCh38
rs779516170 Chr.1: 150984354-150984354 on Build GRCh38
rs587734141 Chr.1: 150984548-150984548 on Build GRCh38
rs373255994 Chr.1: 150984435-150984435 on Build GRCh38
rs752104682 Chr.1: 150984643-150984643 on Build GRCh38
rs1030002345 Chr.1: 150984383-150984383 on Build GRCh38
rs753977252 Chr.1: 150984346-150984346 on Build GRCh38
rs766387608 Chr.1: 150984313-150984313 on Build GRCh38
rs764428573 Chr.1: 150984541-150984541 on Build GRCh38
rs899002930 Chr.1: 150984634-150984634 on Build GRCh38
rs750550955 Chr.1: 150984637-150984637 on Build GRCh38
rs888533905 Chr.1: 150984308-150984308 on Build GRCh38
rs746358214 Chr.1: 150984355-150984355 on Build GRCh38
rs775501751 Chr.1: 150984585-150984585 on Build GRCh38
rs200014201 Chr.1: 150984658-150984658 on Build GRCh38
rs749264529 Chr.1: 150984249-150984249 on Build GRCh38
rs1057046159 Chr.1: 150984220-150984220 on Build GRCh38
rs936221646 Chr.1: 150984521-150984521 on Build GRCh38
rs772809082 Chr.1: 150984299-150984299 on Build GRCh38
rs368693418 Chr.1: 150984433-150984433 on Build GRCh38
rs762318748 Chr.1: 150984312-150984312 on Build GRCh38
rs777546593 Chr.1: 150984602-150984602 on Build GRCh38
rs147958031 Chr.1: 150984585-150984585 on Build GRCh38
rs371333894 Chr.1: 150984519-150984519 on Build GRCh38
rs756241527 Chr.1: 150984231-150984231 on Build GRCh38
rs776494015 Chr.1: 150984269-150984269 on Build GRCh38
rs587665708 Chr.1: 150984623-150984623 on Build GRCh38
rs772536305 Chr.1: 150984253-150984253 on Build GRCh38
rs750461135 Chr.1: 150984564-150984564 on Build GRCh38
rs370721285 Chr.1: 150984551-150984551 on Build GRCh38
rs978600835 Chr.1: 150984507-150984507 on Build GRCh38
rs7536645 Chr.1: 150984353-150984353 on Build GRCh38
rs1057016658 Chr.1: 150984538-150984538 on Build GRCh38
rs752173761 Chr.1: 150984232-150984233 on Build GRCh38
rs774400960 Chr.1: 150984415-150984415 on Build GRCh38
rs987211308 Chr.1: 150984465-150984465 on Build GRCh38
rs759003848 Chr.1: 150984567-150984567 on Build GRCh38
rs777936603 Chr.1: 150984342-150984342 on Build GRCh38
rs767379819 Chr.1: 150984432-150984432 on Build GRCh38
rs888512909 Chr.1: 150984574-150984574 on Build GRCh38
rs267598009 Chr.1: 150984289-150984289 on Build GRCh38
rs763792434 Chr.1: 150984626-150984626 on Build GRCh38
rs587648021 Chr.1: 150984648-150984648 on Build GRCh38
rs746293734 Chr.1: 150984252-150984252 on Build GRCh38
rs1007203914 Chr.1: 150984661-150984661 on Build GRCh38
rs112049993 Chr.1: 150984407-150984407 on Build GRCh38
rs769592594 Chr.1: 150984400-150984400 on Build GRCh38
rs778455474 Chr.1: 150984337-150984337 on Build GRCh38
rs765434593 Chr.1: 150984632-150984632 on Build GRCh38
rs945841046 Chr.1: 150984479-150984479 on Build GRCh38
rs759390638 Chr.1: 150984331-150984331 on Build GRCh38
rs934569649 Chr.1: 150984528-150984528 on Build GRCh38
rs1016193351 Chr.1: 150984325-150984325 on Build GRCh38
rs895855091 Chr.1: 150984236-150984236 on Build GRCh38
rs963287330 Chr.1: 150984318-150984318 on Build GRCh38
rs756337765 Chr.1: 150984337-150984337 on Build GRCh38
rs761544489 Chr.1: 150984270-150984270 on Build GRCh38
rs747425403 Chr.1: 150984589-150984589 on Build GRCh38
rs762560905 Chr.1: 150984409-150984409 on Build GRCh38
rs587724810 Chr.1: 150984399-150984399 on Build GRCh38
rs770554982 Chr.1: 150984606-150984606 on Build GRCh38
rs150030788 Chr.1: 150984367-150984367 on Build GRCh38
rs373832843 Chr.1: 150984408-150984408 on Build GRCh38
rs747337432 Chr.1: 150984384-150984384 on Build GRCh38
rs377740400 Chr.1: 150984414-150984414 on Build GRCh38
rs759478610 Chr.1: 150984418-150984418 on Build GRCh38
rs1005107932 Chr.1: 150984213-150984213 on Build GRCh38
rs926160045 Chr.1: 150984518-150984518 on Build GRCh38
rs16832602 Chr.1: 150984368-150984368 on Build GRCh38
rs201687471 Chr.1: 150984348-150984348 on Build GRCh38
rs770540035 Chr.1: 150984244-150984245 on Build GRCh38
rs756894684 Chr.1: 150984662-150984662 on Build GRCh38
rs780106256 Chr.1: 150984491-150984491 on Build GRCh38
rs2305815 Chr.1: 150984471-150984471 on Build GRCh38
rs779242433 Chr.1: 150984251-150984251 on Build GRCh38
rs184419916 Chr.1: 150984558-150984558 on Build GRCh38
rs142083485 Chr.1: 150984652-150984652 on Build GRCh38
rs377474629 Chr.1: 150984550-150984550 on Build GRCh38
rs745330887 Chr.1: 150984345-150984346 on Build GRCh38
rs587753117 Chr.1: 150984250-150984250 on Build GRCh38
rs752428923 Chr.1: 150984332-150984332 on Build GRCh38
rs777848196 Chr.1: 150984247-150984247 on Build GRCh38
rs377306886 Chr.1: 150984600-150984600 on Build GRCh38
rs771623942 Chr.1: 150984560-150984561 on Build GRCh38

View Full Product Details