Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 150984476-150984883 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANXA9
Entrez Gene ID
8416
Gene Name
annexin A9
Gene Aliases
ANX31
UniGene
Hs.3346
Gene Chromosome Location
Chr.1: 150982023-150995638 on Build GRCh38
Transcripts
NM_003568.2 XM_011510058.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
895657 c.395C>T p.A132V Chr.1: 150984599-150984599 on Build GRCh38
4629738 c.403G>A p.V135M Chr.1: 150984607-150984607 on Build GRCh38
3704964 c.472+7A>G p.? Chr.1: 150984683-150984683 on Build GRCh38
5735629 c.406G>A p.A136T Chr.1: 150984610-150984610 on Build GRCh38
5594190 c.433C>T p.P145S Chr.1: 150984637-150984637 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773735585 Chr.1: 150984607-150984607 on Build GRCh38
rs761956643 Chr.1: 150984631-150984631 on Build GRCh38
rs745346533 Chr.1: 150984613-150984613 on Build GRCh38
rs748876350 Chr.1: 150984605-150984605 on Build GRCh38
rs950351609 Chr.1: 150984809-150984809 on Build GRCh38
rs776969564 Chr.1: 150984628-150984628 on Build GRCh38
rs200970169 Chr.1: 150984640-150984640 on Build GRCh38
rs1008962405 Chr.1: 150984681-150984681 on Build GRCh38
rs587734141 Chr.1: 150984548-150984548 on Build GRCh38
rs752104682 Chr.1: 150984643-150984643 on Build GRCh38
rs370841996 Chr.1: 150984710-150984710 on Build GRCh38
rs764428573 Chr.1: 150984541-150984541 on Build GRCh38
rs899002930 Chr.1: 150984634-150984634 on Build GRCh38
rs750550955 Chr.1: 150984637-150984637 on Build GRCh38
rs746935976 Chr.1: 150984716-150984716 on Build GRCh38
rs775501751 Chr.1: 150984585-150984585 on Build GRCh38
rs200014201 Chr.1: 150984658-150984658 on Build GRCh38
rs778582095 Chr.1: 150984677-150984677 on Build GRCh38
rs936221646 Chr.1: 150984521-150984521 on Build GRCh38
rs368882855 Chr.1: 150984732-150984732 on Build GRCh38
rs745436515 Chr.1: 150984697-150984697 on Build GRCh38
rs777546593 Chr.1: 150984602-150984602 on Build GRCh38
rs957552775 Chr.1: 150984756-150984756 on Build GRCh38
rs147958031 Chr.1: 150984585-150984585 on Build GRCh38
rs12128197 Chr.1: 150984798-150984798 on Build GRCh38
rs968483703 Chr.1: 150984702-150984702 on Build GRCh38
rs768583262 Chr.1: 150984724-150984724 on Build GRCh38
rs371333894 Chr.1: 150984519-150984519 on Build GRCh38
rs587665708 Chr.1: 150984623-150984623 on Build GRCh38
rs750461135 Chr.1: 150984564-150984564 on Build GRCh38
rs978600835 Chr.1: 150984507-150984507 on Build GRCh38
rs370721285 Chr.1: 150984551-150984551 on Build GRCh38
rs1057016658 Chr.1: 150984538-150984538 on Build GRCh38
rs977147958 Chr.1: 150984741-150984741 on Build GRCh38
rs759003848 Chr.1: 150984567-150984567 on Build GRCh38
rs888512909 Chr.1: 150984574-150984574 on Build GRCh38
rs763792434 Chr.1: 150984626-150984626 on Build GRCh38
rs587648021 Chr.1: 150984648-150984648 on Build GRCh38
rs1007203914 Chr.1: 150984661-150984661 on Build GRCh38
rs948662337 Chr.1: 150984843-150984843 on Build GRCh38
rs765434593 Chr.1: 150984632-150984632 on Build GRCh38
rs934569649 Chr.1: 150984528-150984528 on Build GRCh38
rs926082586 Chr.1: 150984744-150984744 on Build GRCh38
rs747425403 Chr.1: 150984589-150984589 on Build GRCh38
rs587698524 Chr.1: 150984749-150984749 on Build GRCh38
rs770554982 Chr.1: 150984606-150984606 on Build GRCh38
rs955833224 Chr.1: 150984757-150984757 on Build GRCh38
rs775473741 Chr.1: 150984715-150984715 on Build GRCh38
rs916132739 Chr.1: 150984764-150984764 on Build GRCh38
rs926160045 Chr.1: 150984518-150984518 on Build GRCh38
rs756894684 Chr.1: 150984662-150984662 on Build GRCh38
rs374119963 Chr.1: 150984834-150984834 on Build GRCh38
rs755543222 Chr.1: 150984805-150984805 on Build GRCh38
rs184419916 Chr.1: 150984558-150984558 on Build GRCh38
rs142083485 Chr.1: 150984652-150984652 on Build GRCh38
rs377474629 Chr.1: 150984550-150984550 on Build GRCh38
rs116722976 Chr.1: 150984816-150984816 on Build GRCh38
rs992625649 Chr.1: 150984814-150984814 on Build GRCh38
rs377306886 Chr.1: 150984600-150984600 on Build GRCh38
rs1017713133 Chr.1: 150984683-150984683 on Build GRCh38
rs771623942 Chr.1: 150984560-150984561 on Build GRCh38

View Full Product Details