Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 150988001-150988505 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANXA9
Entrez Gene ID
8416
Gene Name
annexin A9
Gene Aliases
ANX31
UniGene
Hs.3346
Gene Chromosome Location
Chr.1: 150982023-150995638 on Build GRCh38
Transcripts
NM_003568.2 XM_011510058.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5966080 c.787G>A p.G263S Chr.1: 150988180-150988180 on Build GRCh38
1333909 c.807G>T p.K269N Chr.1: 150988296-150988296 on Build GRCh38
1236714 c.754C>T p.R252C Chr.1: 150988147-150988147 on Build GRCh38
3418027 c.779C>T p.A260V Chr.1: 150988172-150988172 on Build GRCh38
3473554 c.804C>T p.I268I Chr.1: 150988293-150988293 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373118397 Chr.1: 150988087-150988087 on Build GRCh38
rs752855411 Chr.1: 150988188-150988188 on Build GRCh38
rs367612287 Chr.1: 150988150-150988150 on Build GRCh38
rs767764082 Chr.1: 150988186-150988186 on Build GRCh38
rs767846739 Chr.1: 150988218-150988221 on Build GRCh38
rs756277622 Chr.1: 150988201-150988201 on Build GRCh38
rs376305826 Chr.1: 150988108-150988108 on Build GRCh38
rs752059226 Chr.1: 150988200-150988200 on Build GRCh38
rs370979889 Chr.1: 150988109-150988109 on Build GRCh38
rs377065099 Chr.1: 150988368-150988368 on Build GRCh38
rs776402629 Chr.1: 150988155-150988155 on Build GRCh38
rs150067239 Chr.1: 150988286-150988286 on Build GRCh38
rs776311888 Chr.1: 150988095-150988095 on Build GRCh38
rs771415277 Chr.1: 150988148-150988148 on Build GRCh38
rs980961174 Chr.1: 150988462-150988475 on Build GRCh38
rs199810684 Chr.1: 150988304-150988304 on Build GRCh38
rs749748076 Chr.1: 150988147-150988147 on Build GRCh38
rs772258896 Chr.1: 150988407-150988407 on Build GRCh38
rs370252595 Chr.1: 150988154-150988154 on Build GRCh38
rs188413793 Chr.1: 150988026-150988026 on Build GRCh38
rs77936236 Chr.1: 150988169-150988169 on Build GRCh38
rs929410833 Chr.1: 150988475-150988475 on Build GRCh38
rs746284951 Chr.1: 150988256-150988256 on Build GRCh38
rs981047058 Chr.1: 150988444-150988444 on Build GRCh38
rs587751896 Chr.1: 150988180-150988180 on Build GRCh38
rs147533891 Chr.1: 150988137-150988137 on Build GRCh38
rs761180071 Chr.1: 150988096-150988096 on Build GRCh38
rs12033744 Chr.1: 150988452-150988452 on Build GRCh38
rs775300431 Chr.1: 150988342-150988342 on Build GRCh38
rs1046954806 Chr.1: 150988227-150988227 on Build GRCh38
rs966589406 Chr.1: 150988146-150988146 on Build GRCh38
rs376529122 Chr.1: 150988081-150988081 on Build GRCh38
rs900768179 Chr.1: 150988436-150988436 on Build GRCh38
rs587763804 Chr.1: 150988052-150988052 on Build GRCh38
rs888357901 Chr.1: 150988263-150988263 on Build GRCh38
rs755540101 Chr.1: 150988218-150988219 on Build GRCh38
rs369064246 Chr.1: 150988098-150988098 on Build GRCh38
rs765886376 Chr.1: 150988374-150988374 on Build GRCh38
rs188733335 Chr.1: 150988277-150988277 on Build GRCh38
rs1028463218 Chr.1: 150988102-150988102 on Build GRCh38
rs753885693 Chr.1: 150988365-150988365 on Build GRCh38
rs779513794 Chr.1: 150988246-150988246 on Build GRCh38
rs968668021 Chr.1: 150988441-150988441 on Build GRCh38
rs758806425 Chr.1: 150988259-150988259 on Build GRCh38
rs774149401 Chr.1: 150988179-150988179 on Build GRCh38
rs764340723 Chr.1: 150988355-150988355 on Build GRCh38
rs762802671 Chr.1: 150988172-150988172 on Build GRCh38
rs754670778 Chr.1: 150988077-150988077 on Build GRCh38
rs746195112 Chr.1: 150988152-150988152 on Build GRCh38
rs200588048 Chr.1: 150988341-150988341 on Build GRCh38
rs1040996582 Chr.1: 150988411-150988411 on Build GRCh38
rs374576176 Chr.1: 150988237-150988237 on Build GRCh38
rs199972366 Chr.1: 150988354-150988354 on Build GRCh38
rs587759130 Chr.1: 150988110-150988110 on Build GRCh38
rs766263552 Chr.1: 150988176-150988176 on Build GRCh38
rs781060123 Chr.1: 150988264-150988264 on Build GRCh38
rs766455618 Chr.1: 150988107-150988107 on Build GRCh38
rs764097530 Chr.1: 150988213-150988213 on Build GRCh38
rs893543369 Chr.1: 150988460-150988460 on Build GRCh38
rs760815613 Chr.1: 150988344-150988344 on Build GRCh38
rs770352593 Chr.1: 150988049-150988049 on Build GRCh38
rs761428752 Chr.1: 150988163-150988163 on Build GRCh38
rs996894731 Chr.1: 150988440-150988440 on Build GRCh38
rs755223923 Chr.1: 150988112-150988112 on Build GRCh38
rs373251195 Chr.1: 150988075-150988075 on Build GRCh38
rs374591781 Chr.1: 150988118-150988118 on Build GRCh38
rs919322462 Chr.1: 150988466-150988466 on Build GRCh38
rs748783160 Chr.1: 150988299-150988299 on Build GRCh38
rs759390463 Chr.1: 150988334-150988334 on Build GRCh38
rs778414328 Chr.1: 150988138-150988138 on Build GRCh38
rs149528939 Chr.1: 150988117-150988117 on Build GRCh38
rs370934834 Chr.1: 150988083-150988083 on Build GRCh38
rs949681498 Chr.1: 150988461-150988462 on Build GRCh38
rs750963108 Chr.1: 150988390-150988390 on Build GRCh38
rs778117590 Chr.1: 150988071-150988071 on Build GRCh38
rs753101833 Chr.1: 150988286-150988287 on Build GRCh38
rs372215070 Chr.1: 150988305-150988305 on Build GRCh38
rs587650739 Chr.1: 150988217-150988217 on Build GRCh38
rs886647329 Chr.1: 150988316-150988316 on Build GRCh38
rs769172343 Chr.1: 150988165-150988165 on Build GRCh38
rs754385534 Chr.1: 150988125-150988125 on Build GRCh38
rs587766028 Chr.1: 150988289-150988289 on Build GRCh38
rs143896675 Chr.1: 150988101-150988101 on Build GRCh38
rs753893020 Chr.1: 150988215-150988215 on Build GRCh38
rs869174401 Chr.1: 150988085-150988087 on Build GRCh38

View Full Product Details