Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 151090544-151091070 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABPB2
Entrez Gene ID
126626
Gene Name
GA binding protein transcription factor beta subunit 2
Gene Aliases
GABPB-2
UniGene
Hs.15671
Gene Chromosome Location
Chr.1: 151070180-151125545 on Build GRCh38
Transcripts
NM_001323906.1 NM_001323907.1 NM_001323908.1 NM_001323909.1 NM_001323910.1 NM_001323911.1 NM_001323912.1 NM_001323913.1 NM_144618.2 XM_017000246.1 XM_017000247.1 XM_017000248.1 XM_017000249.1 XM_017000250.1 XM_017000251.1 XM_017000252.1 XM_017000253.1 XM_017000254.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6297790 c.271G>T p.V91F Chr.1: 151090568-151090568 on Build GRCh38
5531554 c.258C>T p.I86I Chr.1: 151090555-151090555 on Build GRCh38
1207760 c.251C>T p.A84V Chr.1: 151090548-151090548 on Build GRCh38
7131598 c.275G>T p.R92L Chr.1: 151090572-151090572 on Build GRCh38
6668508 c.275G>A p.R92Q Chr.1: 151090572-151090572 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs587657524 Chr.1: 151091037-151091037 on Build GRCh38
rs1029207340 Chr.1: 151090950-151090950 on Build GRCh38
rs79242128 Chr.1: 151090856-151090856 on Build GRCh38
rs369019929 Chr.1: 151091025-151091025 on Build GRCh38
rs369938894 Chr.1: 151090725-151090725 on Build GRCh38
rs922272715 Chr.1: 151090589-151090589 on Build GRCh38
rs181447329 Chr.1: 151090797-151090797 on Build GRCh38
rs780736795 Chr.1: 151090565-151090565 on Build GRCh38
rs764517811 Chr.1: 151090911-151090911 on Build GRCh38
rs1023552056 Chr.1: 151090884-151090884 on Build GRCh38
rs879481702 Chr.1: 151091034-151091034 on Build GRCh38
rs770495025 Chr.1: 151090580-151090580 on Build GRCh38
rs758503026 Chr.1: 151090564-151090564 on Build GRCh38
rs12093562 Chr.1: 151091044-151091044 on Build GRCh38
rs1004953701 Chr.1: 151090929-151090929 on Build GRCh38
rs115161931 Chr.1: 151090823-151090823 on Build GRCh38
rs998214994 Chr.1: 151090947-151090947 on Build GRCh38
rs372788530 Chr.1: 151090613-151090613 on Build GRCh38
rs982033790 Chr.1: 151090924-151090924 on Build GRCh38
rs190635850 Chr.1: 151090618-151090618 on Build GRCh38
rs587671295 Chr.1: 151090906-151090906 on Build GRCh38
rs1056898872 Chr.1: 151090635-151090635 on Build GRCh38
rs780752718 Chr.1: 151090600-151090602 on Build GRCh38
rs139972452 Chr.1: 151090620-151090620 on Build GRCh38
rs587597251 Chr.1: 151090858-151090858 on Build GRCh38
rs1033108818 Chr.1: 151090868-151090868 on Build GRCh38
rs923823946 Chr.1: 151090624-151090624 on Build GRCh38
rs186466848 Chr.1: 151090912-151090912 on Build GRCh38
rs755421541 Chr.1: 151090572-151090572 on Build GRCh38
rs948001933 Chr.1: 151090881-151090881 on Build GRCh38
rs939301132 Chr.1: 151090863-151090863 on Build GRCh38
rs760608061 Chr.1: 151090571-151090571 on Build GRCh38
rs902001256 Chr.1: 151090931-151090931 on Build GRCh38
rs1050619422 Chr.1: 151090781-151090781 on Build GRCh38
rs587722154 Chr.1: 151090954-151090954 on Build GRCh38
rs776146548 Chr.1: 151090704-151090704 on Build GRCh38
rs1005027016 Chr.1: 151090728-151090728 on Build GRCh38
rs953711800 Chr.1: 151090765-151090765 on Build GRCh38
rs796116090 Chr.1: 151091013-151091020 on Build GRCh38
rs758970578 Chr.1: 151090760-151090760 on Build GRCh38
rs1008847645 Chr.1: 151090826-151090826 on Build GRCh38
rs1029776488 Chr.1: 151090764-151090764 on Build GRCh38
rs760617296 Chr.1: 151090570-151090570 on Build GRCh38
rs77350702 Chr.1: 151090966-151090966 on Build GRCh38
rs972212233 Chr.1: 151090997-151091001 on Build GRCh38
rs895142132 Chr.1: 151090678-151090678 on Build GRCh38
rs752903621 Chr.1: 151091044-151091045 on Build GRCh38
rs781398983 Chr.1: 151090574-151090574 on Build GRCh38
rs752449993 Chr.1: 151090871-151090871 on Build GRCh38
rs371156760 Chr.1: 151090591-151090591 on Build GRCh38
rs749036723 Chr.1: 151090579-151090579 on Build GRCh38
rs587637497 Chr.1: 151091045-151091045 on Build GRCh38
rs377582076 Chr.1: 151091012-151091013 on Build GRCh38
rs1015196121 Chr.1: 151090729-151090729 on Build GRCh38
rs997765580 Chr.1: 151090759-151090759 on Build GRCh38
rs966718022 Chr.1: 151090736-151090736 on Build GRCh38
rs967866526 Chr.1: 151090824-151090824 on Build GRCh38
rs772439762 Chr.1: 151090587-151090587 on Build GRCh38
rs587679799 Chr.1: 151090849-151090849 on Build GRCh38
rs760348465 Chr.1: 151091000-151091002 on Build GRCh38
rs763965098 Chr.1: 151090996-151090997 on Build GRCh38
rs796402961 Chr.1: 151090848-151090848 on Build GRCh38
rs910730245 Chr.1: 151091036-151091037 on Build GRCh38
rs939265835 Chr.1: 151090625-151090625 on Build GRCh38
rs762996067 Chr.1: 151090732-151090732 on Build GRCh38
rs112572180 Chr.1: 151091033-151091034 on Build GRCh38
rs887953508 Chr.1: 151090727-151090727 on Build GRCh38
rs930804003 Chr.1: 151091022-151091025 on Build GRCh38
rs1050588799 Chr.1: 151091034-151091038 on Build GRCh38
rs142196255 Chr.1: 151090969-151090969 on Build GRCh38
rs752850756 Chr.1: 151090880-151090880 on Build GRCh38
rs1036965328 Chr.1: 151090930-151090930 on Build GRCh38
rs1018870471 Chr.1: 151090852-151090852 on Build GRCh38
rs916528022 Chr.1: 151090611-151090611 on Build GRCh38
rs200618832 Chr.1: 151090596-151090596 on Build GRCh38
rs775582301 Chr.1: 151090789-151090789 on Build GRCh38

View Full Product Details