Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 151117080-151117586 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABPB2
Entrez Gene ID
126626
Gene Name
GA binding protein transcription factor beta subunit 2
Gene Aliases
GABPB-2
UniGene
Hs.15671
Gene Chromosome Location
Chr.1: 151070180-151125545 on Build GRCh38
Transcripts
NM_001323906.1 NM_001323907.1 NM_001323908.1 NM_001323909.1 NM_001323910.1 NM_001323911.1 NM_001323912.1 NM_001323913.1 NM_144618.2 XM_017000246.1 XM_017000247.1 XM_017000248.1 XM_017000249.1 XM_017000250.1 XM_017000251.1 XM_017000252.1 XM_017000253.1 XM_017000254.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7238677 c.952G>T p.E318* Chr.1: 151117421-151117421 on Build GRCh38
895699 c.1002G>T p.K334N Chr.1: 151117471-151117471 on Build GRCh38
7242147 c.961G>T p.V321L Chr.1: 151117430-151117430 on Build GRCh38
6009478 c.1031G>T p.S344I Chr.1: 151117500-151117500 on Build GRCh38
4980047 c.959C>T p.T320I Chr.1: 151117428-151117428 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs186006651 Chr.1: 151117337-151117337 on Build GRCh38
rs758977182 Chr.1: 151117527-151117527 on Build GRCh38
rs1019789617 Chr.1: 151117121-151117121 on Build GRCh38
rs1036024130 Chr.1: 151117174-151117174 on Build GRCh38
rs767279838 Chr.1: 151117365-151117365 on Build GRCh38
rs1048093229 Chr.1: 151117426-151117426 on Build GRCh38
rs201733299 Chr.1: 151117514-151117514 on Build GRCh38
rs958141304 Chr.1: 151117168-151117168 on Build GRCh38
rs6656450 Chr.1: 151117126-151117126 on Build GRCh38
rs772601366 Chr.1: 151117342-151117342 on Build GRCh38
rs931445855 Chr.1: 151117106-151117106 on Build GRCh38
rs753023577 Chr.1: 151117370-151117370 on Build GRCh38
rs759368024 Chr.1: 151117352-151117352 on Build GRCh38
rs759458053 Chr.1: 151117448-151117448 on Build GRCh38
rs771971461 Chr.1: 151117449-151117449 on Build GRCh38
rs940350373 Chr.1: 151117142-151117142 on Build GRCh38
rs772872073 Chr.1: 151117415-151117415 on Build GRCh38
rs764314049 Chr.1: 151117383-151117383 on Build GRCh38
rs587707948 Chr.1: 151117304-151117304 on Build GRCh38
rs757976779 Chr.1: 151117127-151117127 on Build GRCh38
rs764249137 Chr.1: 151117491-151117491 on Build GRCh38
rs751892423 Chr.1: 151117530-151117530 on Build GRCh38
rs770398317 Chr.1: 151117429-151117429 on Build GRCh38
rs769344127 Chr.1: 151117414-151117414 on Build GRCh38
rs908757994 Chr.1: 151117135-151117135 on Build GRCh38
rs762030686 Chr.1: 151117512-151117512 on Build GRCh38
rs756288440 Chr.1: 151117381-151117381 on Build GRCh38
rs71624522 Chr.1: 151117187-151117187 on Build GRCh38
rs201724065 Chr.1: 151117404-151117404 on Build GRCh38
rs780551976 Chr.1: 151117528-151117528 on Build GRCh38
rs59454556 Chr.1: 151117412-151117412 on Build GRCh38
rs775155410 Chr.1: 151117456-151117456 on Build GRCh38
rs747360509 Chr.1: 151117280-151117280 on Build GRCh38
rs368694486 Chr.1: 151117556-151117556 on Build GRCh38
rs1031449645 Chr.1: 151117137-151117137 on Build GRCh38
rs757367578 Chr.1: 151117393-151117393 on Build GRCh38
rs780275573 Chr.1: 151117410-151117410 on Build GRCh38
rs768103162 Chr.1: 151117454-151117454 on Build GRCh38
rs764248288 Chr.1: 151117442-151117444 on Build GRCh38
rs997259328 Chr.1: 151117215-151117215 on Build GRCh38
rs376385809 Chr.1: 151117511-151117511 on Build GRCh38
rs753819888 Chr.1: 151117493-151117496 on Build GRCh38
rs879485558 Chr.1: 151117331-151117331 on Build GRCh38
rs587658501 Chr.1: 151117289-151117289 on Build GRCh38
rs761037443 Chr.1: 151117429-151117430 on Build GRCh38
rs757378748 Chr.1: 151117542-151117543 on Build GRCh38
rs777332146 Chr.1: 151117552-151117552 on Build GRCh38
rs587663865 Chr.1: 151117460-151117460 on Build GRCh38
rs755329765 Chr.1: 151117542-151117542 on Build GRCh38
rs146084612 Chr.1: 151117427-151117427 on Build GRCh38
rs762466886 Chr.1: 151117345-151117345 on Build GRCh38
rs766326784 Chr.1: 151117349-151117349 on Build GRCh38
rs587750547 Chr.1: 151117267-151117267 on Build GRCh38
rs778393017 Chr.1: 151117561-151117561 on Build GRCh38
rs77696778 Chr.1: 151117525-151117525 on Build GRCh38
rs371833591 Chr.1: 151117430-151117430 on Build GRCh38
rs888964422 Chr.1: 151117291-151117291 on Build GRCh38
rs999296444 Chr.1: 151117351-151117351 on Build GRCh38
rs753881431 Chr.1: 151117389-151117389 on Build GRCh38
rs1011333418 Chr.1: 151117303-151117303 on Build GRCh38
rs1050112884 Chr.1: 151117250-151117250 on Build GRCh38
rs62621412 Chr.1: 151117409-151117409 on Build GRCh38
rs752757873 Chr.1: 151117234-151117234 on Build GRCh38

View Full Product Details