Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 151519217-151519724 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CGN
Entrez Gene ID
57530
Gene Name
cingulin
Gene Aliases
-
UniGene
Hs.591464
Gene Chromosome Location
Chr.1: 151510859-151538692 on Build GRCh38
Transcripts
NM_020770.2 XM_005245365.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1688532 c.776C>T p.P259L Chr.1: 151519295-151519295 on Build GRCh38
6894849 c.705G>C p.W235C Chr.1: 151519224-151519224 on Build GRCh38
2183699 c.852A>T p.A284A Chr.1: 151519371-151519371 on Build GRCh38
5843618 c.714C>A p.S238R Chr.1: 151519233-151519233 on Build GRCh38
895858 c.709T>G p.S237A Chr.1: 151519228-151519228 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754582096 Chr.1: 151519259-151519259 on Build GRCh38
rs777933169 Chr.1: 151519281-151519281 on Build GRCh38
rs766204654 Chr.1: 151519367-151519367 on Build GRCh38
rs1007508358 Chr.1: 151519648-151519648 on Build GRCh38
rs777540738 Chr.1: 151519387-151519387 on Build GRCh38
rs72997943 Chr.1: 151519674-151519674 on Build GRCh38
rs767192392 Chr.1: 151519361-151519361 on Build GRCh38
rs368023619 Chr.1: 151519313-151519313 on Build GRCh38
rs764377450 Chr.1: 151519358-151519358 on Build GRCh38
rs918251857 Chr.1: 151519540-151519540 on Build GRCh38
rs190731321 Chr.1: 151519653-151519653 on Build GRCh38
rs745918732 Chr.1: 151519319-151519319 on Build GRCh38
rs999558694 Chr.1: 151519359-151519359 on Build GRCh38
rs181234406 Chr.1: 151519405-151519405 on Build GRCh38
rs552258366 Chr.1: 151519244-151519244 on Build GRCh38
rs746328738 Chr.1: 151519432-151519432 on Build GRCh38
rs752262021 Chr.1: 151519382-151519382 on Build GRCh38
rs148740134 Chr.1: 151519312-151519312 on Build GRCh38
rs201140925 Chr.1: 151519381-151519381 on Build GRCh38
rs140720174 Chr.1: 151519360-151519360 on Build GRCh38
rs754995870 Chr.1: 151519372-151519372 on Build GRCh38
rs901222052 Chr.1: 151519668-151519668 on Build GRCh38
rs145327068 Chr.1: 151519263-151519263 on Build GRCh38
rs755806371 Chr.1: 151519276-151519276 on Build GRCh38
rs747755927 Chr.1: 151519273-151519273 on Build GRCh38
rs779284563 Chr.1: 151519411-151519411 on Build GRCh38
rs575514202 Chr.1: 151519318-151519318 on Build GRCh38
rs988166498 Chr.1: 151519415-151519415 on Build GRCh38
rs751585906 Chr.1: 151519368-151519368 on Build GRCh38
rs938341943 Chr.1: 151519422-151519422 on Build GRCh38
rs186698061 Chr.1: 151519564-151519564 on Build GRCh38
rs369915954 Chr.1: 151519401-151519401 on Build GRCh38
rs546259060 Chr.1: 151519472-151519472 on Build GRCh38
rs763371627 Chr.1: 151519357-151519377 on Build GRCh38
rs146676976 Chr.1: 151519384-151519384 on Build GRCh38
rs369954904 Chr.1: 151519347-151519347 on Build GRCh38
rs942158111 Chr.1: 151519652-151519652 on Build GRCh38
rs953401847 Chr.1: 151519459-151519459 on Build GRCh38
rs955942744 Chr.1: 151519365-151519365 on Build GRCh38
rs375613032 Chr.1: 151519241-151519241 on Build GRCh38
rs762863722 Chr.1: 151519364-151519364 on Build GRCh38
rs371662224 Chr.1: 151519592-151519592 on Build GRCh38
rs889941811 Chr.1: 151519622-151519622 on Build GRCh38
rs775565862 Chr.1: 151519349-151519349 on Build GRCh38
rs1001329438 Chr.1: 151519280-151519280 on Build GRCh38
rs772103037 Chr.1: 151519342-151519342 on Build GRCh38
rs531029255 Chr.1: 151519683-151519683 on Build GRCh38
rs926813212 Chr.1: 151519615-151519615 on Build GRCh38
rs796177258 Chr.1: 151519696-151519696 on Build GRCh38
rs993859800 Chr.1: 151519699-151519699 on Build GRCh38
rs1041140734 Chr.1: 151519659-151519659 on Build GRCh38
rs949648244 Chr.1: 151519466-151519466 on Build GRCh38

View Full Product Details