Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 180301400-180301906 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GFPT2
Entrez Gene ID
9945
Gene Name
glutamine-fructose-6-phosphate transaminase 2
Gene Aliases
GFAT, GFAT 2, GFAT2
UniGene
Hs.696497
Gene Chromosome Location
Chr.5: 180300690-180353387 on Build GRCh38
Transcripts
NM_005110.3 XM_006714942.2 XM_017010100.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3919832 c.2016C>T p.P672P Chr.5: 180301597-180301597 on Build GRCh38
1066778 c.2018G>T p.R673I Chr.5: 180301595-180301595 on Build GRCh38
4565362 c.2015_2016CC>TT p.P672L Chr.5: 180301597-180301598 on Build GRCh38
132692 c.2014C>T p.P672S Chr.5: 180301599-180301599 on Build GRCh38
3614846 c.2013C>T p.F671F Chr.5: 180301600-180301600 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs770284736 Chr.5: 180301616-180301616 on Build GRCh38
rs992416222 Chr.5: 180301711-180301711 on Build GRCh38
rs1043003233 Chr.5: 180301512-180301512 on Build GRCh38
rs918204380 Chr.5: 180301737-180301737 on Build GRCh38
rs369164869 Chr.5: 180301538-180301538 on Build GRCh38
rs184546685 Chr.5: 180301457-180301457 on Build GRCh38
rs998556735 Chr.5: 180301517-180301517 on Build GRCh38
rs373323205 Chr.5: 180301630-180301630 on Build GRCh38
rs1020702660 Chr.5: 180301662-180301662 on Build GRCh38
rs983615874 Chr.5: 180301780-180301780 on Build GRCh38
rs4700932 Chr.5: 180301860-180301860 on Build GRCh38
rs776903720 Chr.5: 180301429-180301429 on Build GRCh38
rs879652303 Chr.5: 180301799-180301800 on Build GRCh38
rs774109224 Chr.5: 180301628-180301628 on Build GRCh38
rs551731397 Chr.5: 180301553-180301553 on Build GRCh38
rs138920932 Chr.5: 180301458-180301458 on Build GRCh38
rs537515231 Chr.5: 180301587-180301587 on Build GRCh38
rs4700931 Chr.5: 180301707-180301707 on Build GRCh38
rs777112014 Chr.5: 180301652-180301652 on Build GRCh38
rs926943691 Chr.5: 180301828-180301828 on Build GRCh38
rs531822885 Chr.5: 180301449-180301449 on Build GRCh38
rs377354051 Chr.5: 180301833-180301833 on Build GRCh38
rs181393042 Chr.5: 180301643-180301643 on Build GRCh38
rs998441495 Chr.5: 180301874-180301874 on Build GRCh38
rs929500375 Chr.5: 180301529-180301529 on Build GRCh38
rs967803997 Chr.5: 180301664-180301664 on Build GRCh38
rs761131627 Chr.5: 180301514-180301514 on Build GRCh38
rs950896589 Chr.5: 180301769-180301769 on Build GRCh38
rs765803661 Chr.5: 180301659-180301659 on Build GRCh38
rs757667800 Chr.5: 180301822-180301822 on Build GRCh38
rs568549305 Chr.5: 180301554-180301554 on Build GRCh38
rs915423119 Chr.5: 180301845-180301845 on Build GRCh38
rs764536226 Chr.5: 180301521-180301521 on Build GRCh38
rs1011183567 Chr.5: 180301491-180301491 on Build GRCh38
rs142827230 Chr.5: 180301730-180301730 on Build GRCh38
rs202232625 Chr.5: 180301568-180301568 on Build GRCh38
rs762219108 Chr.5: 180301656-180301656 on Build GRCh38
rs769274690 Chr.5: 180301861-180301861 on Build GRCh38
rs754459383 Chr.5: 180301535-180301535 on Build GRCh38
rs942120730 Chr.5: 180301785-180301785 on Build GRCh38
rs764591640 Chr.5: 180301646-180301646 on Build GRCh38
rs979120463 Chr.5: 180301440-180301440 on Build GRCh38
rs568202568 Chr.5: 180301663-180301663 on Build GRCh38
rs376994904 Chr.5: 180301683-180301683 on Build GRCh38
rs1000510896 Chr.5: 180301677-180301677 on Build GRCh38
rs189057525 Chr.5: 180301552-180301552 on Build GRCh38
rs747483441 Chr.5: 180301781-180301781 on Build GRCh38
rs921927681 Chr.5: 180301801-180301801 on Build GRCh38
rs748135971 Chr.5: 180301569-180301569 on Build GRCh38
rs372543131 Chr.5: 180301546-180301546 on Build GRCh38
rs971094349 Chr.5: 180301733-180301733 on Build GRCh38
rs768375471 Chr.5: 180301638-180301638 on Build GRCh38
rs891065725 Chr.5: 180301837-180301837 on Build GRCh38
rs370042303 Chr.5: 180301557-180301557 on Build GRCh38
rs748832217 Chr.5: 180301600-180301600 on Build GRCh38
rs17080160 Chr.5: 180301849-180301849 on Build GRCh38
rs571043674 Chr.5: 180301547-180301547 on Build GRCh38
rs562918481 Chr.5: 180301428-180301428 on Build GRCh38
rs769794640 Chr.5: 180301579-180301579 on Build GRCh38
rs1038975695 Chr.5: 180301786-180301786 on Build GRCh38
rs1051003629 Chr.5: 180301835-180301835 on Build GRCh38
rs527882964 Chr.5: 180301509-180301509 on Build GRCh38
rs758368156 Chr.5: 180301549-180301549 on Build GRCh38
rs1009837464 Chr.5: 180301651-180301651 on Build GRCh38

View Full Product Details